• USA Home
  • Search Results

  • zirconium9122744067711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zirconium

356 Products
Synonym:   Zirconium element
Image of Zirconium
CAS Number:
Empirical Formula (Hill Notation):
 Zr
Molecular Weight:
 91.22
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Additive
  Weight
  Package quantity
  Particle Dimension
  Assay
  Physical Form
  Composite Dimension
Show More

Product #
Description
403296 Zirconium, powder, −100 mesh,
267651 Zirconium, sponge, ≥99% trace metals basis,
419141 Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis,
267724 Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis,
GF84779612 Zirconium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%,
GF53330172 Zirconium, tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%,
GF56980165 Zirconium, rod, 1000mm, diameter 12.7mm, centerless ground, 99.2%,
GF47938821 Zirconium, powder, max. particle size 45 micron, weight 100 g, purity 99.2%,
GF32665002 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 59mm,
756385 Zirconium, powder, in water, -325 mesh, 99.5% trace metals basis (excluding Hf),
GF03685316 Zirconium, foil, 100x100mm, thickness 0.1mm, annealed, 99.2%,
GF86559384 Zirconium, foil, 1m coil, thickness 0.05mm, annealed, 99.2%,
GF69451487 Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%,
GF22277527 Zirconium, wire, 1.0 mm diameter, length 1 m, purity 99.2%,
GF50488577 Zirconium, foil, 100x100mm, thickness 0.5mm, annealed, 99.2%,
GF11484468 Zirconium, rod, 1000mm, diameter 9.5mm, centerless ground, 99.2%,
GF36220227 Zirconium, rod, 1000mm, diameter 5.0mm, centerless ground, 99.2%,
GF50035137 Zirconium, foil, 5m coil, thickness 0.025mm, annealed, 99.2%,
GF83603909 Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%,
GF04513700 Zirconium, rod, 100mm, diameter 50mm, centerless ground, 99.2%,
GF98267417 Zirconium, wire, 1.0 mm diameter, length 20 m, purity 99.2%,
GF10742284 Zirconium, foil, 150x150mm, thickness 2.0mm, annealed, 99.2%,
GF74069837 Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%,
GF29806982 Zirconium, foil, light tested, 200x200mm, thickness 0.01mm, 99.2%,
GF63904989 Zirconium, foil, light tested, 100x100mm, thickness 0.025mm, annealed, 99.2%,
GF84948834 Zirconium, foil, 0.5m coil, thickness 0.075mm, annealed, 99.2%,
GF04668835 Zirconium, foil, 25x25mm, thickness 0.5mm, annealed, 99.2%,
GF30135002 Zirconium, sponge, 100g, max. size 25mm, 99.2%,
GF88851730 Zirconium, tube, 100mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%,
GF00342380 Zirconium, foil, thickness 1.0 mm, size 150 × 150 mm, purity 99.2%,
GF45997925 Zirconium, foil, light tested, 25x25mm, thickness 0.006mm, 99.2%,
GF27595637 Zirconium, rod, 100mm, diameter 6.0mm, centerless ground, 99.2%,
GF90874121 Zirconium, foil, 6mm disks, thickness 0.1mm, annealed, 99.2%,
GF43362660 Zirconium, foil, 1m coil, thickness 0.02mm, 99.2%,
GF11910743 Zirconium, wire, 1.0 mm diameter, length 0.5 m, purity 99.2%,
GF26818971 Zirconium, wire reel, 1m, diameter 0.5mm, as drawn, 99.2%,
GF34962082 Zirconium, wire, 1.0 mm diameter, length 5 m, purity 99.2%,
GF03886443 Zirconium, foil, 10mm disks, thickness 0.5mm, annealed, 99.2%,
GF99338743 Zirconium, rod, 200mm, diameter 2.0mm, centerless ground, 99.2%,
GF87053474 Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%,
GF43414556 Zirconium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.2%,
GF66876014 Zirconium, sponge, 200g, max. size 25mm, 99.2%,
GF68648158 Zirconium, foil, 300x300mm, thickness 0.125mm, annealed, 99.2%,
GF63994708 Zirconium, rod, 100mm, diameter 2.0mm, centerless ground, 99.2%,
GF17343266 Zirconium, rod, 500mm, diameter 2.0mm, centerless ground, 99.2%,
GF38412806 Zirconium, lump, 8 mm max. lump size, weight 20 g, purity 99.2%,
GF27528048 Zirconium, rod, 100mm, diameter 9.5mm, centerless ground, 99.2%,
GF47919875 Zirconium, foil, light tested, 50x50mm, thickness 0.05mm, annealed, 99.2%,
GF92588967 Zirconium, rod, 200mm, diameter 3.0mm, centerless ground, 99.2%,
GF13632410 Zirconium, foil, 25x25mm, thickness 2.0mm, annealed, 99.2%,
GF32349685 Zirconium, foil, light tested, 25x25mm, thickness 0.025mm, annealed, 99.2%,
GF91309035 Zirconium, foil, 25x25mm, thickness 0.1mm, annealed, 99.2%,
GF47333718 Zirconium, rod, 1000mm, diameter 1.5mm, 99.2%,
GF66272081 Zirconium, rod, 100mm, diameter 1.5mm, 99.2%,
GF29496426 Zirconium, foil, 25x25mm, thickness 0.25mm, annealed, 99.2%,
GF58729607 Zirconium, foil, 100x100mm, thickness 0.125mm, annealed, 99.2%,
GF27504402 Zirconium, foil, 300x300mm, thickness 0.5mm, annealed, 99.2%,
GF04051886 Zirconium, foil, 50x50mm, thickness 2.0mm, annealed, 99.2%,
GF14815593 Zirconium, foil, 10mm disks, thickness 0.0125mm, 99.2%,
GF58113019 Zirconium, foil, 10mm disks, thickness 0.015mm, 99.2%,
GF67870468 Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%,
GF58442070 Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%,
GF54823642 Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%,
GF75562869 Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%,
GF55287221 Zirconium, foil, 10mm disks, thickness 0.02mm, 99.2%,
GF38009044 Zirconium, foil, 10mm disks, thickness 0.05mm, annealed, 99.2%,
GF89139594 Zirconium, foil, 10mm disks, thickness 0.075mm, annealed, 99.2%,
GF22007595 Zirconium, foil, 10mm disks, thickness 0.125mm, annealed, 99.2%,
GF50038039 Zirconium, foil, 10mm disks, thickness 0.25mm, annealed, 99.2%,
GF09475027 Zirconium, foil, 15mm disks, thickness 0.009mm, 99.2%,
GF51948872 Zirconium, foil, 15mm disks, thickness 0.0125mm, 99.2%,
GF83808221 Zirconium, foil, 15mm disks, thickness 0.016mm, 99.2%,
GF05259135 Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%,
GF72812370 Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%,
GF45357688 Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%,
GF51130370 Zirconium, foil, 15mm disks, thickness 0.05mm, annealed, 99.2%,
GF42449249 Zirconium, foil, 15mm disks, thickness 0.075mm, annealed, 99.2%,
GF50949232 Zirconium, foil, 15mm disks, thickness 0.125mm, annealed, 99.2%,
GF24728890 Zirconium, foil, 15mm disks, thickness 0.1mm, annealed, 99.2%,
GF54754218 Zirconium, foil, 15mm disks, thickness 0.25mm, annealed, 99.2%,
GF36480506 Zirconium, foil, 1m coil, thickness 0.016mm, 99.2%,
GF16304004 Zirconium, foil, 1m coil, thickness 0.025mm, annealed, 99.2%,
GF61379275 Zirconium, foil, 1m coil, thickness 0.125mm, annealed, 99.2%,
GF88641311 Zirconium, foil, 1m coil, thickness 0.1mm, annealed, 99.2%,
GF46551551 Zirconium, foil, 1m coil, thickness 0.25mm, annealed, 99.2%,
GF84378426 Zirconium, foil, 25mm disks, thickness 0.009mm, 99.2%,
GF93609366 Zirconium, foil, 25mm disks, thickness 0.0125mm, 99.2%,
GF76027675 Zirconium, foil, 25mm disks, thickness 0.016mm, 99.2%,
GF69710856 Zirconium, foil, 25mm disks, thickness 0.025mm, annealed, 99.2%,
GF62866008 Zirconium, foil, 25mm disks, thickness 0.02mm, 99.2%,
GF91296513 Zirconium, foil, 25mm disks, thickness 0.05mm, annealed, 99.2%,
GF06155344 Zirconium, foil, 25mm disks, thickness 0.075mm, annealed, 99.2%,
GF81684917 Zirconium, foil, 100x100mm, thickness 0.25mm, annealed, 99.2%,
GF06485935 Zirconium, foil, 100x100mm, thickness 2.0mm, annealed, 99.2%,
GF41200726 Zirconium, foil, 150x150mm, thickness 0.125mm, annealed, 99.2%,
GF60664877 Zirconium, foil, 150x150mm, thickness 0.1mm, annealed, 99.2%,
GF10614170 Zirconium, foil, 150x150mm, thickness 0.25mm, annealed, 99.2%,
GF11071651 Zirconium, foil, 150x150mm, thickness 0.5mm, annealed, 99.2%,
GF83109135 Zirconium, foil, 200x300mm, thickness 0.1mm, annealed, 99.2%,
GF15792554 Zirconium, foil, 25x25mm, thickness 0.125mm, annealed, 99.2%,
GF21986747 Zirconium, foil, 25x25mm, thickness 0.2mm, annealed, 99.2%,
GF07434008 Zirconium, foil, 4mm disks, thickness 0.25mm, annealed, 99.2%,
GF13235666 Zirconium, foil, 4mm disks, thickness 0.5mm, annealed, 99.2%,
GF03458796 Zirconium, foil, 50mm disks, thickness 0.016mm, 99.2%,
GF79402068 Zirconium, foil, 50mm disks, thickness 0.01mm, 99.2%,
GF13856799 Zirconium, foil, 50mm disks, thickness 0.025mm, annealed, 99.2%,
GF58250099 Zirconium, foil, 50mm disks, thickness 0.02mm, 99.2%,
GF12064867 Zirconium, foil, 50mm disks, thickness 0.05mm, annealed, 99.2%,
GF69418389 Zirconium, foil, 50mm disks, thickness 0.075mm, annealed, 99.2%,
GF81775895 Zirconium, foil, 50mm disks, thickness 0.125mm, annealed, 99.2%,
GF88926340 Zirconium, foil, 50mm disks, thickness 0.1mm, annealed, 99.2%,
GF34608001 Zirconium, foil, 50mm disks, thickness 0.25mm, annealed, 99.2%,
GF20456456 Zirconium, foil, 50mm disks, thickness 0.5mm, annealed, 99.2%,
GF41562615 Zirconium, foil, 5m coil, thickness 0.02mm, 99.2%,
GF90323489 Zirconium, foil, 5m coil, thickness 0.05mm, annealed, 99.2%,
GF48905062 Zirconium, foil, 5m coil, thickness 0.075mm, annealed, 99.2%,
GF08814236 Zirconium, foil, 6mm disks, thickness 0.009mm, 99.2%,
GF06029748 Zirconium, foil, 6mm disks, thickness 0.0125mm, 99.2%,
GF56253315 Zirconium, foil, 6mm disks, thickness 0.015mm, 99.2%,
GF07959549 Zirconium, foil, 6mm disks, thickness 0.016mm, 99.2%,
GF12144541 Zirconium, foil, 6mm disks, thickness 0.01mm, 99.2%,
GF65118630 Zirconium, foil, 6mm disks, thickness 0.01mm, 99.2%,
GF76824030 Zirconium, foil, 6mm disks, thickness 0.025mm, annealed, 99.2%,
GF38685134 Zirconium, foil, 6mm disks, thickness 0.02mm, 99.2%,
GF03874439 Zirconium, foil, 6mm disks, thickness 0.05mm, annealed, 99.2%,
GF12434509 Zirconium, foil, 6mm disks, thickness 0.075mm, annealed, 99.2%,
GF56903801 Zirconium, foil, 6mm disks, thickness 0.125mm, annealed, 99.2%,
GF29251201 Zirconium, foil, 6mm disks, thickness 0.25mm, annealed, 99.2%,
GF54457847 Zirconium, foil, 6mm disks, thickness 0.5mm, annealed, 99.2%,
GF01779802 Zirconium, foil, 8mm disks, thickness 0.009mm, 99.2%,
GF53718521 Zirconium, foil, 8mm disks, thickness 0.0125mm, 99.2%,
GF31664909 Zirconium, foil, 8mm disks, thickness 0.015mm, 99.2%,
GF94942963 Zirconium, foil, 8mm disks, thickness 0.016mm, 99.2%,
GF18067530 Zirconium, foil, 8mm disks, thickness 0.01mm, 99.2%,
GF29532359 Zirconium, foil, 8mm disks, thickness 0.01mm, 99.2%,
GF03004502 Zirconium, foil, 8mm disks, thickness 0.025mm, annealed, 99.2%,
GF35642291 Zirconium, foil, 8mm disks, thickness 0.02mm, 99.2%,
GF84304076 Zirconium, foil, 8mm disks, thickness 0.05mm, annealed, 99.2%,
GF54313118 Zirconium, foil, 8mm disks, thickness 0.075mm, annealed, 99.2%,
GF12976061 Zirconium, foil, 8mm disks, thickness 0.125mm, annealed, 99.2%,
GF68654201 Zirconium, foil, 25mm disks, thickness 0.125mm, annealed, 99.2%,
GF91111418 Zirconium, foil, 25mm disks, thickness 0.1mm, annealed, 99.2%,
GF44276867 Zirconium, foil, 25mm disks, thickness 0.25mm, annealed, 99.2%,
GF25763654 Zirconium, foil, 25mm disks, thickness 0.5mm, annealed, 99.2%,
GF38000281 Zirconium, foil, 2m coil, thickness 0.025mm, annealed, 99.2%,
GF72792928 Zirconium, foil, 0.5m coil, thickness 0.016mm, 99.2%,
GF40397924 Zirconium, foil, 0.5m coil, thickness 0.01mm, 99.2%,
GF88608011 Zirconium, foil, 0.5m coil, thickness 0.025mm, annealed, 99.2%,
GF89479591 Zirconium, foil, 0.5m coil, thickness 0.05mm, annealed, 99.2%,
GF25617988 Zirconium, foil, 0.5m coil, thickness 0.125mm, annealed, 99.2%,
GF18604942 Zirconium, foil, 0.5m coil, thickness 0.1mm, annealed, 99.2%,
GF02248725 Zirconium, foil, 0.5m coil, thickness 0.25mm, annealed, 99.2%,
GF94637728 Zirconium, foil, 0.5m coil, thickness 0.5mm, annealed, 99.2%,
GF95326487 Zirconium, foil, 10m coil, thickness 0.02mm, 99.2%,
GF22475808 Zirconium, foil, 10mm disks, thickness 0.009mm, 99.2%,
GF20584043 Zirconium, foil, light tested, 150x150mm, thickness 0.008mm, 99.2%,
GF50594340 Zirconium, foil, light tested, 150x150mm, thickness 0.009mm, 99.2%,
GF74962525 Zirconium, foil, light tested, 150x150mm, thickness 0.0125mm, 99.2%,
GF88122641 Zirconium, foil, light tested, 150x150mm, thickness 0.025mm, annealed, 99.2%,
GF79439035 Zirconium, foil, light tested, 150x150mm, thickness 0.02mm, 99.2%,
GF48056733 Zirconium, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.2%,
GF59433161 Zirconium, foil, light tested, 150x150mm, thickness 0.075mm, annealed, 99.2%,
GF00880512 Zirconium, foil, light tested, 200x300mm, thickness 0.02mm, 99.2%,
GF29145887 Zirconium, foil, light tested, 200x300mm, thickness 0.075mm, annealed, 99.2%,
GF48392253 Zirconium, foil, light tested, 25x25mm, thickness 0.005mm, 99.2%,
GF05569454 Zirconium, foil, light tested, 25x25mm, thickness 0.007mm, 99.2%,
GF21608052 Zirconium, foil, light tested, 25x25mm, thickness 0.009mm, 99.2%,
GF77749161 Zirconium, foil, light tested, 25x25mm, thickness 0.0125mm, 99.2%,
GF48229409 Zirconium, foil, light tested, 25x25mm, thickness 0.015mm, 99.2%,
GF64127864 Zirconium, foil, light tested, 25x25mm, thickness 0.016mm, 99.2%,
GF27821639 Zirconium, foil, light tested, 25x25mm, thickness 0.01mm, 99.2%,
GF01998322 Zirconium, foil, light tested, 25x25mm, thickness 0.01mm, 99.2%,
GF18296679 Zirconium, foil, light tested, 25x25mm, thickness 0.02mm, 99.2%,
GF97183356 Zirconium, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.2%,
GF08107602 Zirconium, foil, light tested, 25x25mm, thickness 0.075mm, annealed, 99.2%,
GF50539091 Zirconium, foil, light tested, 300x300mm, thickness 0.05mm, annealed, 99.2%,
GF28760564 Zirconium, foil, light tested, 50x50mm, thickness 0.006mm, 99.2%,
GF85362891 Zirconium, foil, light tested, 50x50mm, thickness 0.007mm, 99.2%,
GF88691114 Zirconium, foil, light tested, 50x50mm, thickness 0.008mm, 99.2%,
GF11195164 Zirconium, foil, light tested, 50x50mm, thickness 0.009mm, 99.2%,
GF40011837 Zirconium, foil, light tested, 50x50mm, thickness 0.0125mm, 99.2%,
GF78470433 Zirconium, foil, light tested, 50x50mm, thickness 0.016mm, 99.2%,
GF51665430 Zirconium, foil, light tested, 50x50mm, thickness 0.01mm, 99.2%,
GF50933998 Zirconium, foil, light tested, 50x50mm, thickness 0.01mm, 99.2%,
GF43711231 Zirconium, foil, light tested, 50x50mm, thickness 0.025mm, annealed, 99.2%,
GF16780428 Zirconium, foil, light tested, 50x50mm, thickness 0.02mm, 99.2%,
GF71688831 Zirconium, foil, not light tested, 100x100mm, thickness 0.003mm, 99.2%,
GF29795169 Zirconium, foil, not light tested, 100x100mm, thickness 0.004mm, 99.2%,
GF97843539 Zirconium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.2%,
GF19291462 Zirconium, foil, not light tested, 100x100mm, thickness 0.006mm, 99.2%,
GF65680432 Zirconium, foil, not light tested, 150x150mm, thickness 0.006mm, 99.2%,
GF08011257 Zirconium, foil, not light tested, 150x150mm, thickness 0.008mm, 99.2%,
GF32255096 Zirconium, foil, not light tested, 150x150mm, thickness 0.009mm, 99.2%,
GF63967247 Zirconium, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.2%,
GF06322597 Zirconium, foil, not light tested, 200x300mm, thickness 0.02mm, 99.2%,
GF22950974 Zirconium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.2%,
GF09376768 Zirconium, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.2%,
GF20332505 Zirconium, foil, 2m coil, thickness 0.02mm, 99.2%,
GF19922586 Zirconium, foil, 2m coil, thickness 0.05mm, annealed, 99.2%,
GF04391243 Zirconium, foil, 2m coil, thickness 0.075mm, annealed, 99.2%,
GF80163884 Zirconium, foil, 2m coil, thickness 0.1mm, annealed, 99.2%,
GF69670514 Zirconium, foil, 4mm disks, thickness 0.009mm, 99.2%,
GF65648234 Zirconium, foil, 4mm disks, thickness 0.0125mm, 99.2%,
GF25250026 Zirconium, foil, 4mm disks, thickness 0.015mm, 99.2%,
GF16063679 Zirconium, foil, 4mm disks, thickness 0.016mm, 99.2%,
GF50497151 Zirconium, foil, 4mm disks, thickness 0.01mm, 99.2%,
GF98569412 Zirconium, foil, 4mm disks, thickness 0.01mm, 99.2%,
GF99014597 Zirconium, foil, 4mm disks, thickness 0.025mm, annealed, 99.2%,
GF76309487 Zirconium, foil, 4mm disks, thickness 0.02mm, 99.2%,
GF56830990 Zirconium, foil, 4mm disks, thickness 0.05mm, annealed, 99.2%,
GF41699470 Zirconium, foil, 4mm disks, thickness 0.075mm, annealed, 99.2%,
GF34855230 Zirconium, foil, 4mm disks, thickness 0.125mm, annealed, 99.2%,
GF60726999 Zirconium, foil, 4mm disks, thickness 0.1mm, annealed, 99.2%,
GF67908583 Zirconium, rod, 1000mm, diameter 2.0mm, centerless ground, 99.2%,
GF48459774 Zirconium, rod, 1000mm, diameter 3.0mm, centerless ground, 99.2%,
GF70124442 Zirconium, rod, 1000mm, diameter 6.0mm, centerless ground, 99.2%,
GF35558321 Zirconium, rod, 100mm, diameter 12.7mm, centerless ground, 99.2%,
GF15031662 Zirconium, rod, 100mm, diameter 19.0mm, centerless ground, 99.2%,
GF90246056 Zirconium, rod, 100mm, diameter 3.0mm, centerless ground, 99.2%,
GF88573618 Zirconium, rod, 100mm, diameter 5.0mm, centerless ground, 99.2%,
GF84010496 Zirconium, rod, 200mm, diameter 1.5mm, 99.2%,
GF44790846 Zirconium, rod, 200mm, diameter 12.7mm, centerless ground, 99.2%,
GF78958715 Zirconium, rod, 200mm, diameter 19.0mm, centerless ground, 99.2%,
GF71701512 Zirconium, rod, 200mm, diameter 5.0mm, centerless ground, 99.2%,
GF17709341 Zirconium, rod, 200mm, diameter 6.0mm, centerless ground, 99.2%,
GF41005538 Zirconium, rod, 200mm, diameter 9.5mm, centerless ground, 99.2%,
GF01311950 Zirconium, rod, 500mm, diameter 1.5mm, 99.2%,
GF98855847 Zirconium, rod, 500mm, diameter 12.7mm, centerless ground, 99.2%,
GF49023862 Zirconium, rod, 500mm, diameter 3.0mm, centerless ground, 99.2%,
GF62689764 Zirconium, rod, 500mm, diameter 5.0mm, centerless ground, 99.2%,
GF84412762 Zirconium, rod, 500mm, diameter 6.0mm, centerless ground, 99.2%,
GF25691878 Zirconium, rod, 500mm, diameter 9.5mm, centerless ground, 99.2%,
GF22520258 Zirconium, rod, 50mm, diameter 19.0mm, centerless ground, 99.2%,
GF15765127 Zirconium, sponge, 20g, max. size 25mm, 99.2%,
GF59501257 Zirconium, sponge, 500g, max. size 25mm, 99.2%,
GF38858057 Zirconium, foil, not light tested, 25x25mm, thickness 0.003mm, 99.2%,
GF35651639 Zirconium, foil, not light tested, 25x25mm, thickness 0.004mm, 99.2%,
GF00437161 Zirconium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.2%,
GF47948653 Zirconium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.2%,
GF10405123 Zirconium, foil, not light tested, 50x50mm, thickness 0.001mm, 99.2%,
GF41568543 Zirconium, foil, not light tested, 50x50mm, thickness 0.002mm, 99.2%,
GF19980502 Zirconium, foil, not light tested, 50x50mm, thickness 0.003mm, 99.2%,
GF46075765 Zirconium, foil, not light tested, 50x50mm, thickness 0.004mm, 99.2%,
GF59265580 Zirconium, foil, 8mm disks, thickness 0.1mm, annealed, 99.2%,
GF56400308 Zirconium, foil, 8mm disks, thickness 0.25mm, annealed, 99.2%,
GF07231092 Zirconium, foil, 8mm disks, thickness 0.5mm, annealed, 99.2%,
GF87583113 Zirconium, foil, light tested, 100x100mm, thickness 0.007mm, 99.2%,
GF56385546 Zirconium, foil, light tested, 100x100mm, thickness 0.008mm, 99.2%,
GF33698065 Zirconium, foil, light tested, 100x100mm, thickness 0.009mm, 99.2%,
GF71987226 Zirconium, foil, light tested, 100x100mm, thickness 0.0125mm, 99.2%,
GF98201426 Zirconium, foil, light tested, 100x100mm, thickness 0.015mm, 99.2%,
GF40690582 Zirconium, foil, light tested, 100x100mm, thickness 0.016mm, 99.2%,
GF40579184 Zirconium, foil, light tested, 100x100mm, thickness 0.01mm, 99.2%,
GF56972593 Zirconium, foil, light tested, 100x100mm, thickness 0.01mm, 99.2%,
GF89795132 Zirconium, foil, light tested, 100x100mm, thickness 0.02mm, 99.2%,
GF89984457 Zirconium, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.2%,
GF81275463 Zirconium, foil, light tested, 100x100mm, thickness 0.075mm, annealed, 99.2%,
GF91375960 Zirconium, foil, light tested, 100x300mm, thickness 0.016mm, 99.2%,
GF96239380 Zirconium, foil, light tested, 150x150mm, thickness 0.007mm, 99.2%,
GF08197836 Zirconium, foil, 10mm disks, thickness 0.1mm, annealed, 99.2%,
GF64127460 Zirconium, foil, 15mm disks, thickness 0.015mm, 99.2%,
GF10902923 Zirconium, foil, 15mm disks, thickness 0.02mm, 99.2%,
GF85806108 Zirconium, foil, 15mm disks, thickness 0.5mm, annealed, 99.2%,
GF43235120 Zirconium, foil, 1m coil, thickness 0.075mm, annealed, 99.2%,
GF85670646 Zirconium, foil, 25mm disks, thickness 0.01mm, 99.2%,
GF15330439 Zirconium, foil, light tested, 200x300mm, thickness 0.025mm, annealed, 99.2%,
GF04947172 Zirconium, foil, light tested, 25x25mm, thickness 0.008mm, 99.2%,
GF24807832 Zirconium, foil, light tested, 50x50mm, thickness 0.005mm, 99.2%,
GF67002599 Zirconium, foil, light tested, 50x50mm, thickness 0.015mm, 99.2%,
GF13450205 Zirconium, foil, light tested, 50x50mm, thickness 0.075mm, annealed, 99.2%,
GF57727705 Zirconium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.2%,
GF86222808 Zirconium, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.2%,
GF70641086 Zirconium, sponge, 50g, max. size 25mm, 99.2%,
GF19445519 Zirconium, tube, 100mm, outside diameter 1.6mm, inside diameter 1.1mm, wall thickness 0.25mm, hard, 99.2%,
GF37657424 Zirconium, tube, 100mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, annealed, 99.2%,
GF60025890 Zirconium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%,
GF30689040 Zirconium, tube, 200mm, outside diameter 1.6mm, inside diameter 1.1mm, wall thickness 0.25mm, hard, 99.2%,
GF06467418 Zirconium, tube, 200mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%,
GF07177912 Zirconium, tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, annealed, 99.2%,
GF03237421 Zirconium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%,
GF36130632 Zirconium, tube, 500mm, outside diameter 1.6mm, inside diameter 1.1mm, wall thickness 0.25mm, hard, 99.2%,
GF08426640 Zirconium, tube, 500mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%,
GF97239044 Zirconium, tube, 500mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, annealed, 99.2%,
GF04678230 Zirconium, tube, 500mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%,
GF69666610 Zirconium, wire reel, 0.5m, diameter 0.05mm, as drawn, 99.2%,
GF28665726 Zirconium, wire reel, 10m, diameter 0.125mm, stress relieved, 99.2%,
GF49275074 Zirconium, wire reel, 10m, diameter 0.25mm, as drawn, 99.2%,
GF00785752 Zirconium, wire reel, 10m, diameter 0.28mm, annealed, 99.2%,
GF46889128 Zirconium, wire reel, 10m, diameter 0.5mm, as drawn, 99.2%,
GF08326975 Zirconium, wire reel, 1m, diameter 0.05mm, as drawn, 99.2%,
GF48232515 Zirconium, wire reel, 1m, diameter 0.125mm, stress relieved, 99.2%,
GF53750358 Zirconium, wire reel, 1m, diameter 0.25mm, as drawn, 99.2%,
GF29269749 Zirconium, wire reel, 1m, diameter 0.28mm, annealed, 99.2%,
GF83216246 Zirconium, wire reel, 20m, diameter 0.125mm, stress relieved, 99.2%,
GF59383817 Zirconium, wire reel, 20m, diameter 0.25mm, as drawn, 99.2%,
GF82511782 Zirconium, wire reel, 20m, diameter 0.28mm, annealed, 99.2%,
GF61140080 Zirconium, wire reel, 20m, diameter 0.5mm, as drawn, 99.2%,
GF40871314 Zirconium, wire reel, 2m, diameter 0.05mm, as drawn, 99.2%,
GF08914664 Zirconium, wire reel, 5m, diameter 0.05mm, as drawn, 99.2%,
GF33029322 Zirconium, wire reel, 5m, diameter 0.125mm, stress relieved, 99.2%,
GF18794201 Zirconium, wire reel, 5m, diameter 0.25mm, as drawn, 99.2%,
GF58945731 Zirconium, wire reel, 5m, diameter 0.28mm, annealed, 99.2%,
GF52371941 Zirconium, wire reel, 5m, diameter 0.5mm, as drawn, 99.2%,
GF56264597 Zirconium, foil, not light tested, 50x50mm, thickness 0.006mm, 99.2%,
GF11722249 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 47mm,
GF12151853 Zirconium, lump, 8 mm max. lump size, weight 100 g, purity 99.2%,
GF12674213 Zirconium, lump, 4 mm max. lump size, weight 50 g, purity 99.2%,
GF13882226 Zirconium, foil, thickness 1.0 mm, size 25 × 25 mm, purity 99.2%,
GF15396914 Zirconium, foil, thickness 0.088 mm, length 1 m, purity 99.2%,
GF16109052 Zirconium, foil, thickness 1.0 mm, 50 mm diameter, purity 99.2%,
GF18867932 Zirconium, foil, thickness 1.0 mm, size 50 × 50 mm, purity 99.2%,
GF20037350 Zirconium, foil, thickness 0.3 mm, length 20 m, coil width 45 mm,
GF21534836 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 47mm,
GF24160294 Zirconium, foil, thickness 0.3 mm, length 10 m, coil width 45 mm,
GF24614377 Zirconium, foil, thickness 0.088 mm, length 2 m, purity 99.2%,
GF24800576 Zirconium, rod, 16 mm diameter, length 500 mm, purity 99.2%,
GF26000299 Zirconium, foil, thickness 0.3 mm, length 1 m, coil width 45 mm,
GF29209967 Zirconium, lump, 8 mm max. lump size, weight 50 g, purity 99.2%,
GF33231091 Zirconium, foil, thickness 0.088 mm, 10 mm diameter, purity 99.2%,
GF34346044 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 33mm,
GF34667613 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 104mm,
GF35646802 Zirconium, rod, 10.0 mm diameter, length 1000 mm, purity 99.2%,
GF35929042 Zirconium, foil, thickness 0.088 mm, 25 mm diameter, purity 99.2%,
GF37464150 Zirconium, rod, 10.0 mm diameter, length 200 mm, purity 99.2%,
GF37865841 Zirconium, foil, thickness 0.088 mm, 8 mm diameter, purity 99.2%,
GF39888286 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 83mm,
GF46625418 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 51mm,
GF49034022 Zirconium, wire, 1.0 mm diameter, length 10 m, purity 99.2%,
GF50036745 Zirconium, foil, thickness 0.3 mm, length 2 m, coil width 45 mm,
GF52295564 Zirconium, rod, 16 mm diameter, length 1000 mm, purity 99.2%,
GF53887438 Zirconium, foil, thickness 0.088 mm, 15 mm diameter, purity 99.2%,
GF54593286 Zirconium, foil, thickness 0.3 mm, length 5 m, coil width 45 mm,
GF58702923 Zirconium, foil, thickness 0.088 mm, 6 mm diameter, purity 99.2%,
GF60716000 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 47mm,
GF60874106 Zirconium, rod, length 100 mm, 10.0 mm diameter, purity 99.2%,
GF62285788 Zirconium, foil, thickness 0.088 mm, size 50 × 50 mm, purity 99.2%,
GF63566433 Zirconium, foil, thickness 1.0 mm, size 100 × 100 mm, purity 99.2%,
GF69436390 Zirconium, lump, 4 mm max. lump size, weight 100 g, purity 99.2%,
GF70366057 Zirconium, rod, 16 mm diameter, length 100 mm, purity 99.2%,
GF72246719 Zirconium, lump, 8 mm max. lump size, weight 200 g, purity 99.2%,
GF75287425 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 47mm,
GF76223729 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 127mm,
GF76797354 Zirconium, foil, thickness 0.088 mm, length 5 m, purity 99.2%,
GF77298283 Zirconium, foil, thickness 0.088 mm, size 25 × 25 mm, purity 99.2%,
GF79786276 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 27mm,
GF85723173 Zirconium, foil, thickness 0.088 mm, size 80 × 100 mm, purity 99.2%,
GF85797400 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 21mm,
GF85844740 Zirconium, powder, 45 max. part. size (micron), weight 200 g, purity 99.2%,
GF88868615 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 152mm,
GF91130475 Zirconium, foil, diameter 50 mm, thickness 0.088 mm, purity 99.2%,
GF94778106 Zirconium, rod, 10.0 mm diameter, length 500 mm, purity 99.2%,
GF97117218 Zirconium, lump, 4 mm max. lump size, weight 20 g, purity 99.2%,
GF98211720 Zirconium, foil, thickness 1.0 mm, size 300 × 300 mm, purity 99.2%,
GF98568950 Zirconium, crucible, cylindrical, length 1 pcs, outside diameter 33mm,
GF99669627 Zirconium, foil, thickness 0.088 mm, diameter 4 mm, purity 99.2%,
GF99723684 Zirconium, rod, 16 mm diameter, length 200 mm, purity 99.2%,