• USA Home
  • Search Results

  • zirconiumivchloride233041002611611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zirconium(IV) chloride

4 Products
Synonym:   Zirconium tetrachloride
Image of Zirconium(IV) chloride
CAS Number:
Empirical Formula (Hill Notation):
 Cl4Zr
Molecular Weight:
 233.04
 EC Index Number
 233-058-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More