• USA Home
  • Search Results

  • zirconiumivoxide12322131423411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zirconium(IV) oxide

10 Products
Synonym:   Zirconia
Image of Zirconium(IV) oxide
CAS Number:
Linear Formula:
 ZrO2
Molecular Weight:
 123.22
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  pH Value
  Concentration
  Assay
  Particle Dimension
  Composite Dimension
Show More

Product #
Description
230693 Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis,
544760 Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM),
204994 Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2),
643122 Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O,
643025 Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O,
774030 Zirconium(IV) oxide, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis (excludes 2% HfO2),
769274 Zirconium(IV) oxide, pellets, diam. × thickness 10 mm × 3.5 mm,
769487 Zirconium(IV) oxide, pellets, diam. × thickness 13.5 mm × 9.5 mm,
769282 Zirconium(IV) oxide, pellets, diam. × thickness 18.5 mm × 7.3 mm,
769266 Zirconium(IV) oxide, pellets, diam. × thickness 17.5 mm × 5 mm,