• USA Home
  • Search Results

  • zleuoh26530201866811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Z-Leu-OH

1 Products
Synonym:   N-(Benzyloxycarbonyl)-L-leucine, N-(Benzyloxycarbonyl)leucine, N-(Carbobenzoxy)leucine, N-(Phenylmethoxy)carbonyl-L-leucine, N-Carbobenzoxy-L-leucine, N-Carbobenzyloxy-L-leucine, N-Carboxy-L-leucine N-benzyl ester, N-Cbz-L-leucine, Benzyloxycarbonyl-L-leucine, Benzyloxycarbonylleucine, Carbobenzoxyleucine, L-N-(Benzyloxycarbonyl)leucine, L-(Carbobenzyloxy)leucine, NSC 60039, Z-L-Leucine
Image of Z-Leu-OH
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH(NHCOOCH2C6H5)COOH
Molecular Weight:
 265.30
 Beilstein Registry Number:
 1253861
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
521221 Z-Leu-OH, 97% (HPLC),