• USA Home
  • Search Results

  • zoxamide3366415605268511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zoxamide

1 Products
Synonym:   3,5-Dichloro-N-(3-chloro-1-ethyl-1-methyl-2-oxopropyl)-4-methylbenzamide
Image of Zoxamide
CAS Number:
Empirical Formula (Hill Notation):
 C14H16Cl3NO2
Molecular Weight:
 336.64
 Beilstein/REAXYS Number:
 11413911
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More