• USA Home
  • Search Results

  • ztyroh31532116416511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Z-Tyr-OH

1 Products
Synonym:   N-Carbobenzyloxy-L-tyrosine, Z-L-Tyrosine
Image of Z-Tyr-OH
CAS Number:
Linear Formula:
 HOC6H4CH2CH(NHCOOCH2C6H5)COOH
Molecular Weight:
 315.32
 Beilstein Registry Number:
 2169918
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
470597 Z-Tyr-OH, 97%,