• USA Home
  • Search Results

  • zymosanafromsaccharomycescerevisiae123455885693211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zymosan A from Saccharomyces cerevisiae

1 Products
CAS Number:
 MDL number:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Physical Form
  Application(s)
Show More