Polyamide Resins
Technical Bulletin banner
Number AL-142
Ion-Exchange Resins and
Related Polymeric Adsorbents
Home | Amberlite® and Amberlyst® Resins | Dowex® Resins | Sephadex® Products
  Merrifield Peptide Resins | Polyethylenimine-Based Products | Polyamide Resins
VI. POLYAMIDE RESINS
Polyamide resins are high-molecular-weight polymers which feature amide linkages along the molecular chain. Natural polyamides include a wide range of proteins from which adhesives and plastics may be derived. Synthetic polyamides, such as nylon and aramid resins, are used extensively in commercial applications.
Table XV
Cat. No.
Form
Approx. density (g/cc)
Melt temp.Tm, 0C)
191043
Pellet
0.97
95
191051
Pellet
0.98
130
191043
Pellet
0.98
140
191051
Pellet
1.01
160
161078
Pellet
0.99
200
Home | Amberlite® and Amberlyst® Resins | Dowex® Resins | Sephadex® Products
  Merrifield Peptide Resins | Polyethylenimine-Based Products | Polyamide Resins