Technology Spotlights
EZ-CrotylMix Reagents
EZ-CrotylMix: A Comprehensive Solution to the Enantioselective Crotylation of Aldehydes
Prof. James Leighton has developed the EZ-CrotylMix reagents for the enantioselective crotylation of aldehydes. These reagents show unprecedented selectivity and versatility, and represent one of the only self-contained crotylation reagents to date.
Benefits of EZ-CrotylMix Reagents:
  • Bench stable reagents that allow for easy reaction set-up and manipulation
  • Provide the desired crotylation product in excellent yield and enantioselectivity
  • Self-contained reagents that do not require any additional synthetic steps
  • Represent a broadly reaching solution to the enantioselective crotylation of aldehydes
EZ-CrotylMix Reactivity
Leighton’s EZ-CrotylMix Reagents
  1.  Kim, H.; Ho, S.; Leighton, J. L. J. Am. Chem. Soc. 2011, 133, 6517.