Gulf Coast Conference, Petrochem
Join us at Gulf Coast Conference | Houston, TX | January 17–18, 2018
George R. Brown Convention Center | 1001 Avenida de las Americas
Explore the complete Petrochemical workflow portfolio, now from MilliporeSigma.
EMD Millipore + Sigma-Aldrich have come together as MilliporeSigma, the North American business of Merck KGaA, Darmstadt, Germany. With our combined portfolio, we strive to bring you the most innovative solutions for the petrochemical industry.  Learn more at www.sigmaaldrich.com/petrochem.
Visit Us at Booths 716 and 717
Wednesday, January 17: 9:00 am – 5:00 pm CST
Thursday, January 18: 9:00 am – 5:00 pm CST
Attend Our Training Session  
Determination of Water Content in Petroleum using Solubilized Solvents with Karl Fischer Titration – Abstract 134
Wednesday January 17: 10:00 am - 12:00 pm, Room 382B
Presenters: Bruce Herzig, Application Specialist for MilliporeSigma
George McLean, Application Specialist for Mettler Toledo
Water content in petroleum products can be difficult. Let us show you how saturated hydrocarbons in most cases can be titrated according to standard methods. When overcoming solubility problems of non-polar or weakly polar substances, the addition of a solubilizer to the solvent is necessary. Learn more about why long-chain and cyclic hydrocarbons, long-chain alcohols and chloroform are recommended. In addition, petroleum products are mixtures of long-chain or aromatic hydrocarbons and are hardly soluble in methanol. Learn more about which solubilizer to use for water determination by Karl Fischer and when a KF oven technique should be utilized.  
Learn about Unique Selectivity at our Oral Presentation
More on the Unique Selectivity of Ionic Liquid GC Stationary Phases
Wednesday January 17: 1:30 pm – 3:00 pm, Room 372B
Presenter: Len Sidisky
See What’s New with H2O at the New Product Showcase
NEW Milli-Q® IQ7000 Lab Water System
An ultrapure water solution designed with you in mind
Introducing the Milli-Q® IQ 7000 water purification system. It's intelligent, intuitive and designed to make your work as pleasant and comfortable as possible, maximizing lab productivity.
  • Breakthrough ergonomic design
  • Precision drop by drop dispensing
  • Mercury free UV technology
  • Fully adjustable dispense flow, up to 2 liters
Explore Our Portfolio