Gulf Coast Conference 2018
Gulf Coast Conference Essentials | Houston, TX | October 16–17, 2018
We are very excited to be back at the Gulf Coast Conference ( GCC) in Galveston this October 2018. Stop by booth #704 and let our petrochem experts know about the current challenges you are facing in your petrochemical workflow. Learn more about the latest sample preparation technology, water purification systems to optimize lab productivity, titration solutions to aid troubleshooting, and GC consumables to provide accurate results every time. See below for the list of posters, presentations and product highlights at the GCC Oct 2018. And be sure to test your petrochem knowledge with our interactive quiz at booth #704 to win a prize!
Want to see what we presented at the GCC in January? Click here.
   Request a quote or consultation
Presentations
Title: Method Development and Validation Strategy for the Karl Fischer Titration Process
Abstract: Karl Fischer titrations are used throughout the chemical industry for evaluating moisture content of products and raw materials. This presentation will demonstrate techniques to evaluate the solubility of a sample in the Karl Fischer reagent solution, and make proper choices in co-solvent to ensure an accurate and reliable titration. After developing a suitable solvent system, the titration must be validated and techniques for method validation will also be discussed.
Speaker:  Bruce Herzig
Date & Time: Tuesday 16th October, 2:35 PM -  3:35 PM
Location: Iris Room
Title: Latest Developments in Ionic Liquid Stationary Phase Technology
Abstract: Over the years, extensive evaluations of columns manufactured with ionic liquid stationary phases have occurred. Their main strength was discovered to be unique selectivity. This selectivity is made possible due to the various combinations of cations and anions that are available along with spacer groups used to prepare these geminal dicationic phases. Columns prepared with di- or tricationic phases have the ability to perform many of the same applications as columns made with polysiloxane polymer or polyethylene glycol stationary phases of similar polarity, but with slight elution order changes. This often results in increased resolution and/or shorter run times. This presentation will compare and contrast the selectivity of the ionic liquid stationary phases with traditional phases of similar or like selectivity for applications with a variety of different sample types from petrochemical, pharmaceutical, environmental, food, beverage, flavor or fragrance industries.
Speaker:
Len Sidisky
Date & Time:
Wednesday 17th October, 9:00 am – 9:30am
Location: Exhibit Hall A
New Product Introduction
HPLC-certified hydrophilic PTFE Millex® filters
Our new HPLC-certified Millex® hydrophilic PTFE syringe filters will be featured in the Best Time Saving category.
HPLC-certified hydrophilic PTFE Millex® filters are now available in our 13mm and 33mm overmolded design. These filters are certified for low levels of UV-absorbing extractables. The broad chemical compatibility of hydrophilic PTFE enables filtration of a wide range of aqueous and organic solvents. For hard to filter samples, the 33mm hydrophilic PTFE Millex® filters are also available for our Samplicity® G2 filtration system.
Product Website: More on the HPLC-certified Millex® hydrophilic PTFE syringe filters
Product Highlights
NEW Milli-Q® IQ7000 Lab Water System displayed at the New Product Showcase at the Gulf Coast Conference
An ultrapure water solution designed with you in mind
Introducing the Milli-Q® IQ 7000 water purification system. It is intelligent, intuitive, and designed to make your work as pleasant and comfortable as possible, maximizing lab productivity.
  • Breakthrough ergonomic design
  • Precision drop by drop dispensing
  • Mercury free UV technology
  • Fully adjustable dispense flow, up to 2 liters
Explore Our Portfolio