Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Coupler Configuration
Pure-Pac II® Assembly and Components
Pure-Pac II Assembly and Components
  1. 20L-200 L Stainless steel container (MAWP < 1 bar) with micromatic MAV valve and dip tube
  2. Z560723-1EA – Micromatic coupler with 12 PSI pressure relief
    (1 in. MNPT discharge, 1/4 in. FNPT nitrogen inlet)*
  3. a. Z565547-1EA – Stainless hose connector
       (1/4 in. MNPT – 1/4 in. hose I.D.)*
    b. 22700-U – 1/4 in. MNPT – 1/4 in. tube O.D.
  4. a. Z685658-1PAK – 1/4 in. I.D. Tygon* tubimg to attach nitrogen
    b. Z280240-1PAK – 1/4 in. PTFE tubing
    c. Z562408-1EA – 1/4 in. SS braided line
  5. Z562300-1EA – Stainless hex reducing coupling
    (1 in. FNPT – 1/2 in. FNPT)*
  6. Z562319-1EA – Stainless street elbow
    (1/2 in. MNPT – 1/2 in. FNPT)*
  7. Z263532-1EA – Tube adapter
    (1/2 in. MNPT – 3/8 in. tube/ferrule O.D.)*
  8. Z263605-1EA – SS braided PTFE hose
    (3/8 in. tube adapt I.D., end fitting 3/8 in., 8 ft. length)
  9. Z741030-1EA – PurePac® dispense toggle valve assembly with 3/8 in. tube fitting and PFA tubing
Z104388-1PAK – Note: While connecting metal to
metal parts, be sure to use PTFE sealing tape.*
*Included in Z566020-1KT kit
Pure-Pac II Container Cutaway Micromatic MacroValve Coupler