Products for Positron Emission Tomography
Labeled isotopes used as target materials in imaging have provided insights into understanding various mechanisms of the human physiology and for treatment of diseases. In the PET industry, Water-18O is widely used for the production of 18F radionuclides such as 18FDG (Fluoro-deoxyglucose). 18FDG has been used for PET in conjunction with other imaging modalities (CT/SPECT) to diagnose and treat a wide range of disease from malignancies in the lungs, liver and brain tumors to other neurological disorders such as epilepsy and Alzheimer′s.1-3 Water-16O is a target material for producing 13N radionuclides such as 13N-Ammonia used for PET myocardial perfusion imaging.4,5
ISOTEC offers target materials for the production of radionuclides of 18F, 11C and 15O. In addition to the target materials, we also offer all reagents used in the production of 18FDG. PET research studies have various applications that include Cardiology, Oncology, Neurology, Pharmacology, Neuropsychology and Cognitive Neuroscience. Contact Us for any questions or to place an order.
The Essential Role of Enriched Stable Nuclides in Positron Emission Tomography (PET)
PET Applications
Products for MRI/MRS and PET Imaging - Brochure
Tool Box
18FDG Synthesis Schematic
Structure Search
Molecular Weight Calculator
1. Hoh, C.K. Nucl Med Biol. 2007, 34, 737-742.
2. Morooka, M.;Kubota, K.; Murata, Y.; Shibuya, H,; Ito, K.; Mochizuki, M.; Akashi, T.; Chiba, T.; Nomura, T.; Ito, H.; Morita, Ann, T. Nuc! Med. 2008, 22, 635-639.
3. Fischman, A.J. Cancer Treat Res. 2008, 143, 67-92.
4. Alexanderson, E.; Gomez-Leon, A.; Vargas, A.; Romero, J.L.; Sierra Fernandez, C.; Rodriguez, Valero M,; Garcia-Rojas, L.; Meave, A.; Amigo, M.C. Rheumatology (Oxford). 2008. 47, 894-896.
5. Mullani, N.A.; Herbs, T.S.; O′Neil, R.G.; Gould, K.L.; Barron, B.J.; Abbruzzese, J.L. J Nucl Med. 2008, 49, 517-523.
Stable Isotope Home Page
Product #
Add to Cart
C203 2-Chloro-2-deoxy-D-glucose analytical standard
709573 Desmethylraclopride 98% (CP)
D204 S-(+)-O-Desmethylraclopride hydrobromide solid, ≥98% (HPLC)
F5006 2-Fluoro-2-deoxy-D-glucose glycosylation inhibitor, glucose analog
600911 15N2/O2(RG) Gas Mixture ratio (39:1), 99 atom % 15N
291110 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane 98%
M1568 Mannose triflate For PET imaging, ≥98% (TLC)
602892 Oxygen-18O2 99 atom % 18O, 99% (CP)
329878 Water-18O 97 atom % 18O