Syringe Needle Gauge Chart
Converting Needle Gauges to Syringe Size Measurements
Use the chart below to cross-reference needle gauge sizes 6 through 34 with their nominal outer diameters (in inches and millimeters), outer diameter range (in inches), nominal inner diameter (in inches and millimeters), nominal diameter range (in inches), and wall diameter (in inches).
Our chart is based on the Birmingham wire gauge system, which corresponds to the Stubs iron wire gauges. Sizes and gauges identify hypodermic needles (injection) as well as needle accessories like tubes.
As the gauge number increases, the size of the needle decreases. The steps between needle gauge sizes are uneven.
See our full line of needles and syringes.
Click on the links in the needle gauge column (size range 22-26s) to search for syringe and needle products of that particular gauge size.
  Nominal   Nominal    
Needle
Gauge
Outer Diameter (in) Outer Diameter (mm) Outer Diameter Range (in) Inner Diameter (in) Inner Diameter (mm) Inner Diameter Range (in) Wall (in)
6 0.203 5.16 0.2020 - 0.2040 0.173 4.39 0.1700 - 0.1760 0.015
6.5 0.188 4.78 0.1870 - 0.1890 0.168 4.27 0.1650 - 0.1710 0.01
7 0.18 4.57 0.1790 - 0.1810 0.15 3.81 0.1470 - 0.1530 0.015
7.5 0.172 4.37 0.1710 - 0.1730 0.15 3.81 0.1470 - 0.1530 0.011
8 0.165 4.19 0.1640 - 0.1660 0.135 3.43 0.1320 - 0.1380 0.015
8.5 0.156 3.96 0.1550 - 0.1570 0.136 3.45 0.1330 - 0.1390 0.01
9 0.148 3.76 0.1470 - 0.1490 0.118 3 0.1150 - 0.1210 0.015
9.5 0.141 3.58 0.1400 - 0.1420 0.119 3.02 0.1170 - 0.1210 0.011
10 0.134 3.4 0.1330 - 0.1350 0.106 2.69 0.1040 - 0.1080 0.014
10.5 0.126 3.2 0.1250 - 0.1270 0.106 2.69 0.1040 - 0.1080 0.01
11 0.12 3.05 0.1190 - 0.1210 0.094 2.39 0.0920 - 0.0960 0.013
11.5 0.115 2.92 0.1140 - 0.1160 0.0945 2.4 0.0930 - 0.0960 0.01
12 0.109 2.77 0.1080 - 0.1100 0.085 2.16 0.0830 - 0.0870 0.012
12.5 0.1 2.54 0.0990 - 0.1010 0.082 2.08 0.0800 - 0.0840 0.009
13 0.095 2.41 0.0940 - 0.0960 0.071 1.8 0.0690 - 0.0730 0.012
13.5 0.089 2.26 0.0880 - 0.0900 0.071 1.8 0.0690 - 0.0730 0.009
14 0.083 2.11 0.0820 - 0.0840 0.063 1.6 0.0610 - 0.0650 0.01
14.5 0.078 1.98 0.0770 - 0.0790 0.063 1.6 0.0620 - 0.0640 0.007
15 0.072 1.83 0.0715 - 0.0725 0.054 1.37 0.0525 - 0.0555 0.009
15.5 0.068 1.73 0.0675 - 0.0685 0.054 1.37 0.0525 - 0.0555 0.007
16 0.065 1.65 0.0645 - 0.0655 0.047 1.19 0.0455 - 0.0485 0.009
16.5 0.062 1.57 0.0615 - 0.0625 0.052 1.32 0.0505 - 0.0535 0.005
17 0.058 1.47 0.0575 - 0.0585 0.042 1.07 0.0405 - 0.0435 0.008
17.5 0.056 1.42 0.0555 - 0.0565 0.046 1.17 0.0450 - 0.0470 0.004
18 0.05 1.27 0.0495 - 0.0505 0.033 0.84 0.0315 - 0.0345 0.0085
18.5 0.046 1.17 0.0455 - 0.0465 0.034 0.86 0.0325 - 0.0355 0.006
19 0.042 1.07 0.0415 - 0.0425 0.027 0.69 0.0255 - 0.0285 0.0075
19.5 0.039 0.99 0.0385 - 0.0395 0.027 0.69 0.0255 - 0.0285 0.006
20 0.036 0.91 0.0355 - 0.0360 0.024 0.6 0.0230 - 0.0245 0.006
20.5 0.0343 0.87 0.0340 - 0.0345 0.0265 0.67 0.0255 - 0.0275 0.004
21 0.032 0.82 0.0320 - 0.0325 0.02 0.51 0.0195 - 0.0210 0.006
21.5 0.0303 0.77 0.0300 - 0.0305 0.0238 0.6 0.0230 - 0.0245 0.0035
22 0.028 0.72 0.0280 - 0.0285 0.016 0.41 0.0155 - 0.0170 0.006
22s 0.028 0.718 0.0280 - 0.0285 0.007 0.168 0.0055 - 0.0077 0.011
22.5 0.0263 0.67 0.0260 - 0.0265 0.0203 0.52 0.0195 - 0.0210 0.003
23 0.025 0.64 0.0250 - 0.0255 0.013 0.34 0.0125 - 0.0140 0.006
23s 0.025 0.642 0.0250 - 0.0255 0.005 0.116 0.0040 - 0.0051 0.01
23.5 0.0233 0.59 0.0230 - 0.0235 0.0173 0.44 0.0165 - 0.0180 0.003
24 0.022 0.57 0.0220 - 0.0225 0.012 0.31 0.0115 - 0.0130 0.005
24.5 0.0213 0.54 0.0210 - 0.0215 0.0163 0.41 0.0155 - 0.0170 0.002
25 0.02 0.51 0.0200 - 0.0205 0.01 0.26 0.0095 - 0.0110 0.005
25s 0.02 0.515 0.0200 - 0.0205 0.006 0.153 0.0055 - 0.0065 0.007
25.5 0.0193 0.49 0.0190 - 0.0195 0.0143 0.36 0.0135 - 0.0150 0.002
26 0.018 0.46 0.0180 - 0.0185 0.01 0.26 0.0095 - 0.0110 0.004
26s 0.0187 0.474 0.0184 - 0.0189 0.005 0.127 0.0045 - 0.0055 0.007
27 0.016 0.41 0.0160 - 0.0165 0.008 0.21 0.0075 - 0.0090 0.004
28 0.014 0.36 0.0140 - 0.0145 0.007 0.18 0.0065 - 0.0080 0.0035
29 0.013 0.34 0.0130 - 0.0135 0.007 0.18 0.0065 - 0.0080 0.003
30 0.012 0.31 0.0120 - 0.0125 0.006 0.16 0.0055 - 0.0070 0.003
31 0.01 0.26 0.0100 - 0.0105 0.005 0.12 0.0040 - 0.0055 0.0025
32 0.009 0.23 0.0090 - 0.0095 0.004 0.11 0.0035 - 0.0050 0.0025
33 0.0083 0.21 0.0080 - 0.0085 0.0043 0.11 0.0035 - 0.0050 0.002
34 0.0073 0.19 0.0070 - 0.0075 0.0033 0.08 0.0025 - 0.0040 0.002