分析标准品
TraceCERT®定量NMR (qNMR)认证参考物质
我们提供一系列NIST SRM可溯源性认证参考物质,可作为定量NMR研究的内标。这些产品包括用于1H、31P和19F NMR实验的qNMR标准品。
  • 根据ISO/IEC 17025和ISO指南34生产
  • 含量经定量NMR(qNMR)认证
  • 出色的准确性、计算不确定性和批次特定值
  • 可溯源至NIST SRM
  • 产品附带全面的证明文件
  • 符合ISO指南31的认证证书
定量NMR – 技术详情和TraceCERT® 认证标准物质 (PDF, 1.5 MB)
查看所有可用qNMR CRM的产品表和光谱:
该表概述了16 1H-qNMR标准品、3 31P-qNMR标准品和3 19F-qNMR标准品在不同溶剂中的光谱转移、溶解度和弛豫时间。它们覆盖了全部光谱范围,可用于几乎任何有机分子的qNMR定量。使用市售NMR溶剂在室温下完成溶解度测试。测试从1 mg/ml开始,最高至250 mg/ml(mg CRM/ml溶剂)。该表仅记录CRM 的T1 弛豫时间(在25°C 下,c ≈ 5至20 mg/ml),但在混合物中可能发生变化。因此,建议在qNMR实验前检查T1延迟。(*使用DCl或NaOD)
1H 定量NMR标准品
产品编号 物质名称
D2O
CDCl3
DMSO-d6
CD3OD
CD3CN
δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml)
01380 Ethylene carbonate 4.5 5.5 >250 4.5 7.0 >250 4.5 2.7 >250 4.5 5.3 >250 4.5 2.0 >250
03826 Calcium formate 8.4 3.4 >250 - - - - - - - - - - - -
06185 Benzoic acid - - - 8.1
7.7
7.5
3.7
4.0
3.4
150 8.0
7.6
7.5
3.3
3.7
3.0
>250 8.0
7.6
7.5
4.3
4.4
3.9
>250 8.0
7.7
7.5
4.5
2.5
2.6
80
06856 Duroquinone - - - 2.0 3.3 >250 2.0 3.1 30 2.0 4.0 10 2.0 5.4 20
07038 Dimethyl terephthalate - - - 8.1
4.0
3.6
1.8
160 8.1
3.9
2.9
1.1
20 8.1
4.0
4.4
2.4
4.0 8.1
3.9
4.9
2.6
20
14659 Potassium hydrogen phthalate 7.5 2.5 >250* - - - - - - 7.9 2.5 5.0 - - -
40384 1,2,4,5-Tetrachloro- 3-nitro-benzene - - - 7.8 10.7 >250 8.5 12.6 >250 9.4 9.6 50 6.8 6.4 10
41867 Dimethyl sulfone 3.0 2.9 >250 3.0 2.7 80 3.0 2.4 >250 3.0 3.3 40 2.9 2.6 >250
42582 Ethyl 4 (dimethyl-amino) benzoate - - - 7.9
6.7
4.3
3.1
1.4
3.8
2.4
2.8
2.0
2.5
>250 7.8
6.7
4.2
3.0
1.3
2.5
1.4
1.9
1.5
2.1
>250 7.9
6.7
4.3
3.0
1.4
3.4
2.5
3.3
2.2
2.7
>250 7.9
6.7
4.3
3.0
1.3
5.6
3.7
4.1
3.6
3.6
50
50409 Thymol - - - 7.1
6.8
6.6
3.2
2.3
1.3
3.8
4.5
4.8
4.3
3.1
1.9
>250 7.0
6.7
6.5
3.1
2.2
1.1
2.0
2.2
2.6
2.3
2.0
0.9
>250 7.0
6.6
6.5
3.2
2.2
1.2
3.7
4.4
5.8
4.0
2.8
1.8
>250 7.1
6.7
6.6
3.2
2.2
1.2
4.9
5.7
5.7
5.2
3.5
2.4
>250
55177 Benzyl benzoate - - - 5.4
8.1
4.3
13.6
>250 5.4
8.0
2.2
8.8
>250 5.4
8.1
2.0
3.9
>250 5.4
8.1
1.0
2.4
>250
74599 1,3,5-Trimethoxybenzene - - - 6.1
3.8
4.7
2.2
>250 6.1
3.7
3.2
1.4
>250 6.1
3.8
4.8
2.7
>250 6.7
4.3
3.4
2.6
>250
74658 1,2,4,5- Tetramethyl-benzene - - - 7.0
2.2
6.1
4.0
>250 6.9
2.1
4.7
2.6
10 6.9
2.2
5.9
4.3
10 6.9
2.2
7.7
5.0
50
89151 Dimethylmalonic acid 1.3 1.0 100 - - - 1.3 0.7 250 1.4 1.0 250 1.4 2.0 30
92816 Maleic acid 6.3 6.1 >250 - - - 6.3 3.0 >250 6.3 3.9 10 6.4 2.3 20
94681 Methyl 3,5-dinitrobenzoate - - - 9.3
9.2
4.1
8.0
6.1
2.6
>250 9.1
8.9
4.0
9.4
7.6
1.5
100 - - - 9.1
9.0
4.0
9.0
8.2
3.5
>250
31P 定量NMR标准品
产品编号 物质名称
D2O
CDCl3
DMSO-d6
CD3OD
CD3CN
δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml)
05498 Triphenyl phosphate - - - -17.7 2.7 >250 -17.3 1.2 >100 -17.5 3.1 >10 -17 4.3 >250
92214 Potassium phosphate monobasic 0.08 8.0 >250 - - - - - - - - - - - -
96708 Phosphonoacetic acid 15.7 4.6 >250 - - - 14.9 1.5 >250 17.7 2.9 >250 - - -
19F 定量NMR标准品
产品编号 物质名称
D2O
CDCl3
DMSO-d6
CD3OD
CD3CN
δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml)
07563 4,4’-Difluorobenzophenone
- - - -105.8 2.4 >250 -106.5 1.4 150 -108.1 2.8 30 -108.3 2.3 140
53396 2,4-Dichlorobenzotrifluoride
- - - -62.5 2.3 >250 -61.2 1.2 >250 -65.4 3.3 >250 -63 2.9 >250
80730 2-Chloro-4-fluorotoluene
- - - -115.8 4.4 >250 -115.3 3.3 >250 -117.7 4.8 >250 -117.3 4.7 >250