分散SPE净化管
Supel™ QuE Z-Sep QuEChERS净化管
 去除复杂基质中的脂肪
主要特点和优势
  • 适用于农药残留分析
  • 采用专利技术,大大减少了脂肪基质和色素的干扰
  • 利用“快速,容易,经济,有效,稳定,安全”的QuEChERS方法
索取评估样品
技术概述
Supel™QuE
Brochure
Supel™QuE 手册
—利用QuEChERS方法对食品和农产品进行多残留农药分析
脂肪基质中的化合物
分析
View QuEChERS video
Supel QuE Z-Sep 简介
Supel QuE Z-Sep吸附剂相比传统的QuEChERS固定相,能够更有效地去除样品萃取物中的脂肪和色素,改善了复杂基质样品的净化效果。Z-Sep产品通过消除令人困扰的基质干扰,提供了更强大的LC-MS和GC-MS方法。这种专利技术能够取代现有方法中的C18和PSA吸附剂,无需额外的方法开发。
Supel QuE Z-Sep/C18是Discovery® DSC-18和Z-Sep颗粒的组合,推荐用于脂肪含量低于15%的样品。Supel QuE Z-Sep+是C18和Z-Sep与硅胶双重结合的产物,推荐用于脂肪含量高于15%的样品净化。Supel QuE Z-Sep推荐用于脂肪基质中的疏水性化合物的分析。
支持数据
各种吸附剂去除油脂的测试。该混合物包含溶于90%乙腈:10%水中的1 mL单、二和三油脂溶液,以及25 mg吸附剂。利用HPLC-ELSD测量溶液中的残留油脂。下图显示了不同吸附剂上的油脂保留量。
90%乙腈:10%水混合液中的单、二和三油脂的保留量。
订购信息
查看产品目录
 技术文章及参考文献
在线研讨会
在线研讨会: 固相微萃取(SPME)方法开发的基本原则
文章
Fast Extraction and Low Level Detection of Chloramphenicol in Shrimp
Extraction and Analysis of Neonicotinoid Pesticides from Flower Blossoms Using QuEChERS and LC-MS
Increase Removal of Fat and Pigment from Avocado Extracts Prior to GC-MS Analysis of Pesticide and Metabolite Residue (Reporter US Volume 31.2)
Improve Pigment Removal and Decrease Matrix Interferences for Pesticide Determination in Oranges Using Supel QuE Z-Sep/C18 QuEChERS Sorbent
Reduce Matrix Background and Improve Overall Analyte Recovery for the Analysis of Pesticides in Beef Kidney
参考文献
Łukasz Rajski, Ana Lozano, Ana Uclés, Carmen Ferrer, and Amadeo R Fernández-Alba; Determination of pesticide residues in high oil vegetal commodities by using various multi-residue methods and clean-ups followed by liquid chromatography tandem mass spectrometry. Journal of Chromatography. A, 2013, 1304:109. (Abstract)
Sapozhnikova, Y.; Lehotay, S.J. Multi-class, multi-residue analysis of pesticides, polychlorinated biphenyls, polycyclic aromatic hydrocarbons, polybrominated diphenyl ethers and novel flame retardants in fish using fast, low-pressure gas chromatography–tandem mass spectrometry. Analytica Chimica Acta, 2013, 758, 80-92.
Geis-Asteggiante, L.; Lehotay, S.J.; Lightfield, A. R.; Dutko, T.; Ng, C.; Bluhm, L.; Ruggedness testing and validation of a practical analytical method for >100 veterinary drug residues in bovine muscle by ultrahigh performance liquid chromatography–tandem mass spectrometry. J.Chromatogr A. 2012, 1258, 43-54.