bio-spme
Bio-SPME 视频
Bio-SPME为生物微量采样和样品制备提供了新方法,并且可实现对游离分析物组分的直接检测。BioSPME平台可改善传统LC/MS应用并推动直接MS分析的未来发展。
该技术可从生物样品中选择性萃取各种分析物,同时排除不需要的大分子和基质。目前,我们可提供适用于各种应用的C18和PDMS/DVB固定相萃取头和。更多固定相即将上市!
查看订购信息
索取免费试用品
技术资源
特点 优势
方法简单 采样和样品制备步骤合为一步,节省成本和时间
可实现自动化 96孔形式高通量处理,减小人为误差
微量采样技术 少量和珍贵样品的完美选择
直接采样 无需样品预处理
萃取头选择性 可直接检测游离分析物组分
解吸附体积小 提高样品浓度,检测范围更广泛
萃取探针
96-tip 阵列
萃取头固定相 应用
C18 用于从水性样品中极性和非极性分析物的萃取
PDMS/DVB 用于水性样品或顶空进样时中等极性半挥发性或挥发性分析物的萃取
检测水平比干血斑(DBS)采样法高5-10倍
图1. 通过BioSPME和DBS方法萃取得到的分析物检测结果. 
BioSPME出色的分析结果主要在于萃取涂层的浓缩能力,以及从萃取头上置换分析物所需的解吸附体积较小。相比之下,DBS采样3 mm卡片上获得的样品相对较少而所需的解吸附体积较大,因此,没有浓缩样品的情况下,DBS方法能够检测到的样品量就很有限。所以,分析物的检测水平,SPME LC Tips 比 DBS 卡片高.
比Thermo RED(快速平衡透析)法的分析时间减少4倍
通过药物蛋白质结合研究对比BioSPME和Thermo RED技术。BioSPME实验方案需要约60分钟,而Thermo RED实验方案需要4个多小时。如图所示,在损失灵敏度的情况下,BioSPME更快速、更简单。想要了解更详细的实验步骤和信息,请查看海报“使用固相微萃取法快速确定药物蛋白质结合率 Rapid Determination of Drug Protein Binding Affinity using Solid Phase Microextraction.
技术资源
描述 类型 编号
产品参考文献
BioSPME Fibers and Probes 手册 T413047
SPME LC Tips 数据单页 T714005
SPME LC Fiber Probes 数据单页 T710118
网络研讨会
Biocompatible Solid Phase Microextraction (BioSPME): A New Tool for the Clinical Chemist
Solid Phase Microextraction: Sketching the Future of Microsampling in Bioanalysis; Sheelan Ahmad; Senior Scientist; GlaxoSmithKline R&D and the University of Hertfordshire
Basic Principles of Solid Phase Microextraction (SPME) Method Development
文章
Rapid Determination of Protein Binding Affinity Using Solid Phase Microextraction
Direct Analysis in Real Time (DART®)-MS Analysis of Fentanyl and Related Analogues from Saliva Using Biocompatible Solid Phase Microextraction (BioSPME)
(DART®)-MS Screening Method for Methamphetamine and Amphetamine Using Biocompatible Solid Phase Microextraction (BioSPME)
Bioanalytical Microsampling using Biocompatible SPME and Benefits over Dried Blood Spots
Utilizing Novel SPME LC Tips for Forensic Screening of Drugs of Abuse in Urine Samples
Biocompatible SPME Devices for Bioanalysis and in vivo Sampling
Investigation of Bio-SPME Technology for the Enrichment of Illicit “Bath Salts” Cathinone Compounds from Biological Samples T413032H
C18 SPME Fiber Probes for LC Applications
期刊参考文献
Biocompatible Solid-Phase Microextraction Nanoelectrospray Ionization: An Unexploited Tool in Bioanalysis. Anal Chem. 2016 Jan 19;88(2):1259-65. doi: 10.1021/acs.analchem.5b03668. Epub 2015 Dec 29. Gómez-Ríos GA, Reyes-Garcés N, Bojko B, Pawliszyn J.
Direct ionization of solid-phase microextraction fibers for quantitative drug bioanalysis: from peripheral circulation to mass spectrometry detection. Anal Chem. 2015 Jan 6;87(1):754-9. doi: 10.1021/ac503706n. Epub 2014 Dec 15. Ahmad S1, Tucker M, Spooner N, Murnane D, Gerhard U.
Coupling desorption electrospray ionization with solid-phase microextraction for screening and quantitative analysis of drugs in urine. Kennedy JH, Aurand C, Shirey R, Laughlin BC, Wiseman JM. Anal Chem. 2010 Sep 1;82(17):7502-8. doi: 10.1021/ac101295g.
Analytical methods used in conjunction with solid-phase microextraction: a review of recent bioanalytical applications. Bioanalysis. 2009 Sep;1(6):1081-102. doi: 10.4155/bio.09.88. Musteata ML, Musteata FM.
产品
描述 编号
SPME-LC萃取探针,官能团C18,5个探针/包 57281-U
SPME-LC Pipette Tips,官能团C18,96-tip阵列 57234-U
SPME-LC Pipette Tips,混合型PDMS/DVB,96-tip阵列 57248-U
IonSense® PDMS/DVB SPE-it Tips, 96-tip 阵列 57249-U
IonSense® C18 SPE-it Tips pkg of 96 ea 57264-U
产品商标
  • FTA和Whatman是GE Healthcare公司的注册商标。
  • IonSense是IonSense, Inc的注册商标