化学品 - 技术文库
颗粒尺寸换算表
筛子编号 标称筛孔尺寸
标准 目数 英寸 毫米 微米
25.4 mm 1 in. 1.00 25.4 25400
22.6 mm 7/8 in. 0.875 22.6 22600
19.0 mm 3/4 in. 0.750 19.0 19000
16.0 mm 5/8 in. 0.625 16.0 16000
13.5 mm 0.530 in. 0.530 13.5 13500
12.7 mm 1/2 in. 0.500 12.7 12700
11.2 mm 7/16 in. 0.438 11.2 11200
9.51 mm 3/8 in. 0.375 9.51 9510
8.00 mm 5/16 in. 0.312 8.00 8000
6.73 mm 0.265 in. 0.265 6.73 6730
6.35 mm 1/4 in. 0.250 6.35 6350
5.66 mm No.3 1/2 0.223 5.66 5660
4.76 mm No. 4 0.187 4.76 4760
4.00 mm No. 5 0.157 4.00 4000
3.36 mm No. 6 0.132 3.36 3360
2.83 mm No. 7 0.111 2.83 2830
2.38 mm No. 8 0.0937 2.38 2380
2.00 mm No. 10 0.0787 2.00 2000
1.68 mm No. 12 0.0661 1.68 1680
1.41 mm No. 14 0.0555 1.41 1410
1.19 mm No. 16 0.0469 1.19 1190
1.00 mm No. 18 0.0394 1.00 1000
0.841 mm No. 20 0.0331 0.841 841
0.707 mm No. 25 0.0278 0.707 707
0.595 mm No. 30 0.0234 0.595 595
0.500 mm No. 35 0.0197 0.500 500
0.420 mm No. 40 0.0165 0.420 420
0.354 mm No. 45 0.0139 0.354 354
0.297 mm No. 50 0.0117 0.297 297
0.250 mm No. 60 0.0098 0.250 250
0.210 mm No. 70 0.0083 0.210 210
0.177 mm No. 80 0.0070 0.177 177
0.149 mm No. 100 0.0059 0.149 149
0.125 mm No. 120 0.0049 0.125 125
0.105 mm No. 140 0.0041 0.105 105
0.088 mm No. 170 0.0035 0.088 88
0.074 mm No. 200 0.0029 0.074 74
0.063 mm No. 230 0.0025 0.063 63
0.053 mm No. 270 0.0021 0.053 53
0.044 mm No. 325 0.0017 0.044 44
0.037 mm No. 400 0.0015 0.037 37
尺寸较大的筛孔(1英寸到1/4英寸)在标识时采用筛网目数,并与筛孔尺寸相对应(单位为英寸)。在 3 1/2至400之间、尺寸较小的筛孔目数在标识时采用筛子上每英寸长度内的筛孔数目来表示。
在筛网目数标识中,鉴定颗粒尺寸的常规约定如下所示:
  • 筛网目数前的“+”表示颗粒留在筛子中;
  • 筛网目数前的“-”表示颗粒可以通过筛子;
  • 通常90%或更多的颗粒会留在指定的范围内。
例如,如果材料的颗粒尺寸为-4 + 40目,那么90%或更多的材料能通过4目筛(颗粒尺寸小于4.76mm),留在40目筛(颗粒尺寸大于0.420mm)中。如果材料是-40目,那么90%或更多的材料能通过40目筛(颗粒尺寸小于0.420mm)。
返回LabBasics技术文库主页