癌症研究
氧化偶氮甲烷
Sigma®高兴能够为癌症研究人员提供这一重要化合物
货号 A2853
氧化偶氮甲烷 (AOM) 是一种用于诱导大鼠和小鼠结肠癌的有效致癌物质。它已被用于评估预防性治疗对氧化偶氮甲烷致癌作用的疗效的研究。1,2,3 氧化偶氮甲烷也常用于在啮齿动物模型中确定特定食物的化学预防效果,如难消化的糖4,5、红肉6、以及绿茶7等。这些从啮齿动物模型中获取的结果有助于确定针对人类结肠癌的可能预防方法。8
在用AOM处理后的小鼠中所生长的肿瘤中检测到转化生长因子β(TGF-β)信号转导的变化或异常。9,10氧化偶氮甲烷处理能够激活EGF受体的内在酪氨酸激酶,同时刺激TGF-α的合成。11
环氧合酶2(COX-2)抑制剂NS-398(货号N194)能够降低氧化偶氮甲烷处理的大鼠中的肿瘤前细胞的发生率。12
参考文献
  1. Escribano, M., et al., Aspirin inhibits endothelial nitric oxide synthase (eNOS) and Flk-1 (vascular endothelial growth factor receptor-2) prior to rat colon tumour development. Clin. Sci. (Lond)., 106: 83-91 (2004).
  2. Marotta, F., et al., Chemopreventive effect of a probiotic preparation on the development of preneoplastic and neoplastic colonic lesions: an experimental study. Hepatogastroenterology, 50: 1914-8 (2003).
  3. Orii, S., et al., Chemoprevention for colorectal tumorigenesis associated with chronic colitis in mice via apoptosis. J. Exp. Clin. Cancer Res., 22: 41-6 (2003).
  4. Pool-Zobel, B., et al., Experimental evidences on the potential of prebiotic fructans to reduce the risk of colon cancer. Br. J. Nutr., Suppl 2: S273-81 (2002).
  5. Nakanishi, S., et al., Effects of high amylose maize starch and Clostridium butyricum on metabolism in colonic microbiota and formation of azoxymethane-induced aberrant crypt foci in the rat colon. Microbiol Immunol., 47: 951-8 (2003).
  6. Pierre, F., et al., Meat and cancer: haemoglobin and haemin in a low-calcium diet promote colorectal carcinogenesis at the aberrant crypt stage in rats. Carcinogenesis, 24: 1683-90 (2003).
  7. Metz, N., et al., Suppression of azoxymethane-induced preneoplastic lesions and inhibition of cyclooxygenase-2 activity in the colonic mucosa of rats drinking a crude green tea extract. Nutr Cancer., 38: 60-4 (2000).
  8. Corpet, D.E. and Pierre, F., Point: From animal models to prevention of colon cancer. Systematic review of chemoprevention in min mice and choice of the model system. Cancer Epidemiol. Biomarkers Prev., 12: 391-400 (2003).
  9. Guda, K., et al., Defective processing of the transforming growth factor-beta1 in azoxymethane-induced mouse colon tumors. Mol. Carcinog., 37: 51-9 (2003).
  10. Guda, K., et al., Aberrant transforming growth factor-beta signaling in azoxymethane-induced mouse colon tumors. Mol Carcinog., 31: 204-13 (2001).
  11. Relan, N.K., Identification and evaluation of the role of endogenous tyrosine kinases in azoxymethane induction of proliferative processes in the colonic mucosa of rats. Biochim Biophys Acta., 1244(2-3): 368-76 (1995
    Jun 9)
  12. Kishimoto, Y., et al., Effects of cyclooxygenase-2 inhibitor NS-398 on APC and c-myc expression in rat colon carcinogenesis induced by azoxymethane. J. Gastroenterol., 37: 186-93 (2002).
返回癌症研究页面