细胞培养
F-12 Ham 营养混合物配方
F12 Ham营养混合物
组分 N4888
[1x] g/L
N6658
[1x] g/L
N6760
g/L
无机盐
Calcium Chloride 0.0333 0.0333 0.0333
Cupric Sulfate • 5H2O 0.0000025 0.0000025 0.0000025
Ferrous Sulfate • 7H2O 0.000834 0.000834 0.000834
Magnesium Chloride 0.0576 0.0576 0.0576
Potassium Chloride 0.224 0.224 0.224
Sodium Bicarbonate 1.176 1.176
Sodium Chloride 7.599 7.599 7.599
Sodium Phosphate Dibasic (anhydrous) 0.14204 0.14204 0.14204
Zinc Sulfate • 7H2O 0.000863 0.000863 0.000863
氨基酸
L-Alanine 0.009 0.009 0.009
L-Arginine • HCl 0.211 0.211 0.211
L-Asparagine • H2O 0.01501 0.01501 0.01501
L-Aspartic Acid 0.0133 0.0133 0.0133
L-Cysteine • HCl • H2O 0.035 0.035 0.035
L-Glutamic Acid 0.0147 0.0147 0.0147
L-Glutamine 0.146 0.146
Glycine 0.00751 0.00751 0.00751
L-Histidine • 3HCl • H2O 0.02096 0.02096 0.02096
L-Isoleucine 0.00394 0.00394 0.00394
L-Leucine 0.0131 0.0131 0.0131
L-Lysine • HCl 0.0365 0.0365 0.0365
L-Methionine 0.00448 0.00448 0.00448
L-Phenylalanine 0.00496 0.00496 0.00496
L-Proline 0.0345 0.0345 0.345
L-Serine 0.0105 0.0105 0.0105
L-Threonine 0.0119 0.0119 0.0119
L-Tryptophan 0.00204 0.00204 0.00204
L-Tyrosine • 2Na • 2H2O 0.00778 0.00778 0.00778
L-Valine 0.0117 0.0117 0.0117
维生素
D-Biotin 0.0000073 0.0000073 0.000073
Choline Chloride 0.01396 0.01396 0.01396
Folic Acid 0.00132 0.00132 0.00132
myo-Inositol 0.018 0.018 0.018
Niacinamide 0.000037 0.000037 0.000037
D-Pantothenic Acid (hemicalcium) 0.00048 0.00048 0.00048
Pyridoxine • HCl 0.000062 0.000062 0.000062
Riboflavin 0.000038 0.000038 0.000038
Thiamine • HCl 0.00034 0.00034 0.00034
Vitamin B12 0.00136 0.00136 0.00136
其他
D-Glucose 1.802 1.802 1.802
Hypoxanthine 0.00408 0.00408 0.00408
Linoleic Acid 0.000084 0.000084 0.000084
Phenol Red • Na 0.0013 0.0013 0.0013
Putrescine • HCl 0.000161 0.000161 0.000161
Pyruvic Acid • Na 0.11 0.11 0.11
Thioctic Acid 0.00021 0.00021 0.00021
Thymidine 0.00073 0.00073 0.00073
添加成分
L-Glutamine 0.146
Sodium Bicarbonate 1.176
返回页首
HEPES改良版F-12 Ham营养混合物
组分 N8641
[1x] g/L
无机盐
Calcium Chloride 0.0333
Cupric Sulfate • 5H2O 0.0000025
Ferrous Sulfate • 7H2O 0.000834
Magnesium Chloride 0.0576
Potassium Chloride 0.224
Sodium Bicarbonate 1.176
Sodium Chloride 7.1
Sodium Phosphate Dibasic (anhydrous) 0.14204
Zinc Sulfate • 7H2O 0.000863
氨基酸
L-Alanine 0.009
L-Arginine • HCl 0.211
L-Asparagine • H2O 0.01501
L-Aspartic Acid 0.0133
L-Cysteine • HCl • H2O 0.035
L-Glutamic Acid 0.0147
Glycine 0.00751
L-Histidine • 3HCl • H2O 0.02096
L-Isoleucine 0.00394
L-Leucine 0.0131
L-Lysine • HCl 0.0365
L-Methionine 0.00448
L-Phenylalanine 0.00496
L-Proline 0.0345
L-Serine 0.0105
L-Threonine 0.0119
L-Tryptophan 0.00204
L-Tyrosine • 2Na • 2H2O 0.00778
L-Valine 0.0117
维生素
D-Biotin 0.0000073
Choline Chloride 0.01396
Folic Acid 0.00132
myo-Inositol 0.018
Niacinamide 0.000037
D-Pantothenic Acid (hemicalcium) 0.000238
Pyridoxine • HCl 0.000062
Riboflavin 0.000038
Thiamine • HCl 0.00034
Vitamin B12 0.00136
其他
D-Glucose 1.802
HEPES 5.958
Hypoxanthine 0.00408
Linoleic Acid 0.000084
Phenol Red • Na 0.0013
Putrescine • HCl 1.000161
Pyruvic Acid • Na 0.11
Thioctic Acid 0.00021
Thymidine 0.00073
添加成分
L-Glutamine 0.146
返回页首
Kaighn改良版F-12 Ham营养混合物
组分 N3520
[1x] g/L
无机盐
Calcium Chloride 0.10207
Cupric Sulfate • 5H2O 0.0000025
Ferrous Sulfate • 7H2O 0.000834
Magnesium Chloride 0.04961
Magnesium Sulfate 0.19264
Potassium Chloride 0.28329
Potassium Phosphate Monobasic 0.058523
Sodium Phosphate Dibasic 0.11502
Sodium Chloride 7.5972
Zinc Sulfate • 7H2O 0.00014375
氨基酸
L-Alanine 0.017818
L-Arginine 0.4214
L-Asparagine • H2O 0.03002
L-Aspartic Acid 0.02662
L-Cysteine • HCl • H2O 0.07024
L-Glutamic Acid 0.02942
L-Glutamine 0.2922
Glycine 0.015014
L-Histidine • HCl • H2O 0.04192
L-Isoleucine 0.007872
L-Leucine 0.02624
L-Lysine • HCl 0.07304
L-Methionine 0.008952
L-Phenylalanine 0.009912
L-Proline 0.06906
L-Serine 0.02102
L-Threonine 0.02382
L-Tryptophan 0.004084
L-Tyrosine 0.010872
L-Valine 0.02342
维生素
D-Biotin 0.00007329
Choline Chloride 0.01396
Folic Acid 0.0013242
Hypoxanthine 0.004083
myo-Inositol 0.01802
Niacinamide 0.00003663
D-Pantothenic Acid (hemicalcium) 0.000477
Putrescine • 2HCl 0.0003222
Pyridoxine • HCl 0.00006168
Riboflavin 0.00003764
Thiamine • HCl 0.0003373
Thymidine 0.0007266
Vitamin B12 0.0013554
其他
D+-Glucose 1.26
Phenol Red • Na 0.00331806
Pyruvic Acid • Na 0.22
DL-6,8-Thioctic Acid 0.0002063
添加成分
Sodium Bicarbonate 2.5
返回页首