生物缓冲液
BIS-TRIS丙烷
BIS-TRIS丙烷是一种具有特别宽泛缓冲范围的两性离子缓冲液,因其两个pKa值非常接近,适用范围约在pH 6至9.5之间。BIS-TRIS丙烷,相比于Tris缓冲液,已被用于增强限制性酶的稳定性和活性。1.
可用pH范围 6.3 - 9.5
工作浓度范围 10mM-50mM4,5
        根据等级或质量水平进行比较
名称 CAS 分子式 分子量 试剂等级 生命科学等级 药物等级   溶液
BIS-TRIS 丙烷 64431-96-5 C11H26N2O6 282.33 RDD014 B6755 B4910 B4679 PHG0005  
互作信息
与Ni、Co、Cu、Zn、Cd及Pb具有强相互作用3
参考文献
  1. Eckert, K. A., and Kunkel, T. A., DNA polymerase fidelity and the polymerase chain reaction. PCR Methods Appl., 1(1), 17-24 (1991).
  2. Dutta K, Natarajan A, Nair P, Shuman S, and Ghose R. “Sequence-specific 1H, 13C and 15N assignments of the phosphoesterase (PE) domain of Pseudomonas aeruginosa DNA ligase D” Biomol. 2011 October.
  3. Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003.
  4. Moldoveanu S, David V, Essentials in Modern HPLC separation, Elsevier 2013 pp. 413.
  5. Makarchikov AF, Brans A, Betterdorf FL, Thiamine diphosphate adenylyl transferase from E. coli: functional characterization of the enzyme synthesizing adenosine thiamine triphosphate, BMC Biochemistry 2007 V8 N17.