生物缓冲液
HEPES
HEPES是两性离子生物缓冲液,也是Good's缓冲液之一。HEPES相比碳酸氢盐缓冲液尽管二氧化碳浓度改变,但在维持细胞培养的生理pH方面效果更好1
pH值范围6.8-8.2的HEPES缓冲液可制备成多种比例。
有效PH范围 6.8 - 8.2
反应浓度范围 10mM-1M7-9
        按等级或质量水平的产品比较
名称 CAS号 分子式 摩尔质量 试剂级 生命科学级 制药级  溶液
HEPES(游离酸) 7365-45-9 C8H18N2O4S 238.30 RDD002
H3375
5310-OP
391338
54457
H7523
H4034
H6147
PHG0001 RES6003H-B7 83264
H0887
H3537
375368
HEPES纳盐 75277-39-3 C8H17N2NaO4S 260.29 H7006 H3784 5380-OP H8651
H3784
RES6007H-A7  H3662 
HEPES半纳盐 103404-87-1 C8H17.5N2O4S • 0.5Na 249.30 H7637 H9897   RES6008H-A7  
HEPES钾盐 82207-62-3 C8H17KN2O4S 276.39 H0527      
互动评论
HEPES不结合镁,钙或锰(II)离子2.。
HEPES不建议对特定蛋白质应用;它会干扰Folin-Ciocalteu蛋白质分析。双缩脲蛋白质测定不受影响3
HEPES在UV范围内显示低吸光度,但是干扰Lowry蛋白质测定产生假阳性结果。3
被铜(II)氧化5
可以形成自由基,不适合氧化还原研究。4
参考文献
1.      Baicu SC, Taylor MJ. Cryobiology 45 (1): 33–48 (2002)
2.      Good, N.E. and Izawa, S., Methods in Enzymology, 24B, 53 (1972).
3.      Himmel, H.M. & Heller, W.: J. Clin. Chem. Clin. Biochem. 25, 909-913 (1987)
4.      Grady, J. K.; Chasteen, N. D.; Harris, D. C. (1988). "Radicals from "Good's" buffers". Anal. Biochem. 173 (1): 111–115.
5.      Fengjiang W, Lawrence MS "Oxidation of Tertiary Amine Buffers by Copper". Inorganic Chemistry 1989 (28; 2): 168-169
6.      Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003
7.      Downs SM, Mastropolo AM, Culture Conditions Affect Meiotic Regulation in Cumulus Cell-Enclosed Mouse Oocytes, Molecular Reproduction and Development 1997(46) 551-566
8.      Gupta B, Taha M, Lee M-J, Buffers more than buffering agent: introducing a new class of stabilizers for the protein BSA, Phys.Chem.Chem.Phys 2015(17) pp 1114-1133
9.      Kebede N, Francis PS, Barbante GJ, Hogan CF, Electrogenerated chemiluminescence of tris (2,2’ biopyridine) ruthenium (II) using common biological buffers as co-reactant, pH buffer and supporting electrolyte, Royal Society of Chemistry 2015(140) pp7142-7145