生物缓冲液
MOPS
别名:3-吗啉丙烷-1-磺酸、3-(N-吗啉代)丙磺酸和3-吗啉丙磺酸
两性离子缓冲液MOPS是MES(生物化学通用的Good's缓冲液中的一种)的结构类似物,主要用作琼脂糖凝胶分离RNA时的缓冲液2
可用的pH范围 6.5 - 7.9
工作浓度范围 20mM-1M7, 8
        按等级或质量水平对产品进行比较
名称 CAS 分子式 分子量 试剂级 生命科学级
制药级
溶液
MOPS 1132-61-2 C7H15NO4S 209.26 6310-OP M1254 RDD003 M3183 M5162 69947 PHG0007 M1442
MOPS钠盐 71119-22-7 C7H14NNaO4S 231.25 M9381 RDD018 475889 M5789 M9024 RES0197M-A7  
MOPS半钠盐 117961-20-3 C7H14.5NO4S • 0.5Na 220.25 M0289 M9027      
相互作用方面的备注
MOPS对福林法测蛋白有干扰5
当在葡萄糖存在下高压灭菌时,MOPS溶液不完全稳定。1
不建议在哺乳动物细胞培养工作中使用20 mM以上的浓度4
MOPS使 BSA稳定3
与Fe强相互作用;与Mg、Mn、Co和Ni弱相互作用6
参考文献
1.      The Merck Index, 12th ed., Entry# 6346. 1996
2.      Molecular Cloning: A Laboratory Manual, 3rd ed., Sambrook, J. and Russell, D.W., CSHL Press (Cold Spring Harbor, NY: 2001), p. 7.32.
3.      Taha, Mohamed. et al. 2011. Macromolecules. 44.21: 8575-8589.
4.      Eagle, H., Science, 174, 500-503 (1971).
5.      Ferguson, W.J. et al. (1980) Anal. Biochem. 104, 300-310 Hydrogen ion buffers for biological research.
6.      Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003
7.      Downs SM, Mastropolo AM, Culture Conditions Affect Meiotic Regulation in Cumulus Cell-Enclosed Mouse Oocytes, Molecular Reproduction and Development 1997(46) 551-566
8.      Gupta B, Taha M, Lee M-J, Buffers more than buffering agent: introducing a new class of stabilizers for the protein BSA, Phys. Chem. Chem. Phys 2015(17) pp 1114-1133