生物学缓冲液
Trizma® 碱规格比较
同义词:2-氨基-2-(羟甲基)-1,3-丙二醇,THAM,Tris碱,三羟甲基氨基甲烷,Trometamol
TRIS是一种用于生物化学和分子生物学的成熟纯度标准品和缓冲液1。其可用于配制pH范围7.5-8.5的缓冲溶液。在细胞和分子生物学应用中,TRIS缓冲溶液广泛用于蛋白质和核酸提取和纯化等流程。
可用的pH范围 7 - 9
工作浓度范围 10-62.5mM9,10
        根据级别和质量级进行的产品比较结果
名称 CAS 分子式 摩尔重量 试剂级 生命科学级 制药级  溶液级
TRIZMA® 77-86-1 C4H11NO3 121.14 T4661 RDD008 93352 T1503 T6066 93362 T6791   T1669
氨丁三醇 77-86-1 C4H11NO3 121.14     RES3137T-A7 RES3193T-A7  
Sigma 7-9® 77-86-1 C4H11NO3 121.14 T1378      
Trizma® 盐酸盐/ TRIS 盐酸盐 1185-53-1 C4H12ClNO3 157.60 RDD009 T3253 9310-OP 648313 93363 T5941 T6666 648317 RES3098T-B7 PHG0002  
交互评论
TRIS 可和醛和酮反应产生Schiff 碱。5
TRIS经报道可干扰一系列酶的活性,因而在研究蛋白质时应谨慎使用2,3
TRIS在温度变化时出现了较大的解离迁移。6
TRIS 干扰Bradford 染料结合蛋白质定量检测实验4
与铬、铁、钴、镍、铜互作强;与镁、钙、锌、镉和铅互作弱8
文献参考资料
1.      Gomori, G., Preparation of Buffers for Use in Enzyme Studies. Methods Enzymol., 1, 138-146 (1955).
2.      Desmarais, W.T., et al. 2002. Structure. 10: 1063-1072. PMID: 12176384
3.      Ghalanbor, Z., et al. 2008. Protein Pept. Lett. 15: 212-214. PMID: 18289113
4.      Stoll, V.S. and Blanchard, J.S. (1990) Buffers: Principles and practice. Meth. Enzmol. 182, 24–38.
5.      Hermanson, G.T. (1996). Bioconjugate Techniques, Academic Press.
6.      El-Harakany, A.A.; Abdel Halima, F.M. and Barakat, A.O. (1984). "Dissociation constants and related thermodynamic quantities of the protonated acid form of tris-(hydroxymethyl)-aminomethane in mixtures of 2-methoxyethanol and water at different temperatures". J. Electroanal. Chem. 162 (1–2): 285–305.
7.      Mohamed Taha, Ming-Jer Lee. Biochemical Engineering Journal. Volume 46, Issue 3, 1 November 2009, Pages 334–344.
8.      Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003
9.      Western blot protocol- Duke University http://neuro.surgery.duke.edu/files/documents/Western_Blot.pdf
10.    Shepherd LD, McLay TGB, Two micro-scale protocols for the isolation of DNA from polysaccharide-rich plant tissue, J Plant Res 2011 Mar;124(2):311-4.