Enzyme Explorer
凝血酶因子IIa
Thrombin IIa header
本页内容
 特异性
凝血酶是一种内切丝氨酸蛋白酶,可选择性地裂解纤维蛋白原的Arg -Gly键,形成纤维蛋白并释放纤维蛋白肽AB1,2
凝血酶的最佳切割位点是:1)AB-Pro-Arg- || -XY,其中A和B是疏水性氨基酸,X和Y是非酸性氨基酸;2)Gly-Arg- || -Gly。2
任意哺乳动物的凝血酶可凝结其他哺乳动物的纤维蛋白原3
凝血酶有两种仅作用于精氨酸位点的纤维蛋白原裂解方式。初级裂解产物纤维蛋白肽A是从纤维蛋白原第16位氨基酸后裂解,有时从第19位氨基酸后裂解,而次级裂解产物纤维蛋白肽B是在第14位氨基酸处裂解产生。4
凝血酶在pH 5-10.5的范围内有活性。5
催化最适pH是8.3。5
凝血酶在pH 5或更低时沉淀。5
凝血酶活化不需要二价金属离子或辅因子。但凝血酶的Na +依赖性变构激活与凝血酶在精氨酸残基位点切割的特异性密切相关。血浆蛋白C活化时,血栓调节蛋白是凝血酶的辅因子。7
 体内活化过程和物理性质
凝血酶在体内的主要形式是它的酶原,凝血酶原(因子II),由肝脏产生。正常人血浆中凝血酶原的浓度约为5-10mg / dL。凝血酶原是一种糖蛋白,含糖量约为~12%。8
凝血酶原在体内被活化的因子X切割,释放活化肽并将凝血酶切割成轻链和重链,产生催化活性的α-凝血酶。 α-凝血酶由轻链(A链)(MW〜6,000)和重链(B链)(〜31,000)组成。这两条链通过一个二硫键连接。12人凝血酶的B链由肽部分(MW 29,485)和三个天冬酰胺位点上具有N连接糖基化的碳水化合物部分(2334)组成。9,10 牛凝血酶含有1.7 %葡糖胺,1.8%唾液酸,0.61%半乳糖和0.95%甘露糖。11凝血酶还含有γ-羧基谷氨酰胺残基。这些修饰的谷氨酰残基是微粒体酶,维生素K依赖性羧化酶羧化的结果。γ-羧基谷氨酰残基对于负电荷磷脂表面的钙依赖性相互作用是必需的,对于凝血酶原向凝血酶的转化也是必不可少的。12在体内,凝血酶原在磷脂膜的表面活化,所述磷脂膜结合氨基末端凝血酶原以及Va和Xa因子。活化起始缓慢,因为 因子V本身是由最初的少量凝血酶激活。
在特定的储存条件下,α-凝血酶的自溶消化导致形成缺乏纤维蛋白溶解活性的β-纤维蛋白和γ-凝血酶,但保留一些除纤维蛋白原以外的抗肽底物和蛋白底物合成活性。13Sigma凝血酶制剂主要是α凝血酶形式。
人凝血酶由几个同工酶组成,等电点范围6.35-7.6。
牛凝血酶等电点范围是7.05  -  7.114
E1%(280nm) = 18.3 (人)15
E1%(280nm) = 19.5 (牛)16
 凝血酶活性测定
Sigma的凝血酶实验方法是以与NIH凝血酶参考标准直接比较获得NIH单位来衡量。
根据改进的Biggs法,NIH测定方案使用0.2 mL稀释血浆(1:1添加生理盐水)和0.1 mL凝血酶样品(在pH 7.35的1%缓冲白蛋白溶液中固定)作为底物。17仅在15-25秒的凝血时间内测定凝血酶浓度。
文献中的凝血酶浓度通常有不同的活性单位。18
凝血酶文献中的几个常识:
1 IOWA unit= 0.83 NIH unit
1 WHO unit = 0.56 NIH unit
1 NIH unit = 0.324 +/- 0.073 µg
1 NIH unit = 1 USP unit
凝血酶(人和牛)将催化几种肽对硝基苯胺,甲苯磺酰基-硝基苄基酯和硫代苄基酯合成底物的水解。19
 应用
血浆中纤维蛋白凝块的形成:
一到两个单位的凝血酶可凝结1毫升的血浆。
融合蛋白的切割:
在20微升50 mM pH 8.0碳酸氢铵溶液中,0.5 NIH unit /nM的凝血酶可在凝血酶识别位点处裂解多种肽。20
凝血酶切割融合蛋白可以在1:500比率下进行。21
融合蛋白可以在由50 mM Tris,pH 8.0,150 mM NaC,2.5 mM CaCl2和0.1%2-巯基乙醇组成的凝血酶切割缓冲液中被切割。将2 mg融合蛋白与4 µg凝血酶在裂解缓冲液中室温孵育20分钟。22
 产品
人重组凝血酶
货号 产品名称 加入购物车
T9326 HEK293细胞中表达的人重组凝血酶,≥2,000 units/mg蛋白
人类凝血酶
货号 产品名称 加入购物车
T1063 人血浆凝血酶,冻干粉末,> 2800 NIH units/mg蛋白质(E1%/280, 18.3)
T6884 人血浆凝血酶,冻干粉末,≥2,000 NIH units/mg蛋白质(E1%/280, 18.3)
T9010 人血浆凝血酶,冻干粉末,适用于凝血酶时间的常规检测
T7009 人血浆凝血酶,冻干粉末,〜1,000 NIH units/mg蛋白质(E1%/ 280,18.3)
T9135 人血浆凝血酶,冻干粉末,适用于凝血酶时间的常规检测
T4393 人血浆凝血酶,冻干粉末,〜1,000 NIH units/mg蛋白质(E1%/ 280,18.3),经过细胞培养检测
T7572 人血浆凝血酶,50-300 NIH units/mg蛋白质
牛凝血酶
货号 产品名称 加入购物车
T7513 牛血浆凝血酶,冻干粉末,≥2,000 NIH units/mg蛋白质(E1%/280 = 19.5)
T9549 牛血浆来源凝血酶,粉末,经过细胞培养测试,1,500-2,500 NIH units/mg蛋白质(E1%/280 = 19.5)
T9681 牛血浆凝血酶,冻干粉末,≥700 NIH units/mg蛋白(考马斯亮蓝)
T6634 牛血浆凝血酶,冻干粉末,600-1,500 NIH units/mg蛋白质(双缩脲)
T4265 牛血浆凝血酶,冻干粉末,175-350个NIH units/mg蛋白质(双缩脲)
T9000 牛血浆缓冲水溶液来源凝血酶,≥125 NIH units/mg蛋白质(双缩脲)
T6200 牛血浆凝血酶,冻干粉末,40-300 NIH units/mg蛋白质(双缩脲)
T3399 牛血浆凝血酶,冻干粉末,50-150 NIH units/mg蛋白质(双缩脲)
鼠凝血酶
货号 产品名称 加入购物车
T8397 鼠血浆凝血酶,冻干粉末,〜1,000 NIH units/mg蛋白质(双缩脲)
兔凝血酶
货号 产品名称 加入购物车
T4897 兔血浆凝血酶
大鼠凝血酶
货号 产品名称 加入购物车
T5772 大鼠血浆来源凝血酶,冻干粉末,≥1,000 units/mg蛋白质(双缩脲)
参考文献
  1. Enzyme Nomenclature: EC 3.4.21.5
  2. Chang, J.Y., Eur. J. Biochem., 151, 217?224 (1985).
  3. The Plasma Proteins, 2nd ed., 2, Putnam, F. W., ed, p. 148.
  4. Machovich, R., The Thrombin, 1, 63-66 (1984)
  5. Machovich, R., The Thrombin, 1, 111 (1984)
  6. Prasad, S., J. Biol. Chem. 279, 10103-10108 (2004)
  7. Kisiel, W., Human plasma protein C: isolation, characterization, and mechanism of activation by alpha-thrombin. J. Clin. Invest. 64, 761-769, (1979)
  8. The Plasma Proteins, 2nd ed., 2, Putnam, F. W., ed: Table 2. See also: The Enzyme Explorer: Plasma and Blood Protein Resource
  9. Qian, W.J., et al., J. Proteome Res., 4, 2070-2080 (2005).
  10. Nilsson, B., et al., Arch. Biochem. Biophys., 224, 127-133 (1983)
  11. Boyer, P.D., The Enzymes, Academic Press (New York), 3rd ed., Vol. III, p. 277-321 (1971).
  12. Expasy/SwissProt: P00743
  13. Boissel, J.P., et al., J. Biol. Chem., 259, 5691-5697 1984).
  14. Righetti, P.G., and Tudor, G., Isoelectric points and molecular weights of proteins, a new table. Journal of Chromatography, 220, 115-194 (1981).
  15. Butkowski, R.J. et al., J. Biol. Chem., 252, 4942 (1977).
  16. Winzor, D. J. and Scheraga, H. A., Arch. Biochem. Biophys. 104, 202-207 (1964)
  17. Human Blood Coagulation, Haemostasis and Thrombosis, 2nd ed., R. Biggs, ed., p. 722 (1976).
  18. The Handbook of Synthetic Substrates, Hemker, H. C., Martinus Nijhoff publisher (1983).
  19. Lottenberg, R., et al., Assay of Coagulation Proteases Using Peptide Chromogenic and Fluorogenic Substrates. Meth. Enzymol., 80-C, 341-361 (1981).
  20. Chang, Y., Thrombin specificity. Requirement for apolar amino acids adjacent to the thrombin cleavage site of polypeptide substrate. Eur. J. Biochem., 151(2), 217-224 (1985).
  21. Hakes, D.J. and Dixon, J.E., Anal. Biochem., 202, 293 (1992).
  22. Gaun, KL and Dixon, JE,, Anal. Biochem., 192, 262, 1991
  23. De Cristofaro, R. and De Candia, E., J. Thromb. Thrombolysis, 15, 151-163 (2003)
  24. Sherwood, J.A., Mol. Biochem. Parisitol., 40, 173-181 (1990)
  25. Berg, D.T., et al., Science, 273, 1389-1391 (1996)
  26. Lundblad, R.L. et al., Methods Enzymol., 45, 156 (1976)
  27. Matsuoka, S., et al., JP. J. Pharmacol., 51, 455-463 (1989)
  28. Wimen, B., Meth. Enzymol., 80, 395-408 (1981)
  29. Magnusson, S. The Enzymes, 3rd ed., III, pp. 277-321, Boyer, P.D., ed., Academic Press (1971)