酶试剂
人白蛋白
Gene ID: ALB
CAS#: 70024-90-7
制备方法:
Sigma-Aldrich使用经由传统Cohn法发展而来的冷醇分馏法以及热休克法来生产人血清白蛋白。
分子量:
66,437 Da8(基于氨基酸组分)。商用制剂包含不同程度的翻译后修饰和遗传变异体,分子量大致在66,437 到66,600 Da的范围内。
完整未变性人血清白蛋白的去卷积质谱
Methods of Preparation:
We produce human serum albumin using a cold alcohol fractionation process derived from the traditional Cohn1,3 method as well as heat shock methods.2,4
Molecular Weight:
66,437 Da8 (based on amino acid composition). Commercial preparations contain varying degrees of post-translational modifications and genetic variants with molecular weight components mainly in the range of 66,437 to 66,600 Da
Deconvoluted mass spectrum of intact Native HSA