互作代谢途径图
该新版标志性IUBMB-Sigma-Nicholson代谢途径图为此标准工具带来了更多的功能。现在,所有的代谢物、酶和被选定的途径都是可被搜索和实现互动的。该图的“骨架”是糖酵解途径,随后是TCA(Krebs)循环和呼吸链,它们一起参与了ATP合成酶合成ATP的过程。ATP是所有生命需要的大部分能量的来源。代谢的许多生物合成和分解途径如碳水化合物、氨基酸、脂质与这种骨架相关并可通过使用颜色进行区分。在可能的情况下,使用黑色箭头代替人体中的代谢。大约550个反应可通过IUBMB酶委员会的(EC)编号而进行鉴定,并将其编入索引。
Interactive Metabolic Pathways Map