plant Profiler
黄芪(Astragalus membranaceous)
Astragalus (Astragalus membranaceus) Image
同义词/常用名/相关术语
黄芪皂苷I-IV,Astragalus gummiferaAstragalus lentiginosusAstragalus membranaceusAstragalus membranaceus japonica, Astragalus mollissimusAstragalus mongholicus,黄芪多糖,Astragalus propinquus,黄芪皂苷,Astragalus trigonus,astragel, baak kei, beg kei, bei qi, buck qi,毛蕊异黄酮,豆科植物(家族),芒硝,gaba-aminobytyric酸,山羊角,山羊刺,绿龙,龙牙胶,黄芪胶,黄芪胶,hoang ky, hog gum, Huang Qi, huang-chi, huangoi, huangqi, hwanggi,异黄酮,ji cao,豆科(家族),疯草,膜状紫云英,紫云英,紫云英,蒙古乳,蒙古紫云英,内蒙黄芪,ogi,芒柄花甙,ougi,膜吸虫,黄芪,spino santo,叙利亚黄蓍胶,tai shen, 黄蓍胶,trigonosides I-III, wong kei, yellow leader,黄野豌豆,Zhongfengnaomitong
作用机制
药理:
  • 成分:黄芪的活性成分已得到鉴定和分析,主要包括:多糖环阿屯糖苷级分(黄芪苷I-IV和三萜苷I-III)、四种主要异黄酮(芒柄花素,黄酮苷,毛蕊异黄酮及其糖苷)、皂苷、几种次要异黄酮和其他生物胺。25,26,27,28,29,8,30,31 黄芪还含有三萜类化合物。三萜类和皂苷类与类固醇激素前体具有结构相似性。
  • 抗糖尿病作用:动物和人体研究表明黄芪中的皂苷、异黄酮和多糖可以降低血糖。24,5,17,32,33,6,7基于实验研究,从黄芪中分离的皂苷可抑制蛋白酪氨酸磷酸化酶和蛋白激酶C活化,以及改善F-肌动蛋白重排,从而改变胰岛素敏感性。30,34 一些研究人员已经观察到体外葡萄糖细胞摄取的增加和脂肪细胞中蛋白质的合成增加。35,4黄芪中的异黄酮包括formononectin和鹰嘴豆素A是过氧化物酶体增殖物激活受体(PPAR)和用于恢复血糖平衡的受体的有效激活剂。33
  • 抗炎症作用:体内抗炎活性可以通过抑制NF-κB的活化和mRNA的表达而介导。36,37
  • 抗微生物作用:黄芪多糖已被证明可在动物和人体内诱导内源性干扰素的产生并增强干扰素在病毒感染中的作用。38,9,22,2与未经黄芪预处理的动物相比,用黄芪预处理并暴露于Coxsachie B3、日本脑炎病毒、NDV和仙台病毒的小鼠具有显着更高的干扰素水平和巨噬细胞产量。39,21一些研究人员也报道说,作为对于黄芪的反应,病毒性心肌炎小鼠可表现出总体转归和心脏功能的改善,以及减少的心肌损伤、病毒载量和病毒复制。40,41,20,10,42另外也有报道表明黄芪多糖组分可刺激家禽细菌感染引起的IgA、IgM、IgG和抗原特异性脾细胞的产生。14,15,16相反,黄芪没有表现出抗HIV感染的体外细胞毒性或抗病毒活性。
  • 抗肿瘤特性/化疗副作用的改善:有体外证据表明黄芪通过刺激免疫系统可以增强疗效并减少化疗的不良反应。18,19,2Wei等人报道黄芪提取物显着增加了肺癌患者血液单核细胞的体外细胞因子分泌和基因表达。44也有研究显示黄芪提取物可在癌症和HIV患者的淋巴细胞中诱导LAK细胞活性,并刺激淋巴细胞对各种癌症患者中的诱变剂的胚细胞应答。45,46,47,48,49,50Lau等人也报道含有黄芪的草药组合可在体内抑制肿瘤生长,同时增强巨噬细胞和淋巴因子激活的杀伤细胞活性。51与用常规化疗治疗的大鼠相比,用含黄芪草药的治疗也可抑制大鼠的肿瘤生长和生存期延长。其他一些作者也报道了动物模型中白细胞介素-2(IL-2)产生的细胞毒性的增强45,52和诱变的减少53
  • 抗氧化和细胞保护特性:黄芪中的成分,特别是类黄酮,可发挥显着的细胞抗氧化作用,在预防心血管、肝、肺和肾脏的病变中似乎可发挥作用。54,55据报道,黄芪在体外可保护肾小管免受由高能冲击波56和辐射损伤所引起并由自由基所介导的损伤23。在健康和具有心血管疾病的动物模型心脏中,黄芪多糖和类黄酮组分被报道可清除线粒体自由基,防止脂质过氧化,保护线粒体免受异丙肾上腺素所诱导的损伤,预防糖尿病引起的心肌肥大,并减弱在充血性心力衰竭、糖尿病和实验性高血压中心脏结构和功能的病理性衰退。57,58,59,11,60,61已有研究表明黄芪多糖和类黄酮提取物在肾功能衰竭动物模型中保护肾脏的功能,以及保护因肝毒素而引起的肝功能。56,62,63,4,64,65,66类似的是,黄芪似乎可以保护细胞免受缺氧所带来的挑战69。黄芪甲苷IV被报道在失血性休克后可在体外保护人内皮细胞和肠上皮细胞免于再灌注损伤。67在短暂的局部性缺血后,培养的神经元68和小鼠大脑中也可观察到细胞对缺氧的保护作用69。黄芪苷IV还可抑制人类和动物疾病模型中的血小板聚集、纤溶酶原活化降低以及促进血纤维蛋白凝块的血管内溶解。18,70Xuan等人报道在一种耳毒性模型中黄芪可通过增强的DNA和RNA合成来保护细胞免受病理损伤。65在一项动物研究中,黄芪在实验性结肠炎中具有预防和治疗的潜力。1抗炎作用涉及抗氧化以及抑制结肠组织中的粘附分子合成。在一项体外研究中,黄芪和黄芪皂苷可通过一氧化氮调节途径有效地保护内皮依赖性舒张反应高半胱氨酸引起的急性损伤,而这其中抗氧化作用发挥了关键作用。31
  • 心血管作用:实验研究表明黄芪总苷会影响动物和人体在细胞、分离组织和全身水平的心血管功能。在孤立工作的大鼠心脏中,Wang等人报道黄芪提取物在较高剂量下可产生正性肌力作用,在较低剂量下产生负性肌力作用。52黄芪还可通过外周血管舒张作用在健康和高血压动物中产生降血压作用。71,72Chen等人报道黄芪可显着降低大鼠肾性高血压模型的动脉压,并改善了实验性充血性心力衰竭的心功能。73这种作用也被Ma等人所证实,他们报道黄芪可提高左心室的收缩功能,并在实验性诱导的慢性心力衰竭大鼠中增强因心房利钠肽而引起的强心、利尿和钠利尿反应。13另有体外证据表明黄芪还可以刺激缺血心肌66和羊膜74的血管生成和血运重建,促进血管内皮细胞增殖和DNA合成74。黄芪也被研究表明可增加红细胞中的钠泵活性。75
  • 激素/生殖作用:黄芪对各种激素系统均有显着影响。据报道,相对于对照,黄芪苷可显着增加大鼠垂体细胞培养物中的生长激素释放。12黄芪多糖成分还被证明可防止去卵巢大鼠的骨质流失。76 已有研究证明可在健康大鼠和男性中显著增强体外精子活力和存活力。77据报道,黄芪还能抑制由镉引起的精子生成和运动性衰退。25Liu等人最近报道了黄芪提取物显着增加了30名不育男性的体外精子活力和活力。78
  • 免疫系统作用:据报道,黄芪黄酮能增加豚鼠79和小鼠体内细胞介导的免疫力42。黄芪通过增加辅助性T细胞2型细胞因子的数量,增加肿瘤坏死因子和白细胞介素-6的水平来增强细胞介导的免疫。80,81,48,82,83,84已有研究表明其可刺激巨噬细胞和自然杀伤细胞的活性。根据动物研究,相对于对照,黄芪可增强针对对细菌感染的细胞和体液免疫应答。从患有下肢缺血的患者采集的骨髓血液和服用黄芪的患者的CD34 +细胞比例显着增加(p <0.05)。
药代动力学:
  • 黄芪苷似乎可通过与细胞膜受体直接相互作用发挥其免疫调节作用。Shao等人报道黄芪多糖组分可激活小鼠B细胞并通过细胞表面上的多糖组分和TLR4之间的直接相互作用刺激巨噬细胞产生。35Hao等人在大鼠内皮细胞中证实了黄芪多糖组分可通过促进细胞表面I-1的表达来刺激嗜中性粒细胞和内皮细胞之间的粘附。87这种作用被认为会促进伤口愈合中的炎症反应并可能是在几种组织中观察到的黄芪多糖的解毒和细胞保护作用的基础。已知黄芪多糖可增加各种组织中的RNA活性和蛋白质合成,并增加调节特定细胞代谢功能的酶的活性。2已有研究表明黄芪可调节鸟氨酸脱羧酶活性并增加2型糖尿病动物模型中肝脏和骨骼肌中胰岛素诱导的蛋白酪氨酸磷酸化酶活性。34
  • 黄芪单一化学成分的药代动力学参数数据不足。
参考文献
  1. Ko, J. K., Lam, F. Y., and Cheung, A. P. Amelioration of experimental colitis by Astragalus membranaceus through anti-oxidation and inhibition of adhesion molecule synthesis. World J Gastroenterol 10-7-2005;11(37):5787-5794. 16270386
  2. Zee-Cheng RK. Shi-quan-da-bu-tang (ten significant tonic decoction), SQT. A potent Chinese biological response modifier in cancer immunotherapy, potentiation and detoxification of anticancer drugs. Methods Find Exp Clin Pharmacol 1992;14(9):725-736. 1294861
  3. Zhao, K. S., Mancini, C., and Doria, G. Enhancement of the immune response in mice by Astragalus membranaceus extracts. Immunopharmacology 1990;20(3):225-233. 2149722
  4. Wang, H. Y. and Chen, Y. P. [Clinical observation on treatment of diabetic nephropathy with compound fructus arctii mixture]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2004;24(7):589-592. 2149722
  5. Liu, Y. H., Yang, L., and Liu, J. [Clinical observation on treatment of early diabetic nephropathy by milkvetch injection combined with captopril]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2005;25(11):993-995. 16355614
  6. Wu, Y., Ou-Yang, J. P., Wu, K., Wang, Y., Zhou, Y. F., and Wen, C. Y. Hypoglycemic effect of Astragalus polysaccharide and its effect on PTP1B. Acta Pharmacol Sin 2005;26(3):345-352. 15715932
  7. Zhang, D. Q., Zhang, J. J., Wang, J. X., Hu, S. M., Li, W., Ou, L. N., Chen, S. H., and Gao, X. M. [Effects of Qilan Tangzhining capsule on glucose and lipid metabolism in rats with diabetes mellitus and hyperlipemia]. Zhongguo Zhong Yao Za Zhi 2005;30(10):773-777. 16075719
  8. Shibata T, Sakai E, Nishi K, and Asetai M. Growth and glycoside content of Astragalus membranaceus (Bunge Leguminaceae) cultivated in different soil groups. Natural Medicine 1996;50:296-299.
  9. Qian, Z. W., Mao, S. J., Cai, X. C., Zhang, X. L., Gao, F. X., Lu, M. F., Shao, X. S., Li, Y. Y., Yang, X. K., Zhuo, Y. A., and . Viral etiology of chronic cervicitis and its therapeutic response to a recombinant interferon. Chin Med J (Engl) 1990;103(8):647-651. 2173655
  10. Yang Y, Jin P, Guo Q, and et al. Effect of Astragulas membranaceus on natural killer cell activity and induction of alpha- and gamma- interferon in patients with coxsackie B viral myocarditis. Chinese Medical Journal 1990;103(4):304-307.
  11. Li, C., Cao, L., and Zeng, Q. Astragalus prevents diabetic rats from developing cardiomyopathy by downregulating angiotensin II type2 receptors' expression. J Huazhong Univ Sci Technolog Med Sci 2004;24(4):379-384. 15587404
  12. Kim, C., Ha, H., Kim, J. S., Kim, Y. T., Kwon, S. C., and Park, S. W. Induction of growth hormone by the roots of Astragalus membranaceus in pituitary cell culture. Arch Pharm Res 2003;26(1):34-39. 12568355
  13. Ma, J., Peng, A., and Lin, S. Mechanisms of the therapeutic effect of astragalus membranaceus on sodium and water retention in experimental heart failure. Chin Med J (Engl) 1998;111(1):17-23. 10322646
  14. Guo, F. C., Kwakkel, R. P., Williams, B. A., Li, W. K., Li, H. S., Luo, J. Y., Li, X. P., Wei, Y. X., Yan, Z. T., and Verstegen, M. W. Effects of mushroom and herb polysaccharides, as alternatives for an antibiotic, on growth performance of broilers. Br Poult Sci 2004;45(5):684-694. 15623224
  15. Guo, F. C., Kwakkel, R. P., Williams, B. A., Parmentier, H. K., Li, W. K., Yang, Z. Q., and Verstegen, M. W. Effects of mushroom and herb polysaccharides on cellular and humoral immune responses of Eimeria tenella-infected chickens. Poult Sci 2004;83(7):1124-1132. 15285503
  16. Guo, F. C., Williams, B. A., Kwakkel, R. P., Li, H. S., Li, X. P., Luo, J. Y., Li, W. K., and Verstegen, M. W. Effects of mushroom and herb polysaccharides, as alternatives for an antibiotic, on the cecal microbial ecosystem in broiler chickens. Poult Sci 2004;83(2):175-182. 14979567
  17. Lu, D. C., Su, Z. J., and Rui, T. [Effect of jian yan ling on serum lipids, apoprotein and lipoprotein-a]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1994;14(3):142-144, 131-2. 7950180
  18. Xu, C. J., Jian, X. C., and Cheng, H. Q. [Influence of astragalus membranaceus injection on the proliferation of rabbit bone marrow stromal cells and differentiation to osteoblast]. Zhong Nan Da Xue Xue Bao Yi Xue Ban 2004;29(4):489-491. 16134616
  19. Li, N. Q. [Clinical and experimental study on shen-qo injection with chemotherapy in the treatment of malignant tumor of digestive tract]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1992;12(10):579, 588-592. 1302539
  20. Tianqing P, Yingzhen Y, Riesemann H, and et al. The inhibitory effect of Astragalus membranaceus on coxsackie B-3 virus RNA replication. Chinese Medical Sciences Journal 1995;10(3):146-150.
  21. Yan, H. J. [Clinical and experimental study of the effect of kang er xin-I on viral myocarditis]. Zhong Xi Yi Jie He Za Zhi 1991;11(8):468-70, 452. 1659497
  22. Yunde H, Guoliang M, Shuhua W, and et al. Effects of radix astragali seu-hedysari on teh interferon system. Chin Med J 1981;94:35-40.
  23. Kim SH, Lee SE, Oh H, and et al. The radioprotective effects of bu-zhong-yi-qi-tang: a prescription of traditional Chinesemedicine astragalus. J Chin Med 2002;30(1):127-137.
  24. Jia, W., Gao, W. Y., and Xiao, P. G. Antidiabetic drugs of plant origin used in China: compositions, pharmacology, and hypoglycemic mechanisms. Zhongguo Zhong Yao Za Zhi 2003;28(2):108-113. 15015278
  25. Xiao, H. B., Krucker, M., Albert, K., and Liang, X. M. Determination and identification of isoflavonoids in Radix astragali by matrix solid-phase dispersion extraction and high-performance liquid chromatography with photodiode array and mass spectrometric detection. J Chromatogr A 4-2-2004;1032(1-2):117-124. 15065786
  26. Wang, Q. J., Lu, W. W., Lu, H., Liu, F., Yang, S. J., Hua, Y. Q., and Ji, S. X. [Protective effect of qi dong yi xin on acute myocardial infarction in dogs]. Zhongguo Zhong Yao Za Zhi 2003;28(5):449-452. 15139132
  27. Kitagawa I, Wang H, Saito M, and et al. Chemical constituents of astragali radix, the root of Astragalus membranaceus bunge. Astragalosides, I, II, IV, acetylastragoloside I and isoastragalosides. Chem Pharm Bull 1983;31:698.
  28. He Z and Findlay JA. Constituents of Astragalus membranaceus. Journal of Natural Products 1991;54(3):810-815.
  29. Ma Y, Tian Z, Kuang H, and et al. Studies of the constituents of Astragalus membranaceus BUNGE. III. Structures of triterpenoidal glycosides, huangqiyenins A and B, from the leaves. Chem Pharm Bull 1997;45(2):359-361.
  30. Li, H. B., Ge, Y. K., Zhang, L., and Zheng, X. X. Astragaloside IV improved barrier dysfunction induced by acute high glucose in human umbilical vein endothelial cells. Life Sci 8-15-2006;79(12):1186-1193. 16650877
  31. Zhang, B. Q., Hu, S. J., Qiu, L. H., Zhu, J. H., Xie, X. J., Sun, J., Zhu, Z. H., Xia, Q., and Bian, K. Effects of Astragalus membranaceus and its main components on the acute phase endothelial dysfunction induced by homocysteine. Vascul Pharmacol 2007;46(4):278-285. 17196887
  32. Park JH, Kim BK, Park MK, and et al. Anti-diabetic activity of herbal drugs. Kor J Pharmacogn 1997;28(2):72-74.
  33. Shen, P., Liu, M. H., Ng, T. Y., Chan, Y. H., and Yong, E. L. Differential effects of isoflavones, from Astragalus membranaceus and Pueraria thomsonii, on the activation of PPARalpha, PPARgamma, and adipocyte differentiation in vitro. J Nutr 2006;136(4):899-905. 16549448
  34. Yong WU, Yand OU, Ping J, Ke WU, and et al. Hypoglycemic effect of astragalus polysaccharide and its effect on PTP1B. Acta Pharmacol Sin 2005;26(2):345-352.
  35. Shao, B. M., Xu, W., Dai, H., Tu, P., Li, Z., and Gao, X. M. A study on the immune receptors for polysaccharides from the roots of Astragalus membranaceus, a Chinese medicinal herb. Biochem Biophys Res Commun 8-6-2004;320(4):1103-1111. 15249203
  36. Zhang, H. M., Chen, S. W., Xie, C. G., Xie, Y. Q., Deng, X. F., and Wang, F. [Effects and mechanism of ShenQi compound recipe on inflammation maker in GK rats]. Zhong Yao Cai 2006;29(3):249-253. 16850723
  37. Zhang, W. J., Hufnagl, P., Binder, B. R., and Wojta, J. Antiinflammatory activity of astragaloside IV is mediated by inhibition of NF-kappaB activation and adhesion molecule expression. Thromb Haemost 2003;90(5):904-914. 14597987
  38. Hou, Y. D., Ma, G. L., Wu, S. H., Li, Y. Y., and Li, H. T. Effect of Radix Astragali seu Hedysari on the interferon system. Chin Med J (Engl) 1981;94(1):35-40. 6164521
  39. Kajimura, K., Takagi, Y., Ueba, N., Yamasaki, K., Sakagami, Y., Yokoyama, H., and Yoneda, K. Protective effect of astragali radix by oral administration against Japanese encephalitis virus infection in mice. Biol Pharm Bull 1996;19(9):1166-1169. 8889035
  40. Guo, Q., Peng, T. Q., and Yang, Y. Z. [Effect of Astragalus membranaceus on Ca2+ influx and coxsackie virus B3 RNA replication in cultured neonatal rat heart cells]. Zhongguo Zhong Xi Yi ie He Za Zhi 1995;15(8):483-485. 8580697
  41. Peng, T. Q., Yang, Y. Z., and Kandolf, R. [Effect and mechanism of Astragalus membranaceus on coxsackie B3 virus RNA in mice]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1994;14(11):664-666. 7703635
  42. Yuan, W. L., Chen, H. Z., Yang, Y. Z., Yang, X. Y., Lin, Y. S., Jin, P. Y., and Zhou, T. S. Effect of astragalus membranaceus on electric activities of cultured rat beating heart cells infected with Coxsackie B-2 virus. Chin Med J (Engl) 1990;103(3):177-182. <2164463/a> 1371029
  43. Yao, X. J., Wainberg, M. A., and Parniak, M. A. Mechanism of inhibition of HIV-1 infection in vitro by purified extract of Prunella vulgaris. Virology 1992;187(1):56-62.
  44. Wei, H., Sun, R., Xiao, W., Feng, J., Zhen, C., Xu, X., and Tian, Z. Traditional Chinese medicine Astragalus reverses predominance of Th2 cytokines and their up-stream transcript factors in lung cancer patients. Oncol Rep 2003;10(5):1507-1512. 12883732
  45. Chu, D. T., Lepe-Zuniga, J., Wong, W. L., LaPushin, R., and Mavligit, G. M. Fractionated extract of Astragalus membranaceus, a Chinese medicinal herb, potentiates LAK cell cytotoxicity generated by a low dose of recombinant interleukin-2. J Clin Lab Immunol 1988;26(4):183-187. 3264344
  46. Chu, D., Sun, Y., Lin, J., Wong, W., and Mavligit, G. [F3, a fractionated extract of Astragalus membranaceus, potentiates lymphokine-activated killer cell cytotoxicity generated by low-dose recombinant interleukin-2]. Zhong Xi Yi Jie He Za Zhi 1990;10(1):34-6, 5. 2350835
  47. Chu, D. T., Lin, J. R., and Wong, W. [The in vitro potentiation of LAK cell cytotoxicity in cancer and aids patients induced by F3--a fractionated extract of Astragalus membranaceus]. Zhonghua Zhong Liu Za Zhi 1994;16(3):167-171. 7956691
  48. Jin, R., Wan, L. L., Mitsuishi, T., Kodama, K., and Kurashige, S. [Immunomodulative effects of Chinese herbs in mice treated with anti- tumor agent cyclophosphamide]. Yakugaku Zasshi 1994;114(7):533-538. 7932098
  49. Sun Y, Hersh EM, Talpax M, and et al. Immune restoration and/or augmentation of local graft versus host reaction by traditional Chinese medicinal herbs. Cancer 7-1-1983;52(1):70-73. 6336578
  50. Sun Y, Hersh EM, Lee S, and et al. Preliminary observations on the effects of the Chinese medicinal herbs Astragalus membranaceus and Ligustrum lucidum on lymphocyte blastogenic responses. Journal of Biological Response Modifiers 1983;2(3):227-237. 6644339
  51. Lau, B. H., Ruckle, H. C., Botolazzo, T., and Lui, P. D. Chinese medicinal herbs inhibit growth of murine renal cell carcinoma. Cancer Biother 1994;9(2):153-161. 7812364
  52. Wang, Q. [Inotropic action of Astragalus membranaceus Bge. saponins and its possible mechanism]. Zhongguo Zhong Yao Za Zhi 1992;17(9):557-559 (inside backcover). 1292500
  53. Wong, B. Y., Lau, B. H., Tadi, P. P., and Teel, R. W. Chinese medicinal herbs modulate mutagenesis, DNA binding and metabolism of aflatoxin B1. Mutat Res 6-1-1992;279(3):209-216. 1377337
  54. Shirataki Y, Takao M, Yoshida S, and et al. Antioxidative components isolated from the roots of Astragalus membranaceus Bunge (Astragali Radix) Phytotherapy Research 1997;11:603-605.
  55. Xu, X. Y., Li, L. H., Wu, L. S., Zhao, C. L., and Lin, H. Y. [Adjustment effect of Radix Astragalus and Radix Angelicae sinensis on TNF-alpha and bFGF on renal injury induced by ischemia reperfusion in rabbit]. Zhongguo Zhong Yao Za Zhi 2002;27(10):771-773. 12776561
  56. Sheng, B. W., Chen, X. F., Zhao, J., He, D. L., and Nan, X. Y. Astragalus membranaceus reduces free radical-mediated injury to renal tubules in rabbits receiving high-energy shock waves. Chin Med J (Engl) 1-5-2005;118(1):43-49. 15642225
  57. Guan, Y., Zhou, Z., Zhang, C., Ruan, Y., and Wu, Z. Effect of A-L tonic capsule on DNA content in rat experimental hepatocarcinogenesis. J Huazhong Univ Sci Technolog Med Sci 2002;22(1):12-3, 16. 12658771
  58. Han, L. and Chen, K. J. [The protective effects of yi-xin-kang capsule on the structure and function of rat myocardial mitochondria]. Zhongguo Zhong Yao Za Zhi 2001;26(11):773-777. 12776352
  59. Hong CY, Lo YC, Tan FC, and et al. Astragalus membranaceus and Polygonum multiflorum protect rat heart mitochondria against lipid peroxidation. American Journal of Chinese Medicine 1994;22(1):63-70. 8030620
  60. Toda, S., Yase, Y., and Shirataki, Y. Inhibitory effects of astragali radix, crude drug in Oriental medicines on lipid peroxidation and protein oxidative modification of mouse brain homogenate by copper. Phytother Res 2000;14(4):294-296. 10861978
  61. Wang, D., Shen, W., Tian, Y., Dong, Z., Liu, G., Sun, Z., Yang, S., and Zhou, S. [Protective effect of total flavonoids of radix Astragali on mammalian cell damage caused by hydroxyl radical]. Zhongguo Zhong Yao Za Zhi 1995;20(4):240-2. 7646797
  62. Wang, D. Q., Critchley, J. A., Ding, B. G., Ma, Y. Q., Zhao, H. L., Thomas, G. N., Tian, Y. P., Tomlinson, B., Critchley, L. A., and Edward, J. A. [Protection against paracetamol-induced hepatic damage using total flavonoids of Astragalus]. Zhongguo Zhong Yao Za Zhi 2001;26(9):617-620. 12776431
  63. Wang, D. Q., Ding, B. G., Ma, Y. Q., Zhao, H. L., Neil, T. G., Brian, T., Tian, Y. P., Wang, C. B., and Critchley, J. A. [Studies on protective effect of total flavonoids of Astragalus on liver damage paracetamol-inducedby]. Zhongguo Zhong Yao Za Zhi 2001;26(7):483-486. 12776364
  64. Wu, L., Liu, H., Xue, P., Lu, Z. G., and Du, K. F. [Influence of a triplex superimposed treatment on HBV replication and mutation during treating chronic hepatitis B]. Zhonghua Shi YanHe Lin Chuang Bing Du Xue Za Zhi 2001;15(3):236-238. 11986694
  65. Xuan, W., Dong, M., and Dong, M. Effects of compound injection of Pyrola rotundifolia L and Astragalus membranaceus Bge on experimental guinea pigs' gentamicin ototoxicity. Ann Otol Rhinol Laryngol 1995;104(5):374-380. 7747908
  66. Zhou, J. Y., Fan, Y., Kong, J. L., Wu, D. Z., and Hu, Z. B. [Effects of components isolated from Astragalus membranaceus Bunge on cardiac function injured by myocardial ischemia reperfusion in rats]. Zhongguo Zhong Yao Za Zhi 2000;25(5):300-302. 12512456
  67. Hei, Z. Q., Zhang, J. J., Lin, S. Q., Liu, K. X., and Chen, B. X. [Effects of Astragalus membranaceus injection on nitric oxide and endothelin concentration of intestinal mucosa after hemorrhage shock-reperfusion in rats]. Zhongguo Zhong Yao Za Zhi 2004;29(5):444-447. 15706900
  68. He, X., Li, C., and Yu, S. Protective effects of radix astragali against anoxic damages to in vitro cultured neurons. J Tongji Med Univ 2000;20(2):126-127. 12845724
  69. Luo, Y., Qin, Z., Hong, Z., Zhang, X., Ding, D., Fu, J. H., Zhang, W. D., and Chen, J. Astragaloside IV protects against ischemic brain injury in a murine model of transient focal ischemia. Neurosci Lett 6-17-2004;363(3):218-223. 15182947
  70. Zhang WJ, Wojta J, and Binder BR. Regulation of the fibrinolytic potential of cultured human umbilical vein endothelial cells: Astragaloside IV downregulates plasminogen activator inhibitor-1 and upregulates tissue-type plasminogen activator expression. J Vasc Res 1997;34:273-280.
  71. Hikino H, Funayama S, and Endo K. Hypotensive principle of Astragalus and Hedysarum roots. Planta Medica 1976;30:297-302.
  72. Ning, L., Chen, C. X., Jin, R. M., Wu, Y. P., Zhang, H. G., Sun, C. L., Song, C. Q., and Hu, Z. B. [Effect of components of dang-gui-bu-xue decoction on hematopenia]. Zhongguo Zhong Yao Za Zhi 2002;27(1):50-53. 12774358
  73. Chen, M. L., Liu, W. J., Wang, C. Y., Zhu, X. M., Yin, J. F., Wang, D. X., and Liu, P. [Effects of yishenjiangyafang on blood pressure and protecting renal function in RPH rats]. Zhongguo Zhong Yao Za Zhi 2003;28(8):746-751. 15015358
  74. Lei, Y., Gao, Q., and Li, Y. S. [Study on effects of Astragalus, Angelica and their combination on vascular endothelial cell proliferation in vitro]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2003;23(10):753-756. 14626189
  75. Jin, C. and Dai, R. H. [Effect of Astragalus membranaceus on erythrocyte sodium content and sodium transport in the coronary heart disease]. Zhong Xi Yi Jie He Za Zhi 1991;11(11):651-3, 643. 1667511
  76. Kim, C., Ha, H., Lee, J. H., Kim, J. S., Song, K., and Park, S. W. Herbal extract prevents bone loss in ovariectomized rats. Arch Pharm Res 2003;26(11):917-924. 14661857
  77. Hong, C. Y., Ku, J., and Wu, P. Astragalus membranaceus stimulates human sperm motility in vitro. Am J Chin Med 1992;20(3-4):289-294. 1471613
  78. Liu, J., Liang, P., Yin, C., Wang, T., Li, H., Li, Y., and Ye, Z. Effects of several Chinese herbal aqueous extracts on human sperm motility in vitro. Andrologia 2004;36(2):78-83. 15084153
  79. Zhuang MX. The effects of polysaccharides of Astragalus membranaceus Codonopsis pilosula and Panax ginseng on some immune functions in guinea-pigs. Zhongguo yaoxue zazhi 1992;27:653-655.
  80. He, J., Li, Y., Wei, S., Guo, M., and Fu, W. [Effects of mixture of Astragalus membranaceus, Fructus Ligustri lucidi and Eclipta prostrata on immune function in mice]. Hua Xi Yi e Da Xue Xue Bao 1992;23(4):408-411. 1304546
  81. Jiao, Y., Wen, J., and Yu, X. [Influence of flavonoid of Astragalus membranaceus's stem and leaves on the function of cell mediated immunity in mice]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1999;19(6):356-358. 11783202
  82. Liang, H., Zhang, Y., and Geng, B. [The effect of astragalus polysaccharides (APS) on cell mediated immunity (CMI) in burned mice]. Zhonghua Zheng Xing Shao Shang Wai Ke Za Zhi 1994;10(2):138-141. 7922816
  83. Yoshida, Y., Wang, M. Q., Liu, J. N., Shan, B. E., and Yamashita, U. Immunomodulating activity of Chinese medicinal herbs and Oldenlandia diffusa in particular. Int J Immunopharmacol 1997;19(7):359-370. 9568540
  84. Zhao, K. W. and Kong, H. Y. [Effect of Astragalan on secretion of tumor necrosis factors in human peripheral blood mononuclear cells]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1993;13(5):263-5, 259. 8219673
  85. Lau BH, Ong P, and Tosk J. Macrophage chemiluminescence modulated by Chinese medicinal herbs Astragalus membranaceus and Ligustrum lucidum. Phytotherapy Research 1989;3(4):148-153.
  86. Yang, B. H., Zhu, L. Q., Zhang, J. Z., Niu, F. L., and Cui, W. [Effects of Astragalus membranaceus and Panax notoginseng on the transformation of bone marrow stem cells and proliferation of EPC in vitro]. Zhongguo Zhong Yao Za Zhi 2005;30(22):1761-1763. 16468374
  87. Hao, Y., Qiu, Q. Y., and Wu, J. [Effect of Astragalus polysaccharides in promoting neutrophil-vascular endothelial cell adhesion and expression of related adhesive molecules]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2004;24(5):427-430. 15199628