柠檬草(香茅)
Lemongrass (Cymbopogon) Image
同义词/常用名/相关术语
Abafado(葡萄牙语),α-柠檬醛,α-松油烯,Andropogon citratus,Andropogon nardus,bai mak nao(老挝语),β-柠檬醛(neral),β-月桂烯,bhustrina(印度),英国印度柠檬草,capim-cidrao,carene-2,caspase-3,Ceylon香茅草,柠檬醛,科钦柠檬香茅,Cymbopogon ambiguus, Cymbopogon citrates, Cymbopogon citratus DC, Cymbopogon excavatus, Cymbopogon flexuosus, Cymbopogon goeringii, Cymbopogon martinii, Cymbopogon nardus, Cymbopogon nardus, Cymbopogon proximus, Cymbopogon schoenanthus (L.), Cymbopogon winterianus, Cymbopogon winterianus Jowitt,东印度柠檬草,erba di limone (意大利语), essência de capim-limão (葡萄牙语),精油,法尼醇,发烧草,香叶醇,天竺葵草,醋酸香叶酯,禾本科(家族),危地马拉柠檬草,Halfa barr, herbe de citron (法语), hierba de limon (西班牙语), 爪哇香茅,柠檬草,柠檬草提取物(LGE),柠檬草有,柠檬草油(LGO),柠檬草茎,柠檬草药,Lg,马达加斯加柠檬草,梅利莎草,MYR,月桂烯,棕榈油,蒎烯,piperitone,檀香科(家族),邻苯二甲酸二甲酯,檀香酸,sera(印度,僧伽罗语),serai(马来语),sere(印度尼西亚语),sereh(印度尼西亚语),苏丹植物区系,takrai(泰语),萜烯beta-myrcene,西印度群岛柠檬草,Zitronengras (德语)。
注意:该综述不包含香茅油或石根。
作用机理
药理学:
  • 成分:柠檬草(Cymbopogon属)的许多不同物种的主要成分之一是柠檬醛(3,7-二甲基-2,6-辛二烯-1-醛)。17,18,19,20,21,22,23,24,25柠檬草有已被发现含有高达75-85%的柠檬醛17,26,21,同时也含有z-柠檬醛,冰片,草蒿,甲基丁香酚,乙酸香叶酯(乙酸3,7-二甲基-2,6-辛二烯-1-醇),香叶醇(某些物种比柠檬醛更高),β-月桂烯(MYR,7 甲基-3-亚甲基-1,6-辛二烯),柠檬烯,胡椒碱,香茅醛,care烯-2,α-萜品烯,蒎烯,法呢醇,邻二甲苯二酚和(+)。11,27,28,29,30,31,32,33,34,35,36,25,37,38,39,40,41,12,42,25来自根部的挥发油含有56.67%的长叶烯(V4)和20.03%的selina-6-en-4-ol。25特别是,一项针对鲁沙香茅的研究分离得到脂肪酸,普通甾醇和16-羟基十五碳-14(z)-烯酸。43
  • 镇痛作用:柠檬草在针对小鼠和大鼠的研究中展现出了具有镇痛的作用。44,45,46腹膜内和皮下注射柠檬草和月桂烯抑制乙酸引起的小鼠扭体反应。.45,46纳洛酮是一种类鸦片拮抗剂,可阻断小鼠的抗伤害作用,表明镇痛作用可能是内源性类鸦片类药物释放的结果。当通过注射角叉菜胶或前列腺素E2诱导痛觉过敏时,口服柠檬草的大鼠可引起剂量依赖性镇痛,但不影响由环AMP诱导的痛觉过敏,表明了作用的外周位点。44镇痛作用不会对大鼠重复注射造成耐受。柠檬草的镇痛成分被确定为月桂烯。44,45
  • 驱虫效果:在体外对鲁沙香茅的驱虫效果进行了测试。47鲁沙香茅中的一种成分香叶醇,被证明具有潜在的驱虫效果并具有66.7 ug/mL的ED50 值。
  • 抗菌效果:柠檬草在多项实验室研究中被证实具有抗菌效果48,49,50,51,52,53,54,55,56,57,58,59,60,61,62,63,64,65,66,67,68,69,70,可针对大肠杆菌、沙门氏菌以及肺炎克雷伯菌等发挥活性。52,53,54,61,65,67柠檬草还被证实可对常见的呼吸道病原体如流感嗜血杆菌、铜绿假单胞菌、金黄色葡萄球菌、化脓性链球菌和金黄色葡萄球菌等发挥作用。55,57,71,69一项研究表明,香茅草可完全抑制幽门螺杆菌的生长而不产生抗性,而在相同条件下克拉霉素已产生抗性。60据报道,柠檬草还可以提高苯氧乙醇抵抗大肠杆菌50,72和金黄色葡萄球菌63的活性。
  • 柠檬草还被发现可引起形态改变,抑制隔膜和球芽的形成,并产生“水泡”或中间体,从而形成异常形状的细胞并发生裂解。61还有研究发现柠檬香草可通过羧基荧光素的渗漏透化细胞质膜。59一项研究表明,柠檬草可能通过影响参与细胞质和细胞代谢的细菌靶标而具有抗菌活性。69
  • 柠檬草也被测试在进行SnCl(2)治疗时抗大肠杆菌野生型(AB 1157)菌株的能力。72在香茅草存在的情况下,SnCl(2)对大肠杆菌存活的影响降低。另一项实验室研究还发现,来自香茅草的精油具有广谱抑制特性,对13种大肠杆菌中的10种具有强活性(MIC在100和500mcg/mL之间):其中3种产肠毒素,2种致肠病,3种具有肠侵袭性,以及2种可产生志贺菌毒素。相似地,C. winterianus可显着抑制2种产肠毒素,1种肠致病性,1种具有肠侵袭性和1种可产生志贺毒素。50
  • 酢浆草饮料是一种玫瑰茄花萼水提取物、橙或菠萝汁和柠檬草的混合物,可对芽孢杆菌属、铜绿假单胞菌、乳酸杆菌属和棒状杆菌属具有抗菌作用,而单独的玫瑰茄花萼水提取物并不具备这些效果。73
  • 抗癌效果:香茅在沙门氏菌突变试验中被发现具有抗致突变特性。74从香茅中提取的柠檬醛可在多项造血系统癌细胞系中诱导死亡性细胞凋亡、DNA断裂以及caspase-3催化活性。20在诱导DNA加合物和异常隐窝灶之前,对大鼠施用柠檬草提取物75,并证实在结肠癌中可显著抑制DNA加合物和异常隐窝的形成。然而,在一项已诱导了肝癌的大鼠研究中,柠檬草提取物仅对肝癌的早期具有作用。76柠檬草的另一种成分柠檬烯也被用于抗肿瘤活性的检测中。
  • 抗真菌作用:柠檬草在实验室研究中被证实具有抗真菌活性77,17,78,79,80,81,82,83,84,85,86,87,88,89,90,91,92,93,94,95,96,97,98,99,100,101,102,103,104,105,106,107
  • 其中的一些真菌包括各种曲霉菌种和白色念珠菌。在一项研究中,柠檬草油能够在7至12天内治疗豚鼠的实验性癣。88该油还被证明可有效防止真菌在食物上生长。针对柠檬草油所具有抗真菌作用机制具有多种不同假设。它对黑曲霉中的菌丝生长、孢子形成以及质膜破裂和线粒体结构解体被证实具有抑制作用。80,85,96香茅草还会导致酵母细胞发生K +渗漏,以及使得酵母细胞膜的组成发生变化。
  • 抗炎症效果:在一项小鼠的药理学研究中,通过每只受试者腹膜内注入5uL的柠檬草油以验证其抗炎特性。108柠檬草油已被证实可通过抑制嗜中性粒细胞积聚来抑制白细胞向腹腔中的招募。在另一项研究中,0.0125至0.1%柠檬草油在体外可抑制肿瘤坏死因子-α(TNF-α)引起的嗜中性粒细胞活化。17
  • 抗疟疾效果:在小鼠中的研究表明柠檬草具有抵抗疟原虫的效果。109,4,110一项研究表明对小鼠每天使用500mg/kg的柠檬草可引起高达86.6%的反应而同时喹啉在每天10mg/kg时可起到100%的效果。另一项小鼠中的研究证实了Cymbopogon giganteus 可对喹啉抵抗的疟疾起到作用。109
  • 抗突变效果:柠檬草的成分已针对环磷酰胺、丝裂霉素C和氯化镍引起的诱变作用进行了抵抗测试。111,112一项在小鼠中的研究表明,柠檬醛可防止因环磷酰胺、丝裂霉素C和氯化镍而引起的核损伤。112
  • 抗氧化效果:柠檬草的抗氧化功能也被评估过。113,114,115,116柠檬草油的主要成分柠檬醛,以60mg/kg的量给予大鼠口服一周。116随后,通过腹腔注射氯化镍这种常见的诱变剂以引起核损伤。柠檬醛显著的抑制了因镍引起的微核形成,而同时其抗氧化活性也在体外得到了测试。柠檬醛的超氧清除活性被观察到具有19ug/mL的EC50值,表明柠檬醛具有针对溴化镍的抗畸变作用。
  • 还使用大鼠肝脏上皮细胞系对柠檬醛对谷胱甘肽S-转移酶(GST)的诱导活性进行了测试,而GST可使多环芳烃解毒。115柠檬醛可剂量和时间依赖性的诱导GST活性。谷胱甘肽(GSH)和蛋白质巯基还可特异性地被异构体香叶醛激活。作者认为这项研究提示了柠檬醛可诱导GST的抗氧化作用。在另一项研究中,Cymbopogon citratus显示出了清除自由基的作用,并在另一项研究中对γ-射线具有抵抗作用,这再次提示了其自由基清除的机理。113,114
  • 抗血小板活性Cymbopogon citratus精油在豚鼠和大鼠的血浆中被证实具有抗血小板活性。117
  • 抗病毒效果:柠檬草油在体外显示出具有抗单纯疱疹病毒1型(HSV-1)的抗病毒活性。118浓度为0.1%的柠檬草油与病毒粒子在4℃直接相互作用24小时,可完全抑制病毒的复制。
  • 心脏影响:Cymbopogon citratus 水提取物可降低心率但不改变雄性大鼠离体心脏的收缩力。14Cymbopogon goeringii挥发油可抑制离体豚鼠乳头肌和心房的收缩并延长功能不应期,表明该物种的抗心律失常作用。15柠檬草油还表现出对分离的、灌注的肠系膜动脉制剂的血管舒张作用,似乎是由不依赖于一氧化氮和非类清蛋白的机制所介导。13,16当向大鼠静脉内给药时,Cymbopogon citratus显示出一些与剂量相关的降血压作用。当大鼠通过口服用药时,还表现出一些弱利尿和抗炎作用。
  • 中枢神经系统影响:对大鼠口服高达相当于人剂量208倍的柠檬草以及高达200mg/kg的柠檬醛,结果显示其对中枢神经系统(CNS)并不存在作为抑制剂、催眠药、精神安定药、抗惊厥药或抗焦虑药的作用。6腹膜注射对大鼠CNS几乎没有影响。在另一项研究中,大鼠和小鼠以1g/kg的量口服了含在玉米油中的柠檬香草成分β-月桂烯27。该研究也表明柠檬草对中枢神经系统不存在作用。同时也有其他一些研究对此影响也进行了研究。119,120
  • 降血糖作用:有两项在四氧嘧啶诱导的糖尿病大鼠中进行的研究确定了柠檬草(Cymbopogon proximus)的降血糖特性。两项研究均显示,柠檬草在大鼠的四周治疗中可使葡萄糖水平恢复正常。在一项研究中,四氧嘧啶诱导的糖尿病大鼠每天按每100g体重1.5mL的量口服柠檬草水悬浮液4周。9柠檬草治疗可恢复四氧嘧啶诱导的糖尿病大鼠的葡萄糖、尿素、肌酐、胆红素、总蛋白和白蛋白,降低谷草转氨酶(AST)、丙氨酸转氨酶(ALT)和乳酸脱氢酶(LDH)。在另一项研究中,四氧嘧啶诱导的糖尿病大鼠每天口服柠檬草,四周后使得升高的血糖恢复到了正常水平。10柠檬草处理也降低了细胞色素P450s及其相关的酶活性,如芳烃(苯并(a)芘)羟化酶(AHH)、N-亚硝基二甲胺N-脱甲基酶I(NDMA-dI)、NADPH-细胞色素C还原酶、谷胱甘肽S-转移酶(GST)和谷胱甘肽(GSH)。而这些酶都是在四氧嘧啶诱导的糖尿病大鼠中有所增加。
  • 杀虫剂和驱虫效果:柠檬草已被证实具有驱虫效果。3,121,122,123,124,125,126多项研究表明柠檬草油在两到三个小时的驱虫过程中具有50-95%的有效性。3,122,123,126柠檬草也被证明可以用于杀虫。2,127,128,129,35,36,130,131,132,133,134研究表明柠檬草油是一种可应对多种昆虫如尘螨、白蚁和蜱虫的有效杀虫剂。
  • 其他:柠檬草经测试以确定其对于5-HT释放以及用于头痛治疗的影响。Cymbopogon ambiguus可抑制腺苷酸二磷酸(ADP)介导的血小板5-HT在56-98%水平的释放,并且并不是因为蛋白结合鞣的非特异性影响。这种抑制是剂量依赖性的,并具有0.758mg/mL的IC50值。C. ambiguus 的二氯甲烷(DCM)提取物还可引起由肾上腺素、花生四烯酸介导的血小板激活的显著抑制,而由胶原介导的抑制会更明显。作者表示这些结果表明这个抑制作用可能是发生在花生四烯酸途径内并存在有环氧合酶抑制剂。
  • 一项测试评估了黄曲霉对黄曲霉毒素B(1) (AFB(1))暴露后大鼠血清黄曲霉毒素-白蛋白(AF-白蛋白)加合物水平的影响。135 大鼠每日经5g/kg的Cymbopogon citratus提取物处理一周后,开始经受AFB(1)的暴露。相对于对照组,经Cymbopogon citratus提取物处理的大鼠并没有出现显著的生物标志物水平差异。
  • 一项体外试验发现,Cymbopogon densiflorus 具有显着的抗脱细胞活性(抗镰状细胞性贫血),而这支持了刚果治疗师对于这种草药的传统使用。
  • 一项体外研究发现,Cymbopogon citratus的乙醇提取物对乙酰胆碱酯酶(AChE)、酰胆碱酯酶(BChE)和脂氧合酶(LO)具有酶抑制能力(≥50%)。137
  • 柠檬草植物提取物与氯代氯酸根离子反应后,可产生高比例的纳米技术中使用的薄而扁平的单晶金纳米三角。
药效动力学:
  • 在雄性大鼠中研究了柠檬草油的主要成分柠檬醛(高达80%的柠檬草油成分)的尿代谢物。按500mg/kg的量单次口服柠檬醛24小时后对尿液进行收集。结果发现其在尿中的消除速度很快,在24小时内约有50%的量被排出。柠檬醛被迅速代谢并作为代谢物被排出体外。代谢产物是胆汁葡萄糖醛酸苷和几种酸。这些被分离出来的代谢产物有:3-羟基-3,7-二甲基-6-辛烯二酸;3,8-二羟基-3,7-二甲基-6-辛烯酸;3,9-二羟基-3,7-二甲基-6-辛烯酸;E-和Z-3,7-二甲基-2,6-辛二烯二酸;3,7-二甲基-6-辛烯二酸以及E-3,7-二甲基-2,6-辛二烯酸。柠檬醛的尿代谢物似乎是由代谢途径产生的,而不是对双键的亲核加成。
参考文献
  1. Rogers, K. L., Grice, I. D., and Griffiths, L. R. Modulation of in vitro platelet 5-HT release by species of Erythrina and Cymbopogon. Life Sci 8-31-2001;69(15):1817-1829. 11665843
  2. Bassole, I. H., Guelbeogo, W. M., Nebie, R., Costantini, C., Sagnon, N., Kabore, Z. I., and Traore, S. A. Ovicidal and larvicidal activity against Aedes aegypti and Anopheles gambiae complex mosquitoes of essential oils extracted from three spontaneous plants of Burkina Faso. Parassitologia 2003;45(1):23-26. 15270540
  3. Ansari, M. A. and Razdan, R. K. Relative efficacy of various oils in repelling mosquitoes. Indian J Malariol 1995;32(3):104-111. 8936292
  4. Tchoumbougnang, F., Zollo, P. H., Dagne, E., and Mekonnen, Y. In vivo antimalarial activity of essential oils from Cymbopogon citratus and Ocimum gratissimum on mice infected with Plasmodium berghei. Planta Med 2005;71(1):20-23. 15678368
  5. Carbajal, D., Casaco, A., Arruzazabala, L., Gonzalez, R., and Tolon, Z. Pharmacological study of Cymbopogon citratus leaves. J Ethnopharmacol 1989;25(1):103-107. 2716341
  6. Carlini, E. A., Contar, J. de DP, Silva-Filho, A. R., Silveira-Filho, N. G., Frochtengarten, M. L., and Bueno, O. F. Pharmacology of lemongrass (Cymbopogon citratus Stapf). I. Effects of teas prepared from the leaves on laboratory animals. J Ethnopharmacol 1986;17(1):37-64. 3762195
  7. Rai, A., Singh, A., Ahmad, A., and Sastry, M. Role of halide ions and temperature on the morphology of biologically synthesized gold nanotriangles. Langmuir 1-17-2006;22(2):736-741. 16401125
  8. Shankar, S. S., Rai, A., Ankamwar, B., Singh, A., Ahmad, A., and Sastry, M. Biological synthesis of triangular gold nanoprisms. Nat Mater 2004;3(7):482-488. 15208703
  9. Mansour, H. A., Newairy, A. S., Yousef, M. I., and Sheweita, S. A. Biochemical study on the effects of some Egyptian herbs in alloxan-induced diabetic rats. Toxicology 1-25-2002;170(3):221-228. 11788159
  10. Sheweita, S. A., Newairy, A. A., Mansour, H. A., and Yousef, M. I. Effect of some hypoglycemic herbs on the activity of phase I and II drug-metabolizing enzymes in alloxan-induced diabetic rats. Toxicology 5-24-2002;174(2):131-139. 11985890
  11. NTP Carcinogenesis Studies of Food Grade Geranyl Acetate (71% Geranyl Acetate, 29% Citronellyl Acetate) (CAS No. 105-87-3) in F344/N Rats and B6C3F1 Mice (Gavage Study). Natl Toxicol Program Tech Rep SerM 1987;252:1-162. 12748693
  12. Zamith, H. P., Vidal, M. N., Speit, G., and Paumgartten, F. J. Absence of genotoxic activity of beta-myrcene in the in vivo cytogenetic bone marrow assay. Braz J Med Biol Res 1993;26(1):93-98. 8220273
  13. Abeywardena, M., Runnie, I., Nizar, M., Suhaila, M., and Head, R. Polyphenol-enriched extract of oil palm fronds (Elaeis guineensis) promotes vascular relaxation via endothelium-dependent mechanisms. Asia Pac J Clin Nutr 2002;11 Suppl 7:S467-S472. 12492636
  14. Gazola, R., Machado, D., Ruggiero, C., Singi, G., and Macedo, Alexandre M. Lippia alba, Melissa officinalis and Cymbopogon citratus: effects of the aqueous extracts on the isolated hearts of rats. Pharmacol Res 2004;50(5):477-480. 15458767
  15. Liu, M. and Feng, G. H. [Effects of Cymbopogon goeringii (Steud.) A. Camus volatile oil on physiologic properties of the isolated guinea pig myocardium]. Zhongguo Zhong Yao Za Zhi 1989;14(10):620-3, 640. 2597328
  16. Runnie, I., Salleh, M. N., Mohamed, S., Head, R. J., and Abeywardena, M. Y. Vasorelaxation induced by common edible tropical plant extracts in isolated rat aorta and mesenteric vascular bed. J Ethnopharmacol 2004;92(2-3):311-316. 15138017
  17. Abe, S., Sato, Y., Inoue, S., Ishibashi, H., Maruyama, N., Takizawa, T., Oshima, H., and Yamaguchi, H. [Anti-Candida albicans activity of essential oils including Lemongrass (Cymbopogon citratus) oil and its component, citral]. Nippon Ishinkin Gakkai Zasshi 2003;44(4):285-291. 14615795
  18. Balbaa, S. I. and Johnson, C. H. The microscopic structure of lemongrass leaves. J Am Pharm Assoc Am Pharm Assoc (Baltim) 1955;44(2):89-98. 14353722
  19. Banthorpe, D. V., Duprey, R. J., Hassan, M., Janes, J. F., and Modawi, B. M. Chemistry of the Sudanese flora. I. Essential oils of some Cymbopogon species. Planta Med 1976;29(1):10-25. 1257323
  20. Dudai, N., Weinstein, Y., Krup, M., Rabinski, T., and Ofir, R. Citral is a new inducer of caspase-3 in tumor cell lines. Planta Med 2005;71(5):484-488. 15931590
  21. Rauber, Cda S., Guterres, S. S., and Schapoval, E. E. LC determination of citral in Cymbopogon citratus volatile oil. J Pharm Biomed Anal. 3-9-2005;37(3):597-601. 15740922
  22. Saleem, M., Afza, N., Anwar, M. A., Hai, S. M., and Ali, M. S. A comparative study of essential oils of Cymbopogon citratus and some members of the genus Citrus. Nat Prod Res 2003;17(5):369-373. 14526919
  23. Saleem, M., Afza, N., Anwar, M. A., Hai, S. M., Ali, M. S., Shujaat, S., and Atta, Ur rahman. Chemistry and biological significance of essential oils of Cymbopogon citratus from Pakistan. Nat Prod Res 2003;17(3):159-163. 12737398
  24. Wilson, N. D., Ivanova, M. S., Watt, R. A., and Moffat, A. C. The quantification of citral in lemongrass and lemon oils by near-infrared spectroscopy. J Pharm Pharmacol 2002;54(9):1257-1263. 12356280
  25. Li, H., Huang, J., Zhang, X., Chen, Y., Yang, J., and Hei, L. [Allelopathic effects of Cymbopogon citratu volatile and its chemical components]. Ying Yong Sheng Tai Xue Bao 2005;16(4):763-767. 16011184
  26. Nhu-Trang, T. T., Casabianca, H., and Grenier-Loustalot, M. F. Authenticity control of essential oils containing citronellal and citral by chiral and stable-isotope gas-chromatographic analysis. Anal Bioanal Chem 2006;386(7-8):2141-2152. 17089103
  27. da Silva, V. A., de Freitas, J. C., Mattos, A. P., Paiva-Gouvea, W., Presgrave, O. A., Fingola, F. F., Menezes, M. A., and Paumgartten, F. J. Neurobehavioral study of the effect of beta-myrcene on rodents. Braz J Med Biol Res 1991;24(8):827-831. 1797273
  28. D'Souza, A. M., Paknikar, S. K., Dev, V., Beauchamp, P. S., and Kamat, S. P. Biogenetic-type synthesis of (+)-cymbodiacetal, a constituent of Cymbopogon martinii. J Nat Prod. 2004;67(4):700-702. 15104509
  29. Dubey, V. S., Bhalla, R., and Luthra, R. An esterase is involved in geraniol production during palmarosa inflorescence development. Phytochemistry 2003;63(3):257-264. 12737976
  30. Dubey, V. S., Bhalla, R., and Luthra, R. Sucrose mobilization in relation to essential oil biogenesis during palmarosa (Cymbopogon martinii Roxb. Wats. var. motia) inflorescence development. J Biosci. 2003;28(4):479-487. 12799494
  31. Dubey, V. S. and Luthra, R. Biotransformation of geranyl acetate to geraniol during palmarosa (Cymbopogon martinii, Roxb. wats. var. motia) inflorescence development. Phytochemistry 2001;57(5):675-680. 11397433
  32. Dudai, N., Weinberg, Z. G., Larkov, O., Ravid, U., Ashbell, G., and Putievsky, E. Changes in essential oil during enzyme-assisted ensiling of lemongrass (Cymbopogon citratus Stapf.) and lemon eucalyptus (Eucalyptus citriodora Hook). J Agric Food Chem 2001;49(5):2262-2266. 11368586
  33. Fujita, S. I. and Fujita, Y. [Miscellaneous contributions to the essential oils of the plants from various territories. XXX. On the components of the essential oils of Cymbopogon goeringii A. Camus]. Yakugaku Zasshi 1972;92(10):1285-1288. 4674731
  34. Hegnauer, R. [Microscopical study of the leaves of Cymbopogon flexuosus Stapf (lemon grass).]. Pharm Weekbl 6-18-1955;90(13):445-447. 14394824
  35. Ketoh, G. K., Glitho, A. I., and Huignard, J. Susceptibility of the bruchid Callosobruchus maculatus (Coleoptera: Bruchidae) and its parasitoid Dinarmus basalis (Hymenoptera: Pteromalidae) to three essential oils. J Econ Entomol 2002;95(1):174-182. 11942754
  36. Ketoh, G. K., Koumaglo, H. K., Glitho, I. A., and Huignard, J. Comparative effects of Cymbopogon schoenanthus essential oil and piperitone on Callosobruchus maculatus development. Fitoterapia 7-16-2006;16938411
  37. Marongiu, B., Piras, A., Porcedda, S., and Tuveri, E. Comparative analysis of the oil and supercritical CO(2) extract of Cymbopogon citratus Stapf. Nat Prod Res 5-10-2006;20(5):455-459. 16644543
  38. Matsuda, B. M., Surgeoner, G. A., Heal, J. D., Tucker, A. O., and Maciarello, M. J. Essential oil analysis and field evaluation of the citrosa plant "Pelargonium citrosum" as a repellent against populations of Aedes mosquitoes. J Am Mosq Control Assoc 1996;12(1):69-74. 8723261
  39. Naves, Y. R. and Grampoloff, A. V. [p-Menthadienols in the essential oils of Cymbopogon martini Stapf var. sofia (essence of gingergrass) and of Cymbopogon densiflorus Stapf.]. C.R. Hebd Seances Acad Sci 7-15-1959;249(2):306-308. 13671817
  40. Radwan, A. S. An analytical method for proximadiol, the active principle of Cymbopogon proximus. Planta Med 1975;27(1):93-97. 1161881
  41. Schaneberg, B. T. and Khan, I. A. Comparison of extraction methods for marker compounds in the essential oil of lemon grass by GC. J Agric Food Chem 3-13-2002;50(6):1345-1349. 11879000
  42. Iten, F. and Saller, R. [Fennel tea: risk assessment of the phytogenic monosubstance estragole in comparison to the natural multicomponent mixture]. Forsch Komplementarmed Klass Naturheilkd 2004;11(2):104-108. 15138375
  43. Siddiqi, S. A. and Misra, L. Fatty acids and sterols from Cymbopogon martinii var. motia roots. Z Naturforsch [C] 2000;55(9-10):843-845. 11098842
  44. Lorenzetti, B. B., Souza, G. E., Sarti, S. J., Santos, Filho D., and Ferreira, S. H. Myrcene mimics the peripheral analgesic activity of lemongrass tea. J Ethnopharmacol 1991;34(1):43-48. 1753786
  45. Rao, V. S., Menezes, A. M., and Viana, G. S. Effect of myrcene on nociception in mice. J Pharm Pharmacol 1990;42(12):877-878. 1753786
  46. Viana, G. S., Vale, T. G., Pinho, R. S., and Matos, F. J. Antinociceptive effect of the essential oil from Cymbopogon citratus in mice. J Ethnopharmacol 2000;70(3):323-327. 10837994
  47. Kumaran, A. M., D'Souza, P., Agarwal, A., Bokkolla, R. M., and Balasubramaniam, M. Geraniol, the putative anthelmintic principle of Cymbopogon martinii. Phytother Res 2003;17(8):957. 13680833
  48. Betoni, J. E., Mantovani, R. P., Barbosa, L. N., Di Stasi, L. C., and Fernandes, Junior A. Synergism between plant extract and antimicrobial drugs used on Staphylococcus aureus diseases. Mem Inst Oswaldo Cruz 2006;101(4):387-390. 16951808
  49. Cimanga, K., Kambu, K., Tona, L., Apers, S., De Bruyne, T., Hermans, N., Totte, J., Pieters, L., and Vlietinck, A. J. Correlation between chemical composition and antibacterial activity of essential oils of some aromatic medicinal plants growing in the Democratic Republic of Congo. J Ethnopharmacol 2002;79(2):213-220. 11801384
  50. Duarte, M. C., Leme, E. E., Delarmelina, C., Soares, A. A., Figueira, G. M., and Sartoratto, A. Activity of essential oils from Brazilian medicinal plants on Escherichia coli. J Ethnopharmacol 12-3-2006;17210236
  51. Dorman, H. J. and Deans, S. G. Antimicrobial agents from plants: antibacterial activity of plant volatile oils. J Appl Microbiol 2000;88(2):308-316. 10736000
  52. El Kamali, H. H., Hamza, M. A., and El Amir, M. Y. Antibacterial activity of the essential oil from Cymbopogon nervatus inflorescence. Fitoterapia 2005;76(5):446-449. 15896926
  53. Friedman, M., Henika, P. R., Levin, C. E., and Mandrell, R. E. Antibacterial activities of plant essential oils and their components against Escherichia coli O157:H7 and Salmonella enterica in apple juice. J Agric Food Chem 9-22-2004;52(19):6042-6048. 15366861
  54. Friedman, M., Henika, P. R., and Mandrell, R. E. Bactericidal activities of plant essential oils and some of their isolated constituents against Campylobacter jejuni, Escherichia coli, Listeria monocytogenes, and Salmonella enterica. J Food Prot 2002;65(10):1545-1560. 12380738
  55. Inouye, S., Takizawa, T., and Yamaguchi, H. Antibacterial activity of essential oils and their major constituents against respiratory tract pathogens by gaseous contact. J Antimicrob Chemother 2001;47(5):565-573. 11328766
  56. Inouye, S., Uchida, K., and Yamaguchi, H. In-vitro and in-vivo anti-Trichophyton activity of essential oils by vapour contact. Mycoses 2001;44(3-4):99-107. 11413931
  57. Inouye, S., Yamaguchi, H., and Takizawa, T. Screening of the antibacterial effects of a variety of essential oils on respiratory tract pathogens, using a modified dilution assay method. J Infect Chemother 2001;7(4):251-254. 11810593
  58. Mejlholm, O. and Dalgaard, P. Antimicrobial effect of essential oils on the seafood spoilage micro-organism Photobacterium phosphoreum in liquid media and fish products. Lett Appl Microbiol 2002;34(1):27-31. 11849488
  59. Nguefack, J., Budde, B. B., and Jakobsen, M. Five essential oils from aromatic plants of Cameroon: their antibacterial activity and ability to permeabilize the cytoplasmic membrane of Listeria innocua examined by flow cytometry. Lett Appl Microbiol 2004;39(5):395-400. 15482428
  60. Ohno, T., Kita, M., Yamaoka, Y., Imamura, S., Yamamoto, T., Mitsufuji, S., Kodama, T., Kashima, K., and Imanishi, J. Antimicrobial activity of essential oils against Helicobacter pylori. Helicobacter 2003;8(3):207-215. 12752733
  61. Ogunlana, E. O., Hoglund, S., Onawunmi, G., and Skold, O. Effects of lemongrass oil on the morphological characteristics and peptidoglycan synthesis of Escherichia coli cells. Microbios 1987;50(202):43-59. 3299005
  62. Onawunmi, G. O., Yisak, W. A., and Ogunlana, E. O. Antibacterial constituents in the essential oil of Cymbopogon citratus (DC.) Stapf. J Ethnopharmacol 1984;12(3):279-286. 6442749
  63. Onawunmi, G. O. In vitro studies on the antibacterial activity of phenoxyethanol in combination with lemon grass oil. Pharmazie 1988;43(1):42-44. 3287398
  64. Pattnaik, S., Subramanyam, V. R., Kole, C. R., and Sahoo, S. Antibacterial activity of essential oils from Cymbopogon: inter- and intra-specific differences. Microbios 1995;84(341):239-245. 8643027
  65. Pattnaik, S., Subramanyam, V. R., and Rath, C. C. Effect of essential oils on the viability and morphology of Escherichia coli (SP-11). Microbios 1995;84(340):195-199. 8820244
  66. Raybaudi-Massilia, R. M., Mosqueda-Melgar, J., and Martin-Belloso, O. Antimicrobial activity of essential oils on Salmonella enteritidis, Escherichia coli, and Listeria innocua in fruit juices. J Food Prot 2006;69(7):1579-1586. 16865889
  67. Rojas-Grau, M. A., Avena-Bustillos, R. J., Friedman, M., Henika, P. R., Martin-Belloso, O., and McHugh, T. H. Mechanical, barrier, and antimicrobial properties of apple puree edible films containing plant essential oils. J Agric Food Chem 11-29-2006;54(24):9262-9267. 17117819
  68. Takaisi-Kikuni, N. B., Tshilanda, D., and Babady, B. Antibacterial activity of the essential oil of Cymbopogon densiflorus. Fitoterapia 2000;71(1):69-71. 11449474
  69. Takaisi-Kikuni, N. B., Kruger, D., Gnann, W., and Wecke, J. Microcalorimetric and electron microscopic investigation on the effects of essential oil from Cymbopogon densiflorus on Staphylococcus aureus. Microbios 1996;88(354):55-62. 9121380
  70. Wannissorn, B., Jarikasem, S., Siriwangchai, T., and Thubthimthed, S. Antibacterial properties of essential oils from Thai medicinal plants. Fitoterapia 2005;76(2):233-236. 15752638
  71. Pattnaik, S., Rath, C., and Subramanyam, V. R. Characterization of resistance to essential oils in a strain of Pseudomonas aeruginosa (VR-6). Microbios 1995;81(326):29-31. 7731390
  72. Melo, S. F., Soares, S. F., da Costa, R. F., da Silva, C. R., de Oliveira, M. B., Bezerra, R. J., Caldeira-de-Araujo, A., and Bernardo-Filho, M. Effect of the Cymbopogon citratus, Maytenus ilicifolia and Baccharis genistelloides extracts against the stannous chloride oxidative damage in Escherichia coli. Mutat Res 9-20-2001;496(1-2):33-38. 11551478
  73. Oboh, G. and Elusiyan, C. A. Nutrient composition and antimicrobial activity of sorrel drinks (soborodo). J Med Food 2004;7(3):340-342. 15383229
  74. Vinitketkumnuen, U., Puatanachokchai, R., Kongtawelert, P., Lertprasertsuke, N., and Matsushima, T. Antimutagenicity of lemon grass (Cymbopogon citratus Stapf) to various known mutagens in salmonella mutation assay. Mutat Res 1994;341(1):71-75. 7523944
  75. Suaeyun, R., Kinouchi, T., Arimochi, H., Vinitketkumnuen, U., and Ohnishi, Y. Inhibitory effects of lemon grass (Cymbopogon citratus Stapf) on formation of azoxymethane-induced DNA adducts and aberrant crypt foci in the rat colon. Carcinogenesis 1997;18(5):949-955. 9163680
  76. Puatanachokchai, R., Kishida, H., Denda, A., Murata, N., Konishi, Y., Vinitketkumnuen, U., and Nakae, D. Inhibitory effects of lemon grass (Cymbopogon citratus, Stapf) extract on the early phase of hepatocarcinogenesis after initiation with diethylnitrosamine in male Fischer 344 rats. Cancer Lett 9-8-2002;183(1):9-15. 12049809
  77. Essential oils of peppermint, orange or lemongrass kill most strains of fungal and bacterial infections. Posit Health News 1998;(No 17):26-27. 11366554
  78. Anthony, S., Abeywickrama, K., Dayananda, R., Wijeratnam, S. W., and Arambewela, L. Fungal pathogens associated with banana fruit in Sri Lanka, and their treatment with essential oils. Mycopathologia 2004;157(1):91-97. 15008351
  79. Bankole, S. A., Joda, A. O., and Ashidi, J. S. The use of powder and essential oil of Cymbopogon citratus against mould deterioration and aflatoxin contamination of "egusi" melon seeds. J Basic Microbiol 2005;45(1):20-30. 15678559
  80. de Billerbeck, V. G., Roques, C. G., Bessiere, J. M., Fonvieille, J. L., and Dargent, R. Effects of Cymbopogon nardus (L.) W. Watson essential oil on the growth and morphogenesis of Aspergillus niger. Can J Microbiol 2001;47(1):9-17. 15049444
  81. Duarte, M. C., Figueira, G. M., Sartoratto, A., Rehder, V. L., and Delarmelina, C. Anti-Candida activity of Brazilian medicinal plants. J Ethnopharmacol 2-28-2005;97(2):305-311. 15707770
  82. Fandohan, P., Gbenou, J. D., Gnonlonfin, B., Hell, K., Marasas, W. F., and Wingfield, M. J. Effect of essential oils on the growth of Fusarium verticillioides and fumonisin contamination in corn. J Agric Food Chem 11-3-2004;52(22):6824-6829. 15506822
  83. Guynot, M. E., Ramos, A. J., Seto, L., Purroy, P., Sanchis, V., and Marin, S. Antifungal activity of volatile compounds generated by essential oils against fungi commonly causing deterioration of bakery products. J Appl Microbiol 2003;94(5):893-899. 12694455
  84. Hammer, K. A., Carson, C. F., and Riley, T. V. Antimicrobial activity of essential oils and other plant extracts. J Appl Microbiol 1999;86(6):985-990. 10438227
  85. Helal, G. A., Sarhan, M. M., Abu Shahla, A. N., and Abou El-Khair, E. K. Effects of Cymbopogon citratus L. essential oil on the growth, lipid content and morphogenesis of Aspergillus niger ML2-strain. J Basic Microbiol 2006;46(6):456-469. 17139611
  86. Inouye, S., Tsuruoka, T., Watanabe, M., Takeo, K., Akao, M., Nishiyama, Y., and Yamaguchi, H. Inhibitory effect of essential oils on apical growth of Aspergillus fumigatus by vapour contact. Mycoses 2000;43(1-2):17-23. 10838841
  87. Inouye, S., Watanabe, M., Nishiyama, Y., Takeo, K., Akao, M., and Yamaguchi, H. Antisporulating and respiration-inhibitory effects of essential oils on filamentous fungi. Mycoses 1998;41(9-10):403-410. 9916464
  88. Kishore, N., Mishra, A. K., and Chansouria, J. P. Fungitoxicity of essential oils against dermatophytes. Mycoses 1993;36(5-6):211-215. 8264720
  89. Lima, E. O., Gompertz, O. F., Giesbrecht, A. M., and Paulo, M. Q. In vitro antifungal activity of essential oils obtained from officinal plants against dermatophytes. Mycoses 1993;36(9-10):333-336. 8015567
  90. Mishra, A. K. and Dubey, N. K. Evaluation of some essential oils for their toxicity against fungi causing deterioration of stored food commodities. Appl Environ Microbiol 1994;60(4):1101-1105. 8017906
  91. Nguefack, J., Leth, V., Amvam Zollo, P. H., and Mathur, S. B. Evaluation of five essential oils from aromatic plants of Cameroon for controlling food spoilage and mycotoxin producing fungi. Int J Food Microbiol 8-1-2004;94(3):329-334. 15246244
  92. Palhano, F. L., Vilches, T. T., Santos, R. B., Orlando, M. T., Ventura, J. A., and Fernandes, P. M. Inactivation of Colletotrichum gloeosporioides spores by high hydrostatic pressure combined with citral or lemongrass essential oil. Int J Food Microbiol 8-15-2004;95(1):61-66. 15240075
  93. Paranagama, P. A., Abeysekera, K. H., Abeywickrama, K., and Nugaliyadde, L. Fungicidal and anti-aflatoxigenic effects of the essential oil of Cymbopogon citratus (DC.) Stapf. (lemongrass) against Aspergillus flavus Link. isolated from stored rice. Lett Appl Microbiol 2003;37(1):86-90. 12803563
  94. Pattnaik, S., Subramanyam, V. R., and Kole, C. R. Antifungal activity of essential oils from Cymbopogon: inter- and intraspecific differences. Cytobios 1999;97(386):153-159. 10610298
  95. Pattnaik, S., Subramanyam, V. R., and Kole, C. Antibacterial and antifungal activity of ten essential oils in vitro. Microbios 1996;86(349):237-246. 8893526
  96. Pawar, V. C. and Thaker, V. S. In vitro efficacy of 75 essential oils against Aspergillus niger. Mycoses 2006;49(4):316-323. 16784447
  97. Prashar, A., Hili, P., Veness, R. G., and Evans, C. S. Antimicrobial action of palmarosa oil (Cymbopogon martinii) on Saccharomyces cerevisiae. Phytochemistry 2003;63(5):569-575. 12809717
  98. Qureshi, S., Rai, M. K., and Agrawal, S. C. In vitro evaluation of inhibitory nature of extracts of 18-plant species of Chhindwara against 3-keratinophilic fungi. Hindustan Antibiot Bull 1997;39(1-4):56-60. 10386016
  99. Ramos, Ruiz A., De la Torre, R. A., Alonso, N., Villaescusa, A., Betancourt, J., and Vizoso, A. Screening of medicinal plants for induction of somatic segregation activity in Aspergillus nidulans. J Ethnopharmacol 7-5-1996;52(3):123-127. 8771452
  100. Rolz, C., de Leon, R., de Arriola, M. C., and de Cabrera, S. Biodelignification of Lemon Grass and Citronella Bagasse by White-Rot Fungi. Appl Environ Microbiol  1986;52(4):607-611. 16347155
  101. Sarbhoy, A. K., Varshney, J. L., Maheshwari, M. L., and Saxena, D. B. Efficacy of some essential oils and their constituents on few ubiquitous molds. Zentralbl Bakteriol Naturwiss 1978;133(7-8):723-725. 749414
  102. Suhr, K. I. and Nielsen, P. V. Antifungal activity of essential oils evaluated by two different application techniques against rye bread spoilage fungi. J Appl Microbiol 2003;94(4):665-674. 12631202
  103. Tampieri, M. P., Galuppi, R., Macchioni, F., Carelle, M. S., Falcioni, L., Cioni, P. L., and Morelli, I. The inhibition of Candida albicans by selected essential oils and their major components. Mycopathologia 2005;159(3):339-345. 15883716
  104. Tanu, Prakash, A., and Adholeya, A. Effect of different organic manures/composts on the herbage and essential oil yield of Cymbopogon winterianus and their influence on the native AM population in a marginal alfisol. Bioresour Technol 2004;92(3):311-319. 14766166
  105. Velluti, A., Sanchis, V., Ramos, A. J., Egido, J., and Marin, S. Inhibitory effect of cinnamon, clove, lemongrass, oregano and palmarose essential oils on growth and fumonisin B1 production by Fusarium proliferatum in maize grain. Int J Food Microbiol 12-31-2003;89(2-3):145-154. 14623380
  106. Velluti, A., Sanchis, V., Ramos, A. J., Turon, C., and Marin, S. Impact of essential oils on growth rate, zearalenone and deoxynivalenol production by Fusarium graminearum under different temperature and water activity conditions in maize grain. J Appl Microbiol 2004;96(4):716-724. 15012810
  107. Wannissorn, B., Jarikasem, S., and Soontorntanasart, T. Antifungal activity of lemon grass oil and lemon grass oil cream. Phytother Res 1996;10:551.
  108. Abe, S., Maruyama, N., Hayama, K., Inouye, S., Oshima, H., and Yamaguchi, H. Suppression of neutrophil recruitment in mice by geranium essential oil. Mediators Inflamm 2004;13(1):21-24. 15203560
  109. Kimbi, H. K. and Fagbenro-Beyioku, A. F. Efficacy of Cymbopogon giganteus and Enantia chrantha against chloroquine resistant Plasmodium yoelii nigeriensis. East Afr Med J 1996;73(10):636-637. 8997841
  110. Williams, G. O. and Omoh, L. E. Mitotic effects of the aqueous leaf extract of Cymbopogon citratus in Allium cepa root tips. Cytobios 1996;87(350):161-168. 9172394
  111. Kauderer, B., Zamith, H., Paumgartten, F. J., and Speit, G. Evaluation of the mutagenicity of beta-myrcene in mammalian cells in vitro. Environ Mol Mutagen 1991;18(1):28-34. 1864266
  112. Rabbani, S. I., Devi, K., and Zahra, N. Anti-clastogenic effects of citral. Iranian Journal of Pharmacology and Therapuetics 2005;4:28-31.
  113. Cheel, J., Theoduloz, C., Rodriguez, J., and Schmeda-Hirschmann, G. Free radical scavengers and antioxidants from Lemongrass (Cymbopogon citratus (DC.) Stapf.). J Agric Food Chem 4-6-2005;53(7):2511-2517. 15796587
  114. Fuentes, J. L., Alonso, A., Cuetara, E., Vernhe, M., Alvarez, N., Sanchez-Lamar, A., and Llagostera, M. Usefulness of the SOS Chromotest in the study of medicinal plants as radioprotectors. Int J Radiat Biol 2006;82(5):323-329. 16782649
  115. Nakamura, Y., Miyamoto, M., Murakami, A., Ohigashi, H., Osawa, T., and Uchida, K. A phase II detoxification enzyme inducer from lemongrass: identification of citral and involvement of electrophilic reaction in the enzyme induction. Biochem Biophys Res Commun 3-14-2003;302(3):593-600. 12615076
  116. Rabbani, S. I., Devi, K., Khanam, S., and Zahra, N. Citral, a component of lemongrass oil inhibits the clastogenic effect of nickel chloride in mouse micronucleus test system. Pak J Pharm Sci 2006;19(2):108-113. 16751120
  117. Tognolini, M., Barocelli, E., Ballabeni, V., Bruni, R., Bianchi, A., Chiavarini, M., and Impicciatore, M. Comparative screening of plant essential oils: phenylpropanoid moiety as basic core for antiplatelet activity. Life Sci 2-23-2006;78(13):1419-1432. 16274702
  118. Minami, M., Kita, M., Nakaya, T., Yamamoto, T., Kuriyama, H., and Imanishi, J. The inhibitory effect of essential oils on herpes simplex virus type-1 replication in vitro. Microbiol Immunol 2003;47(9):681-684. 14584615
  119. Kokate, C. K., Rao, R. E., and Varma, K. C. Pharmacological investigations of essential oil of Cymbopogon nardus (L.) Rendle: studies on central nervous system. Indian J Exp Biol 1971;9(4):515-516. 5147185
  120. Seth, G., Kokate, C. K., and Varma, K. C. Effect of essential oil of cymbopogon citratus stapf. on central nervous system. Indian J Exp Biol 1976;14(3):370-371. 992793
  121. Gilbert, B., Teixeira, D. F., Carvalho, E. S., De Paula, A. E., Pereira, J. F., Ferreira, J. L., Almeida, M. B., Machado, Rda S., and Cascon, V. Activities of the Pharmaceutical Technology Institute of the Oswaldo Cruz Foundation with medicinal, insecticidal and insect repellent plants. An Acad Bras Cienc 1999;71(2):265-271. 10412493
  122. Govere, J., Durrheim, D. N., Du, Toit N., Hunt, R. H., and Coetzee, M. Local plants as repellents against Anopheles arabiensis, in Mpumalanga Province, South Africa. Cent Afr J Med 2000;46(8):213-216. 11317593
  123. Oyedele, A. O., Gbolade, A. A., Sosan, M. B., Adewoyin, F. B., Soyelu, O. L., and Orafidiya, O. O. Formulation of an effective mosquito-repellent topical product from lemongrass oil. Phytomedicine 2002;9(3):259-262. 12046869
  124. Scott, I. M., Jensen, H., Scott, J. G., Isman, M. B., Arnason, J. T., and Philogene, B. J. Botanical insecticides for controlling agricultural pests: piperamides and the Colorado Potato Beetle Leptinotarsa decemlineata say (Coleoptera: Chrysomelidae). Arch Insect Biochem Physiol 2003;54(4):212-225. 14635182
  125. Tawatsin, A., Wratten, S. D., Scott, R. R., Thavara, U., and Techadamrongsin, Y. Repellency of volatile oils from plants against three mosquito vectors. J Vector Ecol 2001;26(1):76-82. 11469188
  126. Trongtokit, Y., Rongsriyam, Y., Komalamisra, N., and Apiwathnasorn, C. Comparative repellency of 38 essential oils against mosquito bites. Phytother Res 2005;19(4):303-309. 16041723
  127. Cavalcanti, E. S., Morais, S. M., Lima, M. A., and Santana, E. W. Larvicidal activity of essential oils from Brazilian plants against Aedes aegypti L. Mem Inst Oswaldo Cruz 2004;99(5):541-544. 15543421
  128. de Mendonca, F. A., da Silva, K. F., dos Santos, K. K., Ribeiro Junior, K. A., and Sant'Ana, A. E. Activities of some Brazilian plants against larvae of the mosquito Aedes aegypti. Fitoterapia 2005;76(7-8):629-636. 16253435
  129. Heimerdinger, A., Olivo, C. J., Molento, M. B., Agnolin, C. A., Ziech, M. F., Scaravelli, L. F., Skonieski, F. R., Both, J. F., and Charao, P. S. [Alcoholic extract of lemongrass (Cymbopogon citratus) on the control of Boophilus microplus in cattle]. Rev Bras Parasitol Vet 2006;15(1):37-39. 16647001
  130. Morsy, T. A., Mazyad, S. A., and el Sharkawy, I. M. The larvicidal activity of solvent extracts of three medicinal plants against third instar larvae of Chrysomyia albiceps. J Egypt Soc Parasitol 1998;28(3):699-709. 9914694
  131. Raja, N., Albert, S., Ignacimuthu, S., and Dorn, S. Effect of plant volatile oils in protecting stored cowpea Vigna unguiculata (L.) Walpers against Callosobruchus maculatus (F.) (Coleoptera: Bruchidae) infestation. J Stored Prod Res 2001;37(2):127-132. 11124376
  132. Rim, I. S. and Jee, C. H. Acaricidal effects of herb essential oils against Dermatophagoides farinae and D. pteronyssinus (Acari: Pyroglyphidae) and qualitative analysis of a herb Mentha pulegium(pennyroyal). Korean J Parasitol 2006;44(2):133-138. 16809961
  133. Tiwari, B. K., Bajpai, V. N., and Agarwal, P. N. Evaluation of insecticidal, fumigant and repellent properties of lemongrass oil. Indian J Exp Biol 1966;4(2):128-129. 4380199
  134. Zhu, B. C., Henderson, G., Chen, F., Fei, H., and Laine, R. A. Evaluation of vetiver oil and seven insect-active essential oils against the Formosan subterranean termite. J Chem Ecol 2001;27(8):1617-1625. 11521400
  135. Vinitketkumnuen, U., Chewonarin, T., Dhumtanom, P., Lertprasertsuk, N., and Wild, C. P. Aflatoxin-albumin adduct formation after single and multiple doses of aflatoxin B(1) in rats treated with Thai medicinal plants. Mutat Res 7-16-1999;428(1-2):345-351. 10518006
  136. Mpiana, P. T., Tshibangu, D. S., Shetonde, O. M., and Ngbolua, K. N. In vitro antidrepanocytary actvity (anti-sickle cell anemia) of some congolese plants. Phytomedicine 11-16-2006;17113273
  137. Khattak, S., Saeed, Ur Rehman, Shah, H. U., Khan, T., and Ahmad, M. In vitro enzyme inhibition activities of crude ethanolic extracts derived from medicinal plants of Pakistan. Nat Prod Res 2005;19(6):567-571. 16010821
  138. Diliberto, J. J., Srinivas, P., Overstreet, D., Usha, G., Burka, L. T., and Birnbaum, L. S. Metabolism of citral, an alpha,beta-unsaturated aldehyde, in male F344 rats. Drug Metab Dispos 1990;18(6):866-875. 1981530