样品制备
蛋白去除产品
Protein Depletion enables greater analysis of the proteome. Whether you use one or two dimensional electrophoresis, single or multidimensional chromatography or go straight to mass spectrometry. Sigma offers a complete line of products optimized for the depletion of highly abundant proteins from a variety of biological samples. Not only are a range of depletion media available for your sample of interest; human, mouse, rat or plant. But these are also provided in spin column, LC, or automated formats for your specific application.