不含EDTA的SigmaFAST™蛋白酶抑制剂混合物片剂
 最广谱的蛋白酶抑制
不含EDTA的SigmaFAST™蛋白酶抑制剂混合物片剂 (货号 S8830) 是便利的水溶性蛋白酶抑制剂混合物片剂,可抑制:
  • 丝氨酸蛋白酶
  • 半胱氨酸蛋白酶
  • 天冬氨酸蛋白酶
  • 金属蛋白酶
由于S8830不含EDTA,因此不会干扰下游应用,如纯化组氨酸标签蛋白。
在组织或细胞开始裂解时,提取物充满降解蛋白质的蛋白水解酶。在提取和纯化过程中保护蛋白免受酶促降解至关重要。具有功能性并与随后的下游应用相容的分离蛋白同样重要。该片剂对抑制丝氨酸、半胱氨酸、金属蛋白酶和天冬氨酸(酸性)蛋白酶具有广泛特异性,可用于一般目的或更专门的应用。该混合物以便利的水溶性片剂提供,可在不存在EDTA螯合剂或有机溶剂DMSO的情况下提供优异的保护。
 与镍树脂相容
Sigma-Aldrich的不含EDTA的片剂不会干扰HIS-tag蛋白纯化等下游应用。
通过在溶解的不含EDTA的片剂存在下反复从Sigma的HIS-Select镍树脂中回收HIS标签蛋白来进行测试。作为对照,在PBS(磷酸盐缓冲盐溶液)存在下回收相同的HIS标签蛋白。根据测量的蛋白浓度,两个样品组的重复(4次)回收率相同。
Compatible with Nickel Resin
 与竞品的比较
使用荧光蛋白酶抑制试验进行蛋白酶抑制。
Protease Inhibition using Fluorescent Protease Inhibition Assay
SDS-PAGE 分析显示酶消化具有优越的保护作用。
 SDS-PAGE analysis of enzyme digest showing superior protection
A. 对照,只含BSA
B. BSA, 胃蛋白酶
C. BSA, 胃蛋白酶、SIGMAFAST蛋白酶抑制剂混合物、不含EDTA的片剂
D. BSA, 胃蛋白酶、不含EDTA的竞品片剂
SDS-PAGE analysis of enzyme digest showing superior protection
A. 对照,只含BSA
B. BSA, 胰酶提取物
C. BSA, 胰酶提取物、SIGMAFASTä蛋白酶抑制剂混合物、不含EDTA的片剂
 相关产品
蛋白酶抑制剂混合物
蛋白酶抑制剂混合物
磷酸酶抑制剂混合物
HIS-Select 产品
 参考资料
  1. Tang, Y., et al., Promotion of tumor development in prostate cancer by progerin. Cancer Cell Int., 10(1), 47 (2010). Abstract
  2. Short-chain fatty acids induced autophagy serves as an adaptive strategy for retarding mitochondria-mediated apoptotic cell death. Tang, Y., et al. Cell Death Differ. 18, 602-618, (2011). Abstract
  3. Ali, B.R., et al., Endoplasmic reticulum quality control is involved in the mechanism of endoglin-mediated hereditary haemorrhagic telangiectasia. PLoS ONE, 6(10), e26206 (2011). Abstract
  4. Waugh, J.L., et al., Association between regulator of G protein signaling 9-2 and body weight. PLoS ONE, 6(11), e27984 (2011). Abstract
  5. Reber, S., “Isolation of Centromeres from Cultured Cells”, from Microtubule Dynamics: Methods and Protocols (ed. Anne Straube). Methods in Molecular Biology, Vol. 777 (ISBN 978-1-61779-251-9), pp. 107-116 (2011).
  6. Celver, J., et al., D(2)-Dopamine receptors target regulator of G protein signaling 9-2 to detergent-resistant membrane fractions. J. Neurochem., 120(1), 56-69 (2012). Abstract
  7. Young, E.T., et al., The AMP-activated protein kinase Snf1 regulates transcription factor binding, RNA polymerase II activity, and mRNA stability of glucose-repressed genes in Saccharomyces cerevisiae. J. Biol. Chem., 287(34), 29021-29034 (2012). Abstract
  8. Nitsche, B.M., et al., The carbon starvation response of Aspergillus niger during submerged cultivation: Insights from the transcriptome and secretome. BMC Genomics, 13, 380 (2012). Abstract
  9. Lee, D.Z., et al., Reactive oxygen species (ROS) modulate AMPA receptor phosphorylation and cell-surface localization in concert with pain-related behavior. Pain, 153, 1905-1915, (2012). Abstract
  10. Mauracher, S.G., et al., High level protein-purification allows the unambiguous polypeptide determination of latent isoform PPO4 of mushroom tyrosinase. Phytochemistry, 99, 14-25, (2014). Abstract