纯化 & 检测
HIS-Select® 技术
A Highly Selective Chemistry for HIS-tagged Protein Purification
Sigma's HIS-Select products have the ability to purify histidine-tagged proteins quickly and with high selectivity. This is due to HIS-Select's patented, non-charged, hydrophilic nickel chelate linkage. This non-charged linkage greatly diminishes non-specific binding. Most other systems for histidine-tagged protein purification utilize a charged chelate linkage. The tetradentate chelator used in the HIS-Select products results in greatly reduced nickel leaching and in high specificity for purification or affinity capture of your histidine-tagged protein. In addition, HIS-Select eliminates the need for costly and time-consuming secondary purification of your protein, allowing for One-step-purification.
阅读更多..
HIS-tagged proteins
  • High Selectivity for Higher Purity
  • Non-charged, hydrophilic linkage reduces non-specific binding
  • Pure tetradentate chelate for minimal metal leaching
  • One-step purification
What`s New !
Resins
Convenient Formats
Plates
  • Nickel Affinity Gel
  • High Flow Nickel Affinity Gel
  • HIS-Select Cobalt Affinity Gel
  • IMAC-Select Affinity Gel
  • HIS-Select iLAP® 5 ml Column
  • Cartridge
  • Spin Column
  • EZview™ Red Nickel Affinity Gel
  • High Capacity Nickel Coated Plates
  • High Sensitivity Nickel Coated Plates
  • HIS-Select Filter Plates
Kits
  • Trypsin
  • Maldi Kits
  • CelLytics
  • ProteoQwest Kits
  • ProteoSilver
  • HIS-Select Wash & Elution Buffers
Technical Publications & Literature
Recent R&D Posters
Automation Protocols
Additional Literature
Use the "Login" button on the tool bar (at the top of the page) to be able to view package sizes and prices when you click on the product numbers.
返回 纯化 & 检测页面
HIS-Select and products are covered by US Patent No 6,623,655 and are sold for research use only. Commercial use of Sigma's HIS-Select, PHOS-Select, iLAP and IMAC products require additional licenses.