树枝状大分子
树枝状大分子是一种具有纳米级尺寸、高度支化的星形大分子。树枝状大分子这一定义包括三个部分:中心核,内部树枝状结构(分支)和具有功能表面基团的外表面。这些组成的不同组合会产生不同形状和尺寸的产品,同时将内部核保护起来,是生物和材料科学应用的理想选择。尽管附着的表面基团会影响溶解度和螯合能力,但各种核可以实现各种独特的性质,包括不同空腔尺寸,吸附能力和负载-释放特性。最近文献中关注的应用包括药物递送,基因转染,催化,能量收集,光活性,分子量和尺寸测定,流变学修饰和纳米尺度科学与技术1,2。单分散树枝状大分子可以通过逐步化学法合成,以得到具有窄分子量分布,均一大小和形状,以及多种(多价)表面基团Z分子的不同代产品(G 0,G 1,G 2,...)。树枝化基元指的是具有单一焦点活性功能的单分散树枝状大分子楔形部分。超支化聚合物指的是通过较低成本的聚合方法所合成的多分散树枝状大分子。
(1) Functional Polymers and Dendrimers: From Synthesis to Application.
Proceedings of the American Chemical Society Division of Polymeric Materials: Science & Engineering, San Diego, CA, April 1-5, 2001; ACS, 2001
(2) Dendrimers and Other Dendritic Polymers, Jean M.J. Frechet and Donald A. Tomalia, J. Wiley & Sons (2001)