4-甲基伞型酮-β-D-葡糖苷酸水合物
货号 M9130
CAS RN 6160-80-1
别名: MUG
产品描述
来自大肠杆菌的β-葡萄糖醛酸糖苷酶(GUS)已成为转基因植物研究的首选报告酶。4-甲基伞型酮®-D-葡糖苷酸(MUG)是检测植物中GUS基因表达的常用底物。2 由于植物中不含有GUS基因编码的酶,因此,该系统在鉴定转基因植物方面非常实用。MUG具有很弱的荧光或无荧光。但是,当使用β-葡萄糖醛酸糖苷酶处理时,产生的4-甲基伞型酮产物具有荧光。在pH 10.3 的环境中,其荧光强度约为pH 7.0(0.15 M甘氨酸缓冲液)的100倍。1 MUG的这种荧光性质使其成为非常有效的GUS底物。3,4
MUG还可用于鉴定饮用水的大肠杆菌污染以及快速鉴定血液培养物的细菌污染。6
注意事项和免责声明
本品仅供研发使用,不可用于药物、家庭或其他用途。有关危险和安全操作信息,请参阅安全数据表。
制备说明
将本品以0.35 mg/ml的浓度溶于水。亦可以0.1 mM的浓度溶于0.1 M Sorensen缓冲液。6
储存/稳定性
将100mg MUG溶于100 ml H2O中并加入2滴TRITON® X-100,经过无菌过滤后,可在冰箱中稳定保存6个月。7
产品概况
分子式:C16H16O9
分子量:352.2(无水)
熔点:97-100 °C
比旋光度:-105°(0.25% (w/v),溶剂为水)
最大激发波长:365 nm1
最大发射波长:445 nm1
材料
参考文献
  1. J. Org. Chem., 27, 1074 (1962).
  2. Plant Mol. Biol. Rep., 5, 387-405 (1987).
  3. Biochem. J., 61, 569 (1955).
  4. Brot, F.E., et al., Purification and properties of beta-glucuronidase from human placenta. Biochemistry, 17, 385-391 (1978).
  5. Edberg, S.C., et al., National field evaluation of a defined substrate method for the simultaneous detection of total coliforms and Escherichia coli from drinking water: comparison with presenceabsence techniques. Appl. Environ. Microbiol., 55, 1003-1008 (1989).
  6. Sepulveda, J.L.et al., Rapid presumptive identification of gram-negative rods directly from blood cultures by simple enzymatic tests. J. Clin. Microbiol., 28, 177-181 (1990).
  7. Thompson, J.S., et al., Rapid biochemical test to identify verocytotoxin-positive strains of Escherichia coli serotype O157., J. Clin. Microbiol., 28, 2165-2168 (1990).
  8. Feng, P.C. and Hartman, P.A., Fluorogenic assays for immediate confirmation of Escherichia coli. Appl. Environ. Microbiol., 43, 1320-1329 (1982).
TRITON 是Union Carbide的注册商标。