脂多糖
糖生物学分析手册,第2版
脂多糖(LPS)是革兰氏阴性细菌外膜的主要成分。脂多糖位于胞膜的外层,在非包封菌株中暴露在细胞表面上。
 脂多糖的结构
结构体完整的细菌脂多糖是分子量为10-20 kDa的大分子,由三个结构组分组成(参见图1):
  • 疏水脂质部分,脂质A,造成分子毒性,
  • 亲水核心多糖链,
  • 重复的亲水性O-抗原性寡糖侧链,对细菌血清型具有特异性1
  • 细菌脂多糖的一般结构
图1. 细菌脂多糖的一般结构。有关每个部分的详细说明,请参阅文本。缩写:KDO:3脱氧-α-D-甘露辛酮糖酸;Hep:庚糖(庚酮糖);NGa:半乳糖胺;NGc:氨基葡萄糖。
脂质A核心由β-葡糖胺-(1→6)-葡糖胺-1-磷酸碱基构成,其中脂肪酸酯附着在两种碳水化合物上。酰基链长度和酰基数可以随细菌种类的不同而不同,但在同一物种内变化相对较小。内多糖核心通常含有1至4个与二糖核心连接的KDO(3脱氧-α-D-甘露辛酮糖酸)分子。KDO与脂多糖特异性结合,并且具有生物活性的脂质A过去被认为需要至少一个KDO残基才能使细菌存活。2 然而,缺乏KDO的大肠杆菌K-12抑制剂菌株证明,是否有KDO对于细菌的存活并不是绝对的。3  
含有KDO的内核也用庚糖(庚酮糖)单糖修饰,其中最常见的是L-丙三基-α-D-甘露-吡喃庚糖。内核聚糖残基通常用含磷酸基团磷酸化或修饰,例如焦磷酸盐或2-氨基乙基磷酸盐。脂多糖的磷酸基团增加细胞膜的总负电荷,并有助于稳定结构。 
脂多糖的外核含有更常见的己糖,包括葡萄糖、半乳糖和N-乙酰葡糖胺,并且在结构上比内核更多样化。 
O-抗原是重复的寡糖单元,通常由2-6个糖组成。O-抗原是脂多糖的主要结构成分,基于它细菌被区分开来。已经使用独特的O-抗原结构来鉴定和分类大肠杆菌、肠沙门氏菌和霍乱弧菌的血清群。4 来自大肠杆菌的粗糙突变菌株的脂多糖缺乏该结构的O-抗原部分。 
脂多糖的核心部分和脂质A部分可能具有一些结构可变性,而O-抗原具有高度的结构可变性以及重复单元数量的可变性。这些差异导致LPS制剂的显著异质。由于LPS是异质的,并且倾向于形成不同大小的聚集体,有报道称这些聚集体的“分子量”范围为1-4百万道尔顿或更高。当用十二烷基硫酸钠(SDS)和加热处理LPS时,分子量为~50-100 kDa。5 
 脂多糖的功能和应用
在革兰氏阴性菌中,胞膜脂多糖可保护细菌免受胆汁盐和亲脂性抗生素的作用6
脂多糖是热稳定的内毒素,并且长期以来被认为是人类感染性休克(败血症)的关键因素1,7 更一般而言,它是在正常哺乳动物细胞中诱导强烈免疫应答的关键因素。已经鉴定脂质A基团对脂多糖的内毒素活性是关键的。通过在合成和天然来源的大肠杆菌脂质A制剂之间发现相同的生物活性结果(包括内毒素活性),Galanos等证明了点。8 脂多糖的活性受体已被鉴定为CD14 / TLR4 / MD2受体复合物,它促进促炎细胞因子的分泌,包括肿瘤坏死因子-α和白细胞介素-1。9 虽然脂质A成分主要负责免疫反应激活,但肠道沙门氏菌LPS的多糖成分也是NF-κB活化所必需的。10
脂多糖制剂已被用于研究LPS结构、11 代谢、12 免疫学、13 生理学、14 毒性、15 和生物合成。16 它们还被用于诱导白细胞介素等生长促进因子的合成和分泌。17,18由于脂多糖与败血症的关系,已经对脂多糖进行了研究,以确定抗体和LPS生物合成抑制剂的可能靶标。19,20
 脂多糖的提取和纯化
脂多糖可以通过从TCA、21苯酚、22,23或苯酚-氯仿-石油醚(对粗糙菌株)提取来制备。24 TCA提取的脂多糖在结构上类似于苯酚提取的脂多糖,具有相似的电泳图案和内毒性。主要差异在于提取后残留的核酸和蛋白质污染物的量。TCA提取物含有~2%的RNA和~10%的变性蛋白质,而苯酚提取物含有高达60%的RNA和<1%的蛋白质。随后通过凝胶过滤色谱法纯化除去苯酚提取的LPS中存在的大量蛋白质,但产生含有10-20%核酸的制剂。使用离子交换色谱进一步纯化产生含有<1%蛋白质和<1% RNA的脂多糖产物。
 材料
 参考文献
E T Rietschel, T Kirikae, F U Schade, U Mamat, G Schmidt, H Loppnow, A J Ulmer, U Zähringer, U Seydel, F Di Padova
FASEB Journal 1994-02-01
Endotoxins of Gram-negative microbes fulfill as components of the outer membrane a vital function for bacterial viability and, if set free, induce in mammalians potent pathophysiological effects. Chemically, they are lipopolysaccharides (LPS) consisting of an O-specific chain, a core oligosaccharide, and a lipid component, terme...阅读更多
I M Helander, B Lindner, H Brade, K Altmann, A A Lindberg, E T Rietschel, U Zähringer
European Journal of Biochemistry 1988-11-15
The chemical structure of the lipopolysaccharide of a deep-rough mutant (strain I-69 Rd-/b+) of Haemophilus influenzae was investigated. The hydrophilic backbone of lipid A was shown to consist of a beta-(1',6)-linked D-glucosamine disaccharide with phosphate groups at C-1 of the reducing D-glucosamine and at C-4' of the non-red...阅读更多
Timothy C Meredith, Parag Aggarwal, Uwe Mamat, Buko Lindner, Ronald W Woodard
ACS Chemical Biology 2006-02-17
Gram-negative bacteria possess an asymmetric lipid bilayer surrounding the cell wall, the outer membrane (OM). The OM inner leaflet is primarily composed of various glycerophospholipids, whereas the outer leaflet predominantly contains the unique amphiphilic macromolecule, lipopolysaccharide (LPS or endotoxin). The majority of a...阅读更多
Gabrielle Samuel, Peter Reeves
Carbohydrate Research 2003-11-14
The O-antigen is an important component of the outer membrane of Gram-negative bacteria. It is a repeat unit polysaccharide and consists of a number of repeats of an oligosaccharide, the O-unit, which generally has between two and six sugar residues. O-Antigens are extremely variable, the variation lying in the nature, order and...阅读更多
B Jann, K Reske, K Jann
European Journal of Biochemistry 1975-12-01
Lipopolysaccharide preparations from R(rough) Escherichia coli O8-,SR(semirough) Salmonella typhimurium and S (smooth) strains E. coli O8 and Citrobacter 396 were disintegrated with sodium dodecylsulfate and subjected to polyacrylamide gel electrophoresis in the presence of 1% sodium dodecylsulfate. The results obtained were com...阅读更多
6. Analysis of lipopolysaccharides of Gram-negative bacteria. Mayer, H. et al. Methods Microbiol., 18, 157-207 (1985).
J Schletter, H Heine, A J Ulmer, E T Rietschel
Archives of Microbiology 1995-12-01
Endotoxin (lipopolysaccharide, LPS), a constituent of the outer membrane of the cell wall of gram-negative bacteria, exerts a wide variety of biological effects in humans. This review focuses on the molecular mechanisms underlying these activities and discusses structure-function relationships of the endotoxin molecule, its inte...阅读更多
C Galanos, O Lüderitz, E T Rietschel, O Westphal, H Brade, L Brade, M Freudenberg, U Schade, M Imoto, H Yoshimura
European Journal of Biochemistry 1985-04-01
The recently chemically synthesized Escherichia coli lipid A and the natural free lipid A of E. coli were compared with respect to their endotoxic activities in the following test systems: lethal toxicity, pyrogenicity, local Shwartzman reactivity, Limulus amoebocyte lysate gelation capacity, tumour necrotizing activity, B cell ...阅读更多
Eva M Pålsson-McDermott, Luke A J O'Neill
Immunology 2004-10-01
An understanding of lipopolysaccharide (LPS) signal transduction is a key goal in the effort to provide a molecular basis for the lethal effect of LPS during septic shock and point the way to novel therapies. Rapid progress in this field during the last 6 years has resulted in the discovery of not only the receptor for LPS - Tol...阅读更多
Masashi Muroi, Ken-Ichi Tanamoto
Infection and Immunity 2002-11-01
The lipid A portion has been identified as the active center responsible for lipopolysaccharide (LPS)-induced macrophage activation. However, we found that Salmonella (Salmonella enterica serovars Abortusequi, Minnesota, and Typhimurium) lipid A is inactive in human macrophages, despite its LPS being highly active. Thus we inves...阅读更多
S M Strain, S W Fesik, I M Armitage
Journal of Biological Chemistry 1983-03-10
Lipopolysaccharide (LPS) isolated from Escherichia coli D31m4, a heptoseless mutant, was studied by 13C and 31P NMR spectroscopy. Modified isolation and purification procedures are described which permitted high resolution NMR spectra to be obtained from samples of intact LPS. 31P NMR was used to monitor the purity and native he...阅读更多
R S Munford, J M Andersen, J M Dietschy
Journal of Clinical Investigation 1981-12-01
When gram-negative bacterial lipopolysaccharides (LPS) are injected intravenously into the rabbit or rat, they bind to plasma lipoproteins, particularly high density lipoproteins (HDL). The present studies were performed to examine the mechanisms by which LPS-HDL complexes are removed from the circulation and taken up by various...阅读更多
D C Morrison, J A Rudbach
Contemporary topics in molecular immunology 1981-01-01
14. International Review of Biochemistry. Galanos, C., et al. Biochemistry of Lipids III, Vol. 14, 2309, T. E. Goodwin, ed., University Park Press, Baltimore, MD USA (1977).
H J Kurtz, J Quast
American Journal of Veterinary Research 1982-02-01
Escherichia coli endotoxin was continuously infused IV into 31 pigs. The response was related to the infusion rate. Of 13 pigs given endotoxin at the rate of 15.4 and 23.1 micrograms/kg of body weight/hour, 10 had bilateral renal cortical necrosis similar to a generalized Shwartzman lesion. All 13 pigs given endotoxin at the rat...阅读更多
P D Rick, D A Young
Journal of Bacteriology 1982-05-01
A new mutant of Salmonella typhimurium was isolated which possesses a temperature-sensitive defect in the synthesis of 3-deoxy-D-manno-octulosonic acid. The defect in 3-deoxy-D-manno-octulosonic acid synthesis is due to a temperature-sensitive 3-deoxy-D-manno-octulosonate-8-phosphate synthetase, and the mutant accumulates an inc...阅读更多
R R Skelly, P Munkenbeck, D C Morrison
Infection and Immunity 1979-02-01
The murine immune response to a haptenated lipopolysaccharide (LPS) lacking repeating oligosaccharide determinants was studied. The LPS was extracted from a rough strain of bacteria (Salmonella minnesota R595) and chemically haptenated with either trinitrophenol or fluorescein isothiocyanate. These preparations of hapten-R595 LP...阅读更多
Charles H Lang, Christine Silvis, Nobuko Deshpande, Gerald Nystrom, Robert A Frost
Shock 2003-06-01
The presence of increased levels of proinflammatory cytokines in the blood is associated with decreased muscle protein synthesis and the erosion of lean body mass in many catabolic conditions. However, little is known regarding the role of endogenous cytokine synthesis in muscle per se. The purpose of the present study was to ch...阅读更多
J A Yethon, C Whitfield
Current Drug Targets: Infectious Disorders 2001-08-01
Lipopolysaccharide (LPS) constitutes the lipid portion of the outer leaflet of Gram-negative bacteria, and is essential for growth. LPS is also known to be responsible for the variety of biological effects associated with Gram-negative sepsis. In recent years, tremendous progress has been made in determining the exact chemical s...阅读更多
Sven Muller-Loennies, Lore Brade, C Roger MacKenzie, Franco E Di Padova, Helmut Brade
Journal of Biological Chemistry 2003-07-11
Septic shock due to infections with Gram-negative bacteria is a severe disease with a high mortality rate. We report the identification of the antigenic determinants of an epitope that is present in enterobacterial lipopolysaccharide (LPS) and recognized by a cross-reactive monoclonal antibody (mAb WN1 222-5) regarded as a poten...阅读更多
21. Bacterial lipido-protinopolysaccharides ('O' somatic antigens) extraction with trichloroacetic acid. Staub, A. M. Methods in Carbohydrate Chem., Vol. 5, 92-93, John Wiley & Sons, Inc., Hoboken, NJ USA (1965).
22. Bacterial lipopolysaccharides extraction with phenolwater and further applications of the procedure. Westphal, O. and Jann, K. Methods in Carbohydrate Chem., Vol. 5, 92‑93, John Wiley & Sons, Inc., Hoboken, NJ USA (1965).
23. Isolation of lipopolysaccharides from bacteria. Leive, L. and Morrison, D. C. Methods Enzymol., 28, 254-262 (1972).
C Galanos, O Lüderitz, O Westphal
European Journal of Biochemistry 1969-06-01