用于分子生物学的醋酸钠缓冲溶液
 产品编号:S7899
CAS RN 126-96-5
 产品描述
该产品被定为分子生物学级。它适用于DNA沉淀,已经经过了核酸酶存在分析测定。分析证书提供了相应批次的产品浓度和pH值。
醋酸钠是分子生物学应用中广泛使用的一种试剂。它与醋酸一起用作缓冲剂,缓冲范围为pH 3.6 - 5.6。醋酸钠用于核酸的纯化和沉淀1,2,3,蛋白质结晶4,蛋白质凝胶电泳中的凝胶染色5,以及HPLC6。醋酸钠的大规模应用包括其在塑料生产中作为阻燃剂,在印染和制革中用作媒染剂7
大肠杆菌对不同水平的醋酸钠的反应的DNA微阵列研究已有报道8。在不同离子强度、不同盐(包括醋酸钠)的条件下,反相色谱中的蛋白质去折叠的情况已被研究9。醋酸钠与碳酸钠结合使用,用于增强冻干的枯草杆菌蛋白酶Carlsberg在有机溶剂中的活化10。醋酸钠可用作醋酸激酶的底物11
 储存
室温
 组分
该产品是3 M的溶液。可用18兆欧的水制备,用0.2 μm过滤器过滤。该产品在25℃下的pH为5.2 ± 0.1。
 注意事项及免责声明
本产品仅供研发使用,不适用于药物、家庭或其他用途。有关有害物及其安全处理方法的信息,请参阅安全数据表。
 材料
 参考文献
  1. Evans, J. K., et al., Simultaneous purification of RNA and DNA from liver using sodium acetate precipitation. BioTechniques, 24, 416-418 (1998).
  2. Molecular Cloning: A Laboratory Manual, 3rd ed., Sambrook, J. F., et al., Cold Spring Harbor Laboratory Press (Cold Spring Harbor, NY: 2001), pp. 6.26-6.27, A8.12-A8.16.
  3. Wallace, D. M., Large- and Small-Scale Phenol Extractions, Meth. Enzymol., 152, 33-41 (1987).
  4. Baniecki, M. L., et al., Adenovirus proteinase: crystallization and preliminary X-ray diffraction studies to atomic resolution. Acta Crystallogr. D Biol. Crystallogr., 58 (Pt 9), 1462-1464 (2002).
  5. Bjellqvist, B., et al., A nonlinear wide-range immobilized pH gradient for two-dimensional electrophoresis and its definition in a relevant pH scale. Electrophoresis, 14, 1357-1365 (1993).
  6. Clark, T. N., et al., Determination of 3'-azido-2',3'-dideoxyuridine in maternal plasma, amniotic fluid, fetal and placental tissues by high-performance liquid chromatography. J. Chromatogr. B Biomed. Sci. Appl., 755(1-2), 165-172 (2001).
  7. The Merck Index, 12th ed., Entry# 8711.
  8. Polen, T., et al., DNA microarray analyses of the long-term adaptive response of Escherichia coli to acetate and propionate. Appl. Environ. Microbiol., 69(3), 1759-1774 (2003).
  9. McNay, J. L., et al., Protein unfolding duringreversed-phase chromatography: II. Role of salt type and ionic strength. Biotechnol. Bioeng., 76(3), 233-240 (2001).
  10. Ru, M. T., et al., Towards more active biocatalysts in organic media: increasing the activity of saltactivated enzymes. Biotechnol. Bioeng., 75(2), 187-196 (2001).
  11. 1Rose, I., Acetate Kinase of Bacteria (Acetokinase), Meth. Enzymol., 1, 591-595 (1955)