Salen配体
By: William Sommer and Daniel WeibelAldrich ChemFiles 2008, 8.2, 71.
Salen分子已被研究超过六十年。然而,Jacobsen和Katsuki在1990年单独发表了关于Salen用作不对称环氧化反应锰配体的第一篇报道。1 自从第一篇报道开始,金属-Salen伦催化领域已得到了极大的扩展。大量金属-Salen络合物已被合成用于各种催化的不对称转化。2 例如,铬和钴的Salen络合物可催化具有高水平对映选择性非官能化烯烃的环氧化。3 铝的Salen络合物可催化叠氮化物与不饱和酰亚胺的共轭加合。4 Salen配体骨架的简易合成和修饰使其成为发现新型催化剂和反应的首选平台.
Salen络合物的多功能性举例请见方案1
方案1
方案1
 材料
参考文献
  1. (a) Zhang, W. et al. J. Am. Chem. Soc. 1990112, 2801. (b) Jacobsen, E. N. et al. J. Am. Chem. Soc. 1991113, 6703. (c) Irie, R. et al. Tetrahedron Lett. 199031, 7345.
  2. (a) Jacobsen, E. N. In Comprehensive Organometallic Chemistry II; Abel, E. W.; Stone, F. G. A.; Wilkinson, G., Eds.; Elsevier Science: Oxford, 1995; Vol. 12, p 1097. (b) Canali, L.; Sherrington, D. C. Chem. Soc. Rev. 199928, 85. (c) McGarrigle, E. M.; Gilheany, D. G. Chem. Rev. 2005105, 1563.
  3. (a) Jacobsen, E. N. et al. J. Am. Chem. Soc. 1991113, 7063. (b) Jacobsen, E. N. Acc. Chem. Res200033, 421.
  4. Meyers, J.K.; Jacobsen, E. N. J. Am. Chem. Soc. 1999121, 8959.