Jerome Waser研究小组 - 教授产品门户
教授产品站索引
Jérôme Waser教授
Waser实验室正在利用超价碘试剂的独特性质来开发非传统键断开技术。特别引人注意的是,他们发现了1 - [(三异丙基甲硅烷基)乙炔基] -1,2-苯并碘杂环戊烯-3(1H) - 酮(TIPS-EBX)用于亲电子炔基化的特殊性质。该试剂由Zhdankin及其合作者在1996年首次合成(J.Org.Chem.1996,61,657)其在合成中的应用潜力一直未得到完全发掘,直到Waser研究小组报道了将其用于金催化的富电子直接炔基化化合物才开始引人注意,比如吲哚、噻吩或苯胺。TIPS-EBX也是烯烃钯催化氧化和氨基炔基化的首选试剂。最后,该试剂也可用于酮酯和硫醇的直接炔基化或自由基脱羧炔基化。TIPS-EBX是一种长效稳定的结晶化合物,因此特别方便用户使用。Waser实验室目前正在继续开发用于转移乙炔和其他官能团的超价碘试剂。
Waser研究小组网站
有关适用的产品信息,请点击以下链接:
Heller实验室最近发表的文章:
Reto Frei, Jérôme Waser
Journal of the American Chemical Society, 135(26), 9620-9623 (2013)
A thiol-alkynylation procedure utilizing the hypervalent iodine alkyne transfer reagent TIPS-ethynyl-benziodoxolone has been developed. This scalable reaction proceeds in five minutes at room temperature in an open flask using commercially available...Read More
Stefano Nicolai et. al
Organic Letters, 12(2), 384-387 (2009)
The first example of intramolecular oxyalkynylation of nonactivated alkenes using oxidative Pd chemistry is reported. Both phenol and aromatic or aliphatic acid derivatives could be used under operator-friendly conditions (room tempe...Read More
Jonathan P Brand et. al
Angewandte Chemie (International Edition), 48(49), 9346-9349 (2009)