Tehshik Yoon研究小组 - 教授产品门户
教授产品站索引
       Tehshik Yoon教授
Yoon实验室研究的一个主要课题是通过可见光光化学来开发独特的化学转化反应。Yoon实验室已获得了一系列对可见光有响应的过渡金属光催化剂,还发现已知的光化学可以与环加成反应、芳烃氧化反应、胺官能化反应和硫醇-烯反应等进行偶合。该策略的一个重要特征是能够使用合理的配体修饰来调整催化剂的光物理和电化学性质,例如下面的配合物。
Yoon研究小组网站
有关适用的产品信息,请点击以下链接:
Yoon实验室最近发表的文章:
Zhan Lu, Tehshik P Yoon
Angewandte Chemie. International edition in English 2012-10-08
Shishi Lin, Michael A Ischay, Charles G Fry, Tehshik P Yoon
Journal of the American Chemical Society 2011-12-07
Ruthenium(II) polypyridyl complexes promote the efficient radical cation Diels-Alder cycloaddition of electron-rich dienophiles upon irradiation with visible light. These reactions enable facile [4 + 2] cycloadditions that would be electronically mismatched under thermal conditions. Key to the success of this methodology is the ...Read More
Michael A Ischay, Mary E Anzovino, Juana Du, Tehshik P Yoon
Journal of the American Chemical Society 2008-10-01
We report that Ru(bipy)3Cl2 can serve as a visible light photocatalyst for [2+2] enone cycloadditions. A variety of aryl enones participate readily in the reaction, and the diastereoselectivity in the formation of the cyclobutane products is excellent. We propose a mechanism in which a photogenerated Ru(bipy)3+ complex promotes ...Read More
S K Aruo
Bulletin of Animal Health and Production in Africa 1978-09-01