Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Rotor Guide - Centrifree®
  • 16x95 mm
  • Maximum g-force 2000 x g
  • Fixed-angle rotor
Learn more about Centrifree® centrifugal filter
Note: Centrifuges and rotors listed are not available from. Please confirm product availabilty with the manufacturer.
Beckman
Centrifuge Model Rotor Adapter Required
Allegra X12 FX6100  
back to top
Hermle
Centrifuge Model Rotor Adapter Required
C0200-A C0200-96  
back to top
Hettich    
Centrifuge Model Rotor Adapter Required
EBA20 Included  
EBA21 1416  
MIKRO22R 1015  
ROTANTA460/460R 5615 1451
ROTINA35/35R 1792 1451
ROTOFIX32 1613  
UNIVERSAL32/32R 1613  
UNIVERSAL32/32R 1615  
back to top
Kendro
Centrifuge Model Rotor Adapter Required
Biofuge Primo/Primo R 75007588 76002906
Biofuge Stratos 75003057 76002906
Biofuge Stratos 75003336  
Biofuge Stratos 75003335 76002904
Biofuge Stratos 750003334 76002906
Legend Mach 1.6, Multifuge 1/1R 75002006 76002906
Legend Mach 1.6, Multifuge 1/1R 75002001 75002021
Legend Mach 1.6, Multifuge 1/1R 75002002 75007572
Legend T/RT, Multifuge 3/3R 75003057 76002906
Legend T/RT, Multifuge 3/3R 75006441 75006494
Legend T/RT, Multifuge 3/3R 75006446 75006492
back to top