Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a 7-Component Sulfur Gases Mix on Supel-Q™ PLOT after SPME using 75 μm Carboxen/PDMS Fiber
GC Analysis of a 7-Component Sulfur Gases Mix on Supel-Q™ PLOT after SPME using 75 μm Carboxen/PDMS Fiber
Conditions
sample/matrix sulfur gases in 40 mL vial
desorption process 3 min at 290 °C
extraction 10 min headspace at 40 °C
SPME fiber Carboxen/PDMS, 75 µm (57318)
column Supel-Q PLOT, 30 m x 0.32 mm I.D. (24242)
oven 45 °C (1 min) to 250 °C at 15 °C/min
detector FPD
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Gases & Hydrocarbons, Volatiles, Sulfur Compounds, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Supel-Q is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC
Analyte
  • Carbon disulfide
  • Carbonyl sulfide
  • Dimethyl disulfide
  • Ethanethiol
  • Hydrogen sulfide
  • Methanethiol
  • Sulfur dioxide