Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a QC Test Mix on SLB®-IL76
GC Analysis of a QC Test Mix on SLB®-IL76
Conditions
column SLB-IL76, 30 m x 0.25 mm I.D., 0.20 µm (28913-U)
oven 130 °C
carrier gas helium, 1.2 mL/min (28 cm/sec)
sample polar column test mix (47302), 9 analytes, each at 500 µg/mL in methylene chlorid
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Featured Industry General Analytical