Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN
GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN
Conditions
column Astec CHIRALDEX G-PN, 30 m x 0.25 mm I.D., 0.12 µm (74033AST)
oven 130 °C
carrier gas helium, 30 psi
sample peak 1: l-amphetamine (N-TFA)peak 2: d-amphetamine (N-TFA)
liner 4 mm I.D., double taper
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Achieve elution order reversal (d,l) by using CHIRALDEX G-DP column, 140 °C (see G005151).
Categories Analytical Chromatography, Chiral Applications, GC Applications, Clinical, Forensic & Toxicology, Enantiomers, Pharmaceuticals, Drugs of Abuse, Amines
Featured Industry Clinical
Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes