Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Isothermal Analysis
GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Isothermal Analysis
Conditions
column SLB-IL111i, 60 m x 0.25 mm I.D., 0.20 µm (29884-U)
oven 50 °C (20 min)
carrier gas helium, 21 cm/sec
sample 8 analytes at various concentrations in isooctane
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design (2879925-U)
inj. temp. 250 °C
detector FID, 200 °C
Description
Analysis Note The inert SLB IL111i ionic liquic column produces great peak shapes for alcohols via an isothermal analysis, providing resolution in a mix of aromatics. This is impressive for such a polar column.
Featured Industry Chemicals and Industrial Polymers
Petroleum