Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Bleed Profiles of GC Septa on SLB®-5ms
GC Analysis of Bleed Profiles of GC Septa on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 40 °C (3 min), 15 °C/min to 325 °C (15 min)
carrier gas helium, 25 cm/sec constant
sample solvent extracts of septa, dodecane (as an internal standard) added to 100 µg/mL
injection 1 µLsplitless (1 min)
liner 4 mm I.D., single taper, unpacked
inj. temp. 250 °C
detector FID, 325 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry General Analytical
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes