Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of C1-C12 Primary Alcohols on SLB®-IL76i
GC Analysis of C1-C12 Primary Alcohols on SLB®-IL76i
Conditions
column SLB-IL76i, 30 m x 0.25 mm I.D., 0.20 µm (29869-U)
oven 55 C (2 min), 4 °C/min to 200 °C
carrier gas helium, 1.2 mL/min (28 cm/sec)
sample 12 component primary alcohols mix, each analyte at 500 µg/mL in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note The i-series of ionic liquid capillary GC columns are characterized by their improved inertness for polar analytes. This application shows that the SLB IL76i can generate great peak shapes for a series of alcohols.
Featured Industry General Analytical