Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of FAMEs in Human Plasma on Equity®-1
GC Analysis of FAMEs in Human Plasma on Equity®-1
Conditions
column Equity-1, 30 m x 0.25 mm I.D., x 0.25 µm (28046-U)
column SUPELCOWAX 10, 0.95 m x 0.10 mm x 0.10 µm
oven 180 °C, 3 °C/min to 280 °C (10 min)
carrier gas hydrogen, 75.3 kPa, constant
detector FID
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Clinical
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes