Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH
Conditions
column Petrocol DH, 100m x 0.25 mm I.D., 0.5 µm film (24160-U)
column temp. 35 °C for 15 min, then to 200 °C at 2 °C/min, hold for 5 min
carrier gas helium, 20 cm/sec (set at 35 °C)
sample premium unleaded gasoline, split 100:1,
injection 1 µL
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Petrocol is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3-Trimethylbenzene
  • 1,2,4-Trimethylbenzene
  • 1,3,5-Trimethylbenzene
  • 1-Methyl-3-ethylbenzene
  • 1-Methylnaphthalene
  • 2,2,4-Trimethylpentane
  • 2,3,4-Trimethylpentane
  • 2,3-Dimethylbutane
  • 2,3-Dimethylhexane
  • 2,4-Dimethylhexane
  • 2,4-Dimethylpentane
  • 2,5-Dimethylhexane
  • 2-Methylbutane
  • 2-Methylhexane
  • 2-Methylnaphthalene (β)
  • 2-Methylpentane
  • 2-Methylpropane
  • 3-Methylhexane
  • 3-Methylpentane
  • Benzene
  • Butane
  • Dimethylnaphthalenes
  • Ethylbenzene
  • Hexane
  • Mesitylene
  • m-Xylene
  • Naphthalene
  • o-Xylene
  • Pentane
  • p-Xylene
  • Toluene