Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Lemon Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Lemon Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 °C, 50 °C/min to 320 °C
carrier gas hydrogen, 81.5 cm/sec constant
sample lemon essential oil in hexane
injection 0.4 µL in the split mode (300:1)
inj. temp. 320 °C
detector FID, 320 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (E)-Caryophyllene
  • (E)-Myroxide
  • (E)-β-Ocimene
  • (E)-γ-Bisabolene
  • (Z)-γ-Bisabolene
  • 1-Tetradecene
  • Borneol
  • Camphene
  • Campherenol
  • Camphor
  • Carvone
  • cis-Limonene oxide
  • cis-Sabinene hydrate
  • Citronellal
  • Citronellol
  • Citronellyl acetate
  • Decanal
  • Geranial
  • Geraniol
  • Geranyl acetate
  • Limonene
  • Linalool
  • Linalyl isobutanoate
  • Methyl geranoate
  • Myrcene
  • Neral
  • Nerol
  • Neryl acetate
  • Nonanal
  • Norbomanol
  • Octanal
  • Octanol
  • p-Cymene
  • Perilla aldehyde
  • Sabinene
  • Terpinen-4-ol
  • Terpinolene
  • Tetradecane
  • trans-Limonene oxide
  • trans-α-Bergamotene
  • Tricyclene
  • Undecanal
  • α-Bisabolol
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Thujene
  • β-Bisabolene
  • β-Pinene
  • γ-Terpinene
  • δ-3-Carene