Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Nitrogen and Nitrous Oxide on Carboxen®-1010 PLOT
GC Analysis of Nitrogen and Nitrous Oxide on Carboxen®-1010 PLOT
Conditions
column Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven 150 °C
carrier gas helium, 3.0 mL/min
sample 500 ppm nitrous oxide in nitrogen
injection 10 µL, direct using a sample valve, 150 °C
inj. temp. 200 °C
detector TCD, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes